Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 175881516-175882027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPLX2
Entrez Gene ID
10814
Gene Name
complexin 2
Gene Aliases
921-L, CPX-2, CPX2, Hfb1
UniGene
Hs.193235
Gene Chromosome Location
Chr.5: 175796539-175884021 on Build GRCh38
Transcripts
NM_001008220.1 NM_006650.3 XM_005265798.1 XM_005265799.1 XM_011534419.1 XM_017008964.1

SNP Detail

SNP ID Chromosome Location
rs971677009 Chr.5: 175881801-175881801 on Build GRCh38
rs941617125 Chr.5: 175881840-175881840 on Build GRCh38
rs377275465 Chr.5: 175881895-175881895 on Build GRCh38
rs890131170 Chr.5: 175881672-175881672 on Build GRCh38
rs950320904 Chr.5: 175881554-175881554 on Build GRCh38
rs554341701 Chr.5: 175881556-175881556 on Build GRCh38
rs1009920635 Chr.5: 175881931-175881931 on Build GRCh38
rs1034145820 Chr.5: 175881981-175881981 on Build GRCh38
rs1047313831 Chr.5: 175881577-175881577 on Build GRCh38
rs982990648 Chr.5: 175881561-175881561 on Build GRCh38
rs992914905 Chr.5: 175881768-175881768 on Build GRCh38
rs932698576 Chr.5: 175881922-175881922 on Build GRCh38
rs910154373 Chr.5: 175881823-175881823 on Build GRCh38
rs59258044 Chr.5: 175881596-175881596 on Build GRCh38
rs890390472 Chr.5: 175881629-175881629 on Build GRCh38
rs1051519893 Chr.5: 175881628-175881628 on Build GRCh38
rs958687184 Chr.5: 175881730-175881730 on Build GRCh38
rs1060820 Chr.5: 175881609-175881609 on Build GRCh38
rs571111554 Chr.5: 175881868-175881868 on Build GRCh38
rs982963383 Chr.5: 175881803-175881803 on Build GRCh38
rs754062147 Chr.5: 175881612-175881612 on Build GRCh38
rs765269244 Chr.5: 175881846-175881846 on Build GRCh38
rs572264984 Chr.5: 175881804-175881804 on Build GRCh38
rs186364978 Chr.5: 175881957-175881957 on Build GRCh38
rs377004998 Chr.5: 175881602-175881602 on Build GRCh38
rs921466597 Chr.5: 175881845-175881845 on Build GRCh38
rs1024368840 Chr.5: 175881769-175881769 on Build GRCh38
rs74801908 Chr.5: 175881704-175881704 on Build GRCh38
rs755921647 Chr.5: 175881958-175881958 on Build GRCh38
rs912513052 Chr.5: 175881942-175881943 on Build GRCh38
rs1034172139 Chr.5: 175881764-175881764 on Build GRCh38
rs555479440 Chr.5: 175881693-175881693 on Build GRCh38
rs146910572 Chr.5: 175881878-175881878 on Build GRCh38
rs189818293 Chr.5: 175881686-175881686 on Build GRCh38
rs900234500 Chr.5: 175881632-175881632 on Build GRCh38
rs974191726 Chr.5: 175881874-175881874 on Build GRCh38
rs1010534069 Chr.5: 175881685-175881685 on Build GRCh38
rs755078358 Chr.5: 175881644-175881644 on Build GRCh38
rs1037433712 Chr.5: 175881627-175881627 on Build GRCh38
rs185458303 Chr.5: 175881637-175881637 on Build GRCh38
rs532708189 Chr.5: 175881837-175881837 on Build GRCh38
rs917537933 Chr.5: 175881543-175881544 on Build GRCh38
rs968497429 Chr.5: 175881688-175881688 on Build GRCh38
rs557749137 Chr.5: 175881613-175881613 on Build GRCh38
rs1041797138 Chr.5: 175881950-175881950 on Build GRCh38
rs878887863 Chr.5: 175881919-175881919 on Build GRCh38
rs1039883667 Chr.5: 175881842-175881842 on Build GRCh38
rs541683116 Chr.5: 175881741-175881741 on Build GRCh38
rs183458601 Chr.5: 175881765-175881765 on Build GRCh38
rs908679012 Chr.5: 175881573-175881573 on Build GRCh38
rs757220153 Chr.5: 175881902-175881902 on Build GRCh38
rs1030408282 Chr.5: 175881663-175881663 on Build GRCh38
rs931679884 Chr.5: 175881848-175881848 on Build GRCh38
rs917199896 Chr.5: 175881777-175881777 on Build GRCh38
rs951423659 Chr.5: 175881798-175881798 on Build GRCh38
rs780902141 Chr.5: 175881936-175881936 on Build GRCh38

View Full Product Details