Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176294616-176295146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIMC1
Entrez Gene ID
375484
Gene Name
SUMO interacting motifs containing 1
Gene Aliases
C5orf25, OOMA1, PLEIAD
UniGene
Hs.719847
Gene Chromosome Location
Chr.5: 176238359-176345991 on Build GRCh38
Transcripts
NM_001308195.1 NM_001308196.1 NM_198567.5 NR_131772.1 XM_011534553.2 XM_011534554.2 XM_011534556.2 XM_017009454.1 XM_017009455.1 XM_017009456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6778564 c.1466G>A p.R489Q Chr.5: 176295064-176295064 on Build GRCh38
5091195 c.1500G>T p.E500D Chr.5: 176295098-176295098 on Build GRCh38
4600757 c.1412G>A p.R471K Chr.5: 176295067-176295067 on Build GRCh38
4469241 c.1460C>T p.P487L Chr.5: 176295058-176295058 on Build GRCh38
6778563 c.1409G>A p.R470Q Chr.5: 176295064-176295064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746485306 Chr.5: 176295117-176295117 on Build GRCh38
rs774202988 Chr.5: 176294965-176294965 on Build GRCh38
rs377387901 Chr.5: 176295120-176295120 on Build GRCh38
rs368082308 Chr.5: 176294640-176294640 on Build GRCh38
rs575545659 Chr.5: 176294643-176294643 on Build GRCh38
rs974052794 Chr.5: 176294688-176294688 on Build GRCh38
rs746254659 Chr.5: 176295111-176295111 on Build GRCh38
rs1023597440 Chr.5: 176294985-176294985 on Build GRCh38
rs771648970 Chr.5: 176294965-176294966 on Build GRCh38
rs770221381 Chr.5: 176294963-176294964 on Build GRCh38
rs747614908 Chr.5: 176294965-176294967 on Build GRCh38
rs753670845 Chr.5: 176294987-176294987 on Build GRCh38
rs1003623703 Chr.5: 176294991-176294991 on Build GRCh38
rs377627808 Chr.5: 176295037-176295037 on Build GRCh38
rs188640308 Chr.5: 176294982-176294982 on Build GRCh38
rs774716690 Chr.5: 176295019-176295019 on Build GRCh38
rs768329663 Chr.5: 176295119-176295119 on Build GRCh38
rs557322360 Chr.5: 176294786-176294786 on Build GRCh38
rs778400991 Chr.5: 176294726-176294726 on Build GRCh38
rs765075943 Chr.5: 176295066-176295066 on Build GRCh38
rs779076461 Chr.5: 176295107-176295107 on Build GRCh38
rs748555225 Chr.5: 176294968-176294968 on Build GRCh38
rs536537961 Chr.5: 176295056-176295056 on Build GRCh38
rs139994811 Chr.5: 176295070-176295070 on Build GRCh38
rs778225505 Chr.5: 176295098-176295098 on Build GRCh38
rs190491418 Chr.5: 176294734-176294734 on Build GRCh38
rs369729568 Chr.5: 176295054-176295054 on Build GRCh38
rs766884758 Chr.5: 176294980-176294980 on Build GRCh38
rs551747354 Chr.5: 176294900-176294900 on Build GRCh38
rs772834830 Chr.5: 176294964-176294965 on Build GRCh38
rs771328639 Chr.5: 176295014-176295014 on Build GRCh38
rs778313332 Chr.5: 176295013-176295013 on Build GRCh38
rs764137765 Chr.5: 176294983-176294983 on Build GRCh38
rs755091219 Chr.5: 176295072-176295072 on Build GRCh38
rs577281911 Chr.5: 176294722-176294722 on Build GRCh38
rs781491969 Chr.5: 176295075-176295075 on Build GRCh38
rs560915384 Chr.5: 176294838-176294838 on Build GRCh38
rs549305036 Chr.5: 176294831-176294831 on Build GRCh38
rs773499973 Chr.5: 176294975-176294975 on Build GRCh38
rs747371296 Chr.5: 176295106-176295106 on Build GRCh38
rs748266099 Chr.5: 176295001-176295001 on Build GRCh38
rs953619602 Chr.5: 176295079-176295079 on Build GRCh38
rs780244962 Chr.5: 176294921-176294921 on Build GRCh38
rs894363396 Chr.5: 176294915-176294915 on Build GRCh38
rs752279440 Chr.5: 176294981-176294981 on Build GRCh38
rs1000013784 Chr.5: 176294860-176294860 on Build GRCh38
rs568561545 Chr.5: 176295052-176295052 on Build GRCh38
rs143877714 Chr.5: 176294965-176294965 on Build GRCh38
rs143931126 Chr.5: 176295084-176295084 on Build GRCh38
rs186780886 Chr.5: 176294672-176294672 on Build GRCh38
rs780391683 Chr.5: 176294996-176294996 on Build GRCh38
rs372104813 Chr.5: 176294987-176294987 on Build GRCh38
rs973570865 Chr.5: 176295044-176295044 on Build GRCh38
rs781093286 Chr.5: 176295000-176295000 on Build GRCh38
rs756432742 Chr.5: 176294992-176294992 on Build GRCh38
rs546354407 Chr.5: 176294923-176294923 on Build GRCh38
rs565407993 Chr.5: 176294680-176294680 on Build GRCh38
rs771608867 Chr.5: 176295113-176295113 on Build GRCh38
rs70991529 Chr.5: 176294986-176294986 on Build GRCh38
rs538210044 Chr.5: 176294977-176294977 on Build GRCh38
rs369349173 Chr.5: 176294998-176294998 on Build GRCh38
rs749862630 Chr.5: 176295078-176295079 on Build GRCh38
rs774061160 Chr.5: 176294978-176294978 on Build GRCh38
rs752708366 Chr.5: 176295081-176295081 on Build GRCh38
rs113772037 Chr.5: 176294959-176294959 on Build GRCh38
rs62405163 Chr.5: 176294804-176294804 on Build GRCh38
rs770366551 Chr.5: 176294972-176294972 on Build GRCh38
rs17857141 Chr.5: 176295043-176295043 on Build GRCh38
rs780437165 Chr.5: 176295027-176295028 on Build GRCh38
rs757184123 Chr.5: 176294992-176294992 on Build GRCh38
rs764220921 Chr.5: 176294982-176294983 on Build GRCh38
rs759066697 Chr.5: 176295035-176295035 on Build GRCh38
rs373919516 Chr.5: 176295036-176295036 on Build GRCh38
rs908041204 Chr.5: 176294683-176294683 on Build GRCh38
rs750781334 Chr.5: 176294990-176294992 on Build GRCh38
rs776617514 Chr.5: 176294978-176294979 on Build GRCh38
rs749848042 Chr.5: 176295105-176295105 on Build GRCh38
rs147476048 Chr.5: 176295046-176295046 on Build GRCh38
rs970758890 Chr.5: 176294988-176294988 on Build GRCh38
rs770230817 Chr.5: 176295012-176295012 on Build GRCh38
rs761802142 Chr.5: 176295121-176295121 on Build GRCh38
rs114676330 Chr.5: 176294678-176294678 on Build GRCh38
rs759059793 Chr.5: 176294979-176294979 on Build GRCh38
rs751603034 Chr.5: 176294983-176294984 on Build GRCh38
rs183857393 Chr.5: 176294666-176294666 on Build GRCh38
rs781339144 Chr.5: 176294988-176294991 on Build GRCh38
rs758425538 Chr.5: 176294983-176294987 on Build GRCh38
rs546637106 Chr.5: 176294671-176294671 on Build GRCh38
rs932241580 Chr.5: 176294770-176294770 on Build GRCh38
rs940874816 Chr.5: 176294685-176294685 on Build GRCh38
rs938484415 Chr.5: 176294964-176294964 on Build GRCh38
rs76035021 Chr.5: 176294966-176294966 on Build GRCh38
rs944418755 Chr.5: 176294820-176294820 on Build GRCh38
rs765119980 Chr.5: 176294981-176294982 on Build GRCh38
rs759328159 Chr.5: 176294980-176294981 on Build GRCh38
rs72645800 Chr.5: 176294893-176294893 on Build GRCh38
rs530828030 Chr.5: 176294829-176294829 on Build GRCh38

View Full Product Details