Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176313418-176313942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIMC1
Entrez Gene ID
375484
Gene Name
SUMO interacting motifs containing 1
Gene Aliases
C5orf25, OOMA1, PLEIAD
UniGene
Hs.719847
Gene Chromosome Location
Chr.5: 176238359-176345991 on Build GRCh38
Transcripts
NM_001308195.1 NM_001308196.1 NM_001308200.1 NM_198567.5 NR_131772.1 XM_011534553.2 XM_011534554.2 XM_011534556.2 XM_017009454.1 XM_017009455.1 XM_017009456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
243385 c.1755G>T p.L585L Chr.5: 176313768-176313768 on Build GRCh38
6276651 c.77G>T p.G26V Chr.5: 176313707-176313707 on Build GRCh38
3340249 c.1720G>A p.V574I Chr.5: 176313733-176313733 on Build GRCh38
3827716 c.155A>C p.H52P Chr.5: 176313785-176313785 on Build GRCh38
6276650 c.1694G>T p.G565V Chr.5: 176313707-176313707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971948675 Chr.5: 176313619-176313619 on Build GRCh38
rs770731098 Chr.5: 176313642-176313643 on Build GRCh38
rs1042260758 Chr.5: 176313797-176313797 on Build GRCh38
rs374709500 Chr.5: 176313490-176313491 on Build GRCh38
rs114031020 Chr.5: 176313710-176313710 on Build GRCh38
rs567565220 Chr.5: 176313666-176313666 on Build GRCh38
rs764893300 Chr.5: 176313808-176313808 on Build GRCh38
rs77758389 Chr.5: 176313779-176313779 on Build GRCh38
rs762288341 Chr.5: 176313620-176313620 on Build GRCh38
rs745997796 Chr.5: 176313748-176313748 on Build GRCh38
rs368854039 Chr.5: 176313732-176313732 on Build GRCh38
rs1024770456 Chr.5: 176313896-176313896 on Build GRCh38
rs201347297 Chr.5: 176313894-176313894 on Build GRCh38
rs766334333 Chr.5: 176313812-176313812 on Build GRCh38
rs755095208 Chr.5: 176313560-176313561 on Build GRCh38
rs778810201 Chr.5: 176313484-176313484 on Build GRCh38
rs761089697 Chr.5: 176313778-176313778 on Build GRCh38
rs780198530 Chr.5: 176313861-176313861 on Build GRCh38
rs949851125 Chr.5: 176313854-176313854 on Build GRCh38
rs374763357 Chr.5: 176313715-176313715 on Build GRCh38
rs200331366 Chr.5: 176313733-176313733 on Build GRCh38
rs372198463 Chr.5: 176313670-176313670 on Build GRCh38
rs531702258 Chr.5: 176313482-176313482 on Build GRCh38
rs201494839 Chr.5: 176313657-176313657 on Build GRCh38
rs148729256 Chr.5: 176313736-176313736 on Build GRCh38
rs778040404 Chr.5: 176313730-176313730 on Build GRCh38
rs754953486 Chr.5: 176313821-176313821 on Build GRCh38
rs1050896426 Chr.5: 176313449-176313449 on Build GRCh38
rs763706462 Chr.5: 176313479-176313479 on Build GRCh38
rs546516477 Chr.5: 176313910-176313910 on Build GRCh38
rs199698815 Chr.5: 176313868-176313868 on Build GRCh38
rs751507525 Chr.5: 176313813-176313813 on Build GRCh38
rs545063224 Chr.5: 176313724-176313724 on Build GRCh38
rs750625989 Chr.5: 176313902-176313902 on Build GRCh38
rs755747090 Chr.5: 176313466-176313466 on Build GRCh38
rs935939385 Chr.5: 176313568-176313568 on Build GRCh38
rs774812653 Chr.5: 176313463-176313463 on Build GRCh38
rs772337879 Chr.5: 176313857-176313857 on Build GRCh38
rs745942561 Chr.5: 176313856-176313856 on Build GRCh38
rs945661641 Chr.5: 176313544-176313544 on Build GRCh38
rs556318372 Chr.5: 176313787-176313787 on Build GRCh38
rs867127089 Chr.5: 176313737-176313737 on Build GRCh38
rs898737361 Chr.5: 176313699-176313699 on Build GRCh38
rs1024357695 Chr.5: 176313608-176313608 on Build GRCh38
rs749381346 Chr.5: 176313532-176313532 on Build GRCh38
rs777672760 Chr.5: 176313562-176313562 on Build GRCh38
rs139181945 Chr.5: 176313665-176313665 on Build GRCh38
rs143105269 Chr.5: 176313811-176313811 on Build GRCh38
rs1004195126 Chr.5: 176313680-176313680 on Build GRCh38
rs374242380 Chr.5: 176313659-176313659 on Build GRCh38
rs572719647 Chr.5: 176313703-176313703 on Build GRCh38
rs894452878 Chr.5: 176313832-176313832 on Build GRCh38
rs1017375561 Chr.5: 176313698-176313698 on Build GRCh38
rs115828354 Chr.5: 176313603-176313603 on Build GRCh38
rs762215659 Chr.5: 176313892-176313892 on Build GRCh38
rs775930711 Chr.5: 176313775-176313775 on Build GRCh38
rs776308364 Chr.5: 176313698-176313699 on Build GRCh38
rs146338184 Chr.5: 176313526-176313526 on Build GRCh38
rs753495022 Chr.5: 176313682-176313682 on Build GRCh38
rs745748906 Chr.5: 176313485-176313485 on Build GRCh38
rs375250599 Chr.5: 176313495-176313495 on Build GRCh38
rs1050801254 Chr.5: 176313736-176313737 on Build GRCh38
rs779322331 Chr.5: 176313846-176313846 on Build GRCh38
rs568091736 Chr.5: 176313540-176313540 on Build GRCh38
rs779785356 Chr.5: 176313510-176313510 on Build GRCh38
rs746801607 Chr.5: 176313512-176313512 on Build GRCh38
rs758338112 Chr.5: 176313501-176313501 on Build GRCh38
rs776812429 Chr.5: 176313885-176313885 on Build GRCh38
rs774583573 Chr.5: 176313655-176313655 on Build GRCh38
rs751603042 Chr.5: 176313522-176313522 on Build GRCh38
rs945171241 Chr.5: 176313764-176313764 on Build GRCh38
rs368580789 Chr.5: 176313654-176313654 on Build GRCh38
rs749967357 Chr.5: 176313709-176313709 on Build GRCh38
rs764662477 Chr.5: 176313708-176313708 on Build GRCh38
rs772679526 Chr.5: 176313765-176313765 on Build GRCh38
rs752566710 Chr.5: 176313827-176313827 on Build GRCh38
rs935925413 Chr.5: 176313805-176313805 on Build GRCh38
rs187590792 Chr.5: 176313481-176313481 on Build GRCh38
rs753879109 Chr.5: 176313564-176313564 on Build GRCh38
rs761415553 Chr.5: 176313801-176313801 on Build GRCh38
rs529265725 Chr.5: 176313554-176313554 on Build GRCh38
rs925867849 Chr.5: 176313551-176313551 on Build GRCh38
rs977096359 Chr.5: 176313754-176313754 on Build GRCh38
rs757502195 Chr.5: 176313841-176313841 on Build GRCh38
rs111932354 Chr.5: 176313722-176313722 on Build GRCh38
rs894344763 Chr.5: 176313604-176313604 on Build GRCh38
rs756918023 Chr.5: 176313696-176313696 on Build GRCh38
rs571235824 Chr.5: 176313458-176313458 on Build GRCh38
rs61234855 Chr.5: 176313884-176313884 on Build GRCh38
rs746786294 Chr.5: 176313717-176313717 on Build GRCh38
rs977503503 Chr.5: 176313545-176313545 on Build GRCh38
rs780780652 Chr.5: 176313826-176313826 on Build GRCh38

View Full Product Details