Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176365318-176365823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL10
Entrez Gene ID
285598
Gene Name
ADP ribosylation factor like GTPase 10
Gene Aliases
ARL10A
UniGene
Hs.424552
Gene Chromosome Location
Chr.5: 176365468-176415085 on Build GRCh38
Transcripts
NM_001317948.1 NM_173664.5 XM_011534529.2 XM_011534530.2 XM_011534531.2 XM_017009372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391389 c.173A>G p.D58G Chr.5: 176365736-176365736 on Build GRCh38
4648874 c.104G>A p.G35D Chr.5: 176365667-176365667 on Build GRCh38
4625513 c.162C>T p.L54L Chr.5: 176365725-176365725 on Build GRCh38
3749914 c.39G>C p.L13L Chr.5: 176365602-176365602 on Build GRCh38
6431638 c.74T>C p.F25S Chr.5: 176365637-176365637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900123426 Chr.5: 176365397-176365397 on Build GRCh38
rs766092656 Chr.5: 176365594-176365594 on Build GRCh38
rs535799060 Chr.5: 176365360-176365360 on Build GRCh38
rs369791935 Chr.5: 176365501-176365501 on Build GRCh38
rs1015098378 Chr.5: 176365800-176365800 on Build GRCh38
rs983202633 Chr.5: 176365600-176365600 on Build GRCh38
rs558083807 Chr.5: 176365475-176365476 on Build GRCh38
rs531391251 Chr.5: 176365585-176365585 on Build GRCh38
rs1009329216 Chr.5: 176365378-176365378 on Build GRCh38
rs1010361237 Chr.5: 176365458-176365458 on Build GRCh38
rs941719003 Chr.5: 176365604-176365604 on Build GRCh38
rs767026413 Chr.5: 176365598-176365598 on Build GRCh38
rs921064191 Chr.5: 176365613-176365613 on Build GRCh38
rs1034316350 Chr.5: 176365695-176365695 on Build GRCh38
rs753708130 Chr.5: 176365633-176365633 on Build GRCh38
rs1020348106 Chr.5: 176365379-176365379 on Build GRCh38
rs1054008484 Chr.5: 176365410-176365410 on Build GRCh38
rs1000189102 Chr.5: 176365469-176365469 on Build GRCh38
rs982884993 Chr.5: 176365538-176365538 on Build GRCh38
rs890431516 Chr.5: 176365347-176365347 on Build GRCh38
rs1024509827 Chr.5: 176365746-176365746 on Build GRCh38
rs752562019 Chr.5: 176365599-176365599 on Build GRCh38
rs758544478 Chr.5: 176365635-176365635 on Build GRCh38
rs527538131 Chr.5: 176365490-176365490 on Build GRCh38
rs747332260 Chr.5: 176365691-176365691 on Build GRCh38
rs564436013 Chr.5: 176365546-176365546 on Build GRCh38
rs747210363 Chr.5: 176365648-176365648 on Build GRCh38
rs971069803 Chr.5: 176365536-176365536 on Build GRCh38
rs2303667 Chr.5: 176365602-176365602 on Build GRCh38
rs866606743 Chr.5: 176365421-176365421 on Build GRCh38
rs749724023 Chr.5: 176365778-176365778 on Build GRCh38
rs759102189 Chr.5: 176365596-176365596 on Build GRCh38
rs894737487 Chr.5: 176365708-176365708 on Build GRCh38
rs780437265 Chr.5: 176365688-176365688 on Build GRCh38
rs992959180 Chr.5: 176365516-176365516 on Build GRCh38
rs967903431 Chr.5: 176365398-176365398 on Build GRCh38
rs1033071794 Chr.5: 176365764-176365764 on Build GRCh38
rs559783335 Chr.5: 176365354-176365354 on Build GRCh38
rs781454696 Chr.5: 176365659-176365659 on Build GRCh38
rs903188297 Chr.5: 176365727-176365728 on Build GRCh38
rs36049692 Chr.5: 176365448-176365448 on Build GRCh38
rs755044890 Chr.5: 176365653-176365653 on Build GRCh38
rs945846540 Chr.5: 176365669-176365669 on Build GRCh38
rs748621120 Chr.5: 176365682-176365682 on Build GRCh38
rs1020380112 Chr.5: 176365465-176365465 on Build GRCh38
rs1041217251 Chr.5: 176365386-176365386 on Build GRCh38
rs374648151 Chr.5: 176365747-176365747 on Build GRCh38
rs944217999 Chr.5: 176365383-176365383 on Build GRCh38
rs982897829 Chr.5: 176365785-176365785 on Build GRCh38
rs1014492347 Chr.5: 176365734-176365734 on Build GRCh38
rs973061730 Chr.5: 176365627-176365627 on Build GRCh38
rs762494412 Chr.5: 176365593-176365593 on Build GRCh38
rs890335741 Chr.5: 176365665-176365665 on Build GRCh38
rs932484726 Chr.5: 176365629-176365629 on Build GRCh38
rs773727539 Chr.5: 176365595-176365595 on Build GRCh38
rs890554679 Chr.5: 176365371-176365371 on Build GRCh38
rs773414739 Chr.5: 176365769-176365769 on Build GRCh38
rs770246573 Chr.5: 176365549-176365549 on Build GRCh38
rs763826194 Chr.5: 176365610-176365610 on Build GRCh38
rs978899467 Chr.5: 176365432-176365432 on Build GRCh38
rs780382581 Chr.5: 176365647-176365647 on Build GRCh38
rs891057765 Chr.5: 176365663-176365663 on Build GRCh38
rs552177292 Chr.5: 176365545-176365545 on Build GRCh38
rs958962429 Chr.5: 176365499-176365499 on Build GRCh38
rs868540841 Chr.5: 176365466-176365466 on Build GRCh38
rs932889895 Chr.5: 176365404-176365404 on Build GRCh38
rs1042223890 Chr.5: 176365686-176365686 on Build GRCh38
rs773652521 Chr.5: 176365578-176365578 on Build GRCh38

View Full Product Details