Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30544450-30544951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNL1
Entrez Gene ID
2794
Gene Name
G protein nucleolar 1 (putative)
Gene Aliases
HSR1
UniGene
Hs.118354
Gene Chromosome Location
Chr.6: 30541377-30557594 on Build GRCh38
Transcripts
NM_005275.3 XM_005249015.3

SNP Detail

SNP ID Chromosome Location
rs1017717038 Chr.6: 30544533-30544533 on Build GRCh38
rs1009305922 Chr.6: 30544526-30544526 on Build GRCh38
rs965137773 Chr.6: 30544546-30544546 on Build GRCh38
rs569537197 Chr.6: 30544814-30544814 on Build GRCh38
rs997910229 Chr.6: 30544911-30544911 on Build GRCh38
rs982696267 Chr.6: 30544701-30544701 on Build GRCh38
rs1041914804 Chr.6: 30544486-30544486 on Build GRCh38
rs917271270 Chr.6: 30544775-30544775 on Build GRCh38
rs983679903 Chr.6: 30544629-30544629 on Build GRCh38
rs546332443 Chr.6: 30544773-30544773 on Build GRCh38
rs12192673 Chr.6: 30544728-30544728 on Build GRCh38
rs1018022898 Chr.6: 30544541-30544541 on Build GRCh38
rs927390429 Chr.6: 30544723-30544723 on Build GRCh38
rs767954195 Chr.6: 30544670-30544670 on Build GRCh38
rs757361302 Chr.6: 30544882-30544882 on Build GRCh38
rs954771480 Chr.6: 30544478-30544478 on Build GRCh38
rs964168540 Chr.6: 30544567-30544567 on Build GRCh38
rs1029308380 Chr.6: 30544604-30544604 on Build GRCh38
rs17195411 Chr.6: 30544826-30544826 on Build GRCh38
rs997794094 Chr.6: 30544570-30544570 on Build GRCh38
rs954936201 Chr.6: 30544619-30544619 on Build GRCh38
rs893979222 Chr.6: 30544891-30544891 on Build GRCh38
rs899574623 Chr.6: 30544881-30544881 on Build GRCh38
rs1041639753 Chr.6: 30544825-30544826 on Build GRCh38
rs945913771 Chr.6: 30544785-30544785 on Build GRCh38
rs577375329 Chr.6: 30544736-30544736 on Build GRCh38
rs184302063 Chr.6: 30544664-30544664 on Build GRCh38
rs949112349 Chr.6: 30544725-30544725 on Build GRCh38
rs1009689635 Chr.6: 30544910-30544910 on Build GRCh38
rs531724788 Chr.6: 30544921-30544921 on Build GRCh38
rs867126619 Chr.6: 30544841-30544841 on Build GRCh38
rs1039972794 Chr.6: 30544864-30544864 on Build GRCh38
rs760533299 Chr.6: 30544628-30544628 on Build GRCh38
rs574259959 Chr.6: 30544817-30544817 on Build GRCh38
rs935843640 Chr.6: 30544850-30544850 on Build GRCh38
rs137981827 Chr.6: 30544719-30544719 on Build GRCh38
rs753088753 Chr.6: 30544692-30544692 on Build GRCh38

View Full Product Details