Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30552388-30552888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNL1
Entrez Gene ID
2794
Gene Name
G protein nucleolar 1 (putative)
Gene Aliases
HSR1
UniGene
Hs.118354
Gene Chromosome Location
Chr.6: 30541377-30557594 on Build GRCh38
Transcripts
NM_005275.3 XM_005249015.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391629 c.1033G>A p.A345T Chr.6: 30552533-30552533 on Build GRCh38
5737526 c.923G>A p.R308Q Chr.6: 30552643-30552643 on Build GRCh38
6409071 c.976G>A p.E326K Chr.6: 30552590-30552590 on Build GRCh38
5885457 c.930G>C p.K310N Chr.6: 30552636-30552636 on Build GRCh38
3624256 c.993G>A p.E331E Chr.6: 30552573-30552573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142734504 Chr.6: 30552628-30552628 on Build GRCh38
rs112012560 Chr.6: 30552456-30552456 on Build GRCh38
rs569816924 Chr.6: 30552778-30552778 on Build GRCh38
rs748685725 Chr.6: 30552673-30552673 on Build GRCh38
rs545644145 Chr.6: 30552476-30552476 on Build GRCh38
rs768590494 Chr.6: 30552708-30552708 on Build GRCh38
rs760959959 Chr.6: 30552591-30552591 on Build GRCh38
rs762720597 Chr.6: 30552603-30552603 on Build GRCh38
rs767555737 Chr.6: 30552445-30552445 on Build GRCh38
rs944679333 Chr.6: 30552784-30552784 on Build GRCh38
rs149298420 Chr.6: 30552571-30552571 on Build GRCh38
rs774114882 Chr.6: 30552626-30552626 on Build GRCh38
rs747118452 Chr.6: 30552689-30552689 on Build GRCh38
rs73727440 Chr.6: 30552471-30552471 on Build GRCh38
rs755682042 Chr.6: 30552522-30552522 on Build GRCh38
rs1039165236 Chr.6: 30552790-30552790 on Build GRCh38
rs759779412 Chr.6: 30552706-30552706 on Build GRCh38
rs578228176 Chr.6: 30552446-30552446 on Build GRCh38
rs770239997 Chr.6: 30552420-30552420 on Build GRCh38
rs995306367 Chr.6: 30552798-30552798 on Build GRCh38
rs565517610 Chr.6: 30552835-30552835 on Build GRCh38
rs138612204 Chr.6: 30552567-30552567 on Build GRCh38
rs776901989 Chr.6: 30552598-30552598 on Build GRCh38
rs908000756 Chr.6: 30552696-30552696 on Build GRCh38
rs927174875 Chr.6: 30552737-30552737 on Build GRCh38
rs60791005 Chr.6: 30552438-30552438 on Build GRCh38
rs373408800 Chr.6: 30552650-30552650 on Build GRCh38
rs114647077 Chr.6: 30552847-30552847 on Build GRCh38
rs779200731 Chr.6: 30552665-30552665 on Build GRCh38
rs540874668 Chr.6: 30552693-30552693 on Build GRCh38
rs756865539 Chr.6: 30552508-30552508 on Build GRCh38
rs368987354 Chr.6: 30552700-30552700 on Build GRCh38
rs771990292 Chr.6: 30552853-30552854 on Build GRCh38
rs367682974 Chr.6: 30552656-30552656 on Build GRCh38
rs535537681 Chr.6: 30552794-30552794 on Build GRCh38
rs749555843 Chr.6: 30552555-30552555 on Build GRCh38
rs753499648 Chr.6: 30552652-30552652 on Build GRCh38
rs142398523 Chr.6: 30552590-30552590 on Build GRCh38
rs1056936410 Chr.6: 30552791-30552791 on Build GRCh38
rs191921728 Chr.6: 30552562-30552562 on Build GRCh38
rs767298599 Chr.6: 30552723-30552723 on Build GRCh38
rs959558112 Chr.6: 30552671-30552671 on Build GRCh38
rs763947588 Chr.6: 30552613-30552613 on Build GRCh38
rs755802323 Chr.6: 30552649-30552649 on Build GRCh38
rs780822937 Chr.6: 30552516-30552516 on Build GRCh38
rs150890159 Chr.6: 30552826-30552826 on Build GRCh38
rs145669779 Chr.6: 30552582-30552582 on Build GRCh38
rs747816532 Chr.6: 30552673-30552675 on Build GRCh38
rs545909929 Chr.6: 30552419-30552419 on Build GRCh38
rs576124674 Chr.6: 30552448-30552448 on Build GRCh38
rs767165987 Chr.6: 30552643-30552643 on Build GRCh38
rs775667118 Chr.6: 30552427-30552427 on Build GRCh38
rs1042993673 Chr.6: 30552731-30552731 on Build GRCh38
rs780470520 Chr.6: 30552530-30552530 on Build GRCh38
rs917077067 Chr.6: 30552843-30552843 on Build GRCh38
rs769051259 Chr.6: 30552557-30552557 on Build GRCh38
rs146068139 Chr.6: 30552644-30552644 on Build GRCh38
rs752372831 Chr.6: 30552479-30552479 on Build GRCh38
rs745533955 Chr.6: 30552521-30552521 on Build GRCh38
rs183243009 Chr.6: 30552846-30552846 on Build GRCh38
rs765116090 Chr.6: 30552477-30552477 on Build GRCh38
rs761949432 Chr.6: 30552444-30552444 on Build GRCh38
rs771003791 Chr.6: 30552592-30552592 on Build GRCh38
rs200336607 Chr.6: 30552547-30552547 on Build GRCh38
rs762642839 Chr.6: 30552502-30552502 on Build GRCh38
rs758372510 Chr.6: 30552487-30552487 on Build GRCh38
rs549908280 Chr.6: 30552730-30552730 on Build GRCh38
rs777848303 Chr.6: 30552679-30552679 on Build GRCh38
rs768976260 Chr.6: 30552433-30552433 on Build GRCh38
rs370439517 Chr.6: 30552565-30552565 on Build GRCh38
rs749832407 Chr.6: 30552432-30552432 on Build GRCh38

View Full Product Details