Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30552998-30553496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNL1
Entrez Gene ID
2794
Gene Name
G protein nucleolar 1 (putative)
Gene Aliases
HSR1
UniGene
Hs.118354
Gene Chromosome Location
Chr.6: 30541377-30557594 on Build GRCh38
Transcripts
NM_005275.3 XM_005249015.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586563 c.678G>A p.K226K Chr.6: 30553480-30553480 on Build GRCh38
5092594 c.765delT p.P256fs*>8 Chr.6: 30553387-30553387 on Build GRCh38
5531772 c.694C>T p.L232F Chr.6: 30553458-30553458 on Build GRCh38
4614226 c.834delG p.R279fs*13 Chr.6: 30553154-30553154 on Build GRCh38
3624258 c.778C>T p.R260C Chr.6: 30553374-30553374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772014960 Chr.6: 30553325-30553325 on Build GRCh38
rs563460637 Chr.6: 30553247-30553247 on Build GRCh38
rs201236335 Chr.6: 30553059-30553059 on Build GRCh38
rs753005350 Chr.6: 30553075-30553075 on Build GRCh38
rs71561384 Chr.6: 30553169-30553169 on Build GRCh38
rs762540586 Chr.6: 30553379-30553379 on Build GRCh38
rs2074505 Chr.6: 30553360-30553360 on Build GRCh38
rs746882476 Chr.6: 30553453-30553453 on Build GRCh38
rs767505010 Chr.6: 30553047-30553047 on Build GRCh38
rs199763857 Chr.6: 30553193-30553193 on Build GRCh38
rs758752898 Chr.6: 30553084-30553084 on Build GRCh38
rs759118409 Chr.6: 30553367-30553367 on Build GRCh38
rs372722311 Chr.6: 30553229-30553229 on Build GRCh38
rs758805993 Chr.6: 30553207-30553207 on Build GRCh38
rs906481027 Chr.6: 30553239-30553239 on Build GRCh38
rs1049611656 Chr.6: 30553212-30553212 on Build GRCh38
rs376889015 Chr.6: 30553450-30553450 on Build GRCh38
rs1034314713 Chr.6: 30553432-30553432 on Build GRCh38
rs1003388554 Chr.6: 30553317-30553317 on Build GRCh38
rs760634499 Chr.6: 30553077-30553078 on Build GRCh38
rs370068949 Chr.6: 30553340-30553340 on Build GRCh38
rs774244309 Chr.6: 30553041-30553041 on Build GRCh38
rs978372055 Chr.6: 30553107-30553107 on Build GRCh38
rs76843858 Chr.6: 30553294-30553295 on Build GRCh38
rs201780688 Chr.6: 30553158-30553158 on Build GRCh38
rs746105468 Chr.6: 30553146-30553146 on Build GRCh38
rs974956352 Chr.6: 30553383-30553383 on Build GRCh38
rs572313696 Chr.6: 30553068-30553068 on Build GRCh38
rs191144016 Chr.6: 30553213-30553213 on Build GRCh38
rs755163771 Chr.6: 30553423-30553423 on Build GRCh38
rs150038929 Chr.6: 30553408-30553408 on Build GRCh38
rs994929747 Chr.6: 30553411-30553411 on Build GRCh38
rs1033316377 Chr.6: 30553322-30553322 on Build GRCh38
rs147370104 Chr.6: 30553410-30553410 on Build GRCh38
rs371929141 Chr.6: 30553141-30553141 on Build GRCh38
rs764698949 Chr.6: 30553373-30553373 on Build GRCh38
rs1019099016 Chr.6: 30553377-30553377 on Build GRCh38
rs966295838 Chr.6: 30553378-30553378 on Build GRCh38
rs752347137 Chr.6: 30553205-30553205 on Build GRCh38
rs764396007 Chr.6: 30553382-30553382 on Build GRCh38
rs1036935004 Chr.6: 30553231-30553231 on Build GRCh38
rs747515982 Chr.6: 30553110-30553110 on Build GRCh38
rs772999660 Chr.6: 30553439-30553439 on Build GRCh38
rs1010692934 Chr.6: 30553338-30553338 on Build GRCh38
rs868594470 Chr.6: 30553067-30553067 on Build GRCh38
rs375281410 Chr.6: 30553063-30553063 on Build GRCh38
rs777444191 Chr.6: 30553441-30553441 on Build GRCh38
rs754587013 Chr.6: 30553064-30553064 on Build GRCh38
rs1015000708 Chr.6: 30553109-30553109 on Build GRCh38
rs372049810 Chr.6: 30553430-30553430 on Build GRCh38
rs764930170 Chr.6: 30553203-30553203 on Build GRCh38
rs139361425 Chr.6: 30553374-30553374 on Build GRCh38
rs542666619 Chr.6: 30553165-30553165 on Build GRCh38
rs780489835 Chr.6: 30553314-30553314 on Build GRCh38
rs770073126 Chr.6: 30553154-30553154 on Build GRCh38
rs373106261 Chr.6: 30553405-30553405 on Build GRCh38
rs770961116 Chr.6: 30553115-30553115 on Build GRCh38
rs139335715 Chr.6: 30553391-30553391 on Build GRCh38
rs369549010 Chr.6: 30553217-30553217 on Build GRCh38
rs757866378 Chr.6: 30553164-30553164 on Build GRCh38
rs778213877 Chr.6: 30553104-30553104 on Build GRCh38
rs761846710 Chr.6: 30553046-30553046 on Build GRCh38
rs188456748 Chr.6: 30553029-30553029 on Build GRCh38
rs377256021 Chr.6: 30553324-30553324 on Build GRCh38
rs764493803 Chr.6: 30553211-30553211 on Build GRCh38
rs148792484 Chr.6: 30553178-30553178 on Build GRCh38
rs967275299 Chr.6: 30553070-30553070 on Build GRCh38
rs765893337 Chr.6: 30553051-30553051 on Build GRCh38
rs71561383 Chr.6: 30553069-30553069 on Build GRCh38
rs541934202 Chr.6: 30553050-30553050 on Build GRCh38
rs749286575 Chr.6: 30553461-30553461 on Build GRCh38
rs746926257 Chr.6: 30553343-30553343 on Build GRCh38
rs781133892 Chr.6: 30553219-30553219 on Build GRCh38
rs770847074 Chr.6: 30553354-30553354 on Build GRCh38
rs747055154 Chr.6: 30553407-30553407 on Build GRCh38
rs757717783 Chr.6: 30553393-30553393 on Build GRCh38
rs770630082 Chr.6: 30553466-30553466 on Build GRCh38
rs368925664 Chr.6: 30553159-30553159 on Build GRCh38

View Full Product Details