Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30571310-30571813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCF1
Entrez Gene ID
23
Gene Name
ATP binding cassette subfamily F member 1
Gene Aliases
ABC27, ABC50
UniGene
Hs.655285
Gene Chromosome Location
Chr.6: 30571393-30591532 on Build GRCh38
Transcripts
NM_001025091.1 NM_001090.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1244009 c.73+2T>C p.? Chr.6: 30571562-30571562 on Build GRCh38
7124069 c.5C>T p.P2L Chr.6: 30571492-30571492 on Build GRCh38
6421983 c.22C>T p.Q8* Chr.6: 30571509-30571509 on Build GRCh38
1294235 c.26C>T p.P9L Chr.6: 30571513-30571513 on Build GRCh38
1294236 c.59G>A p.S20N Chr.6: 30571546-30571546 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773058674 Chr.6: 30571566-30571566 on Build GRCh38
rs888264502 Chr.6: 30571420-30571420 on Build GRCh38
rs745728667 Chr.6: 30571593-30571593 on Build GRCh38
rs370107811 Chr.6: 30571554-30571554 on Build GRCh38
rs886318389 Chr.6: 30571387-30571387 on Build GRCh38
rs566087434 Chr.6: 30571538-30571538 on Build GRCh38
rs763887332 Chr.6: 30571502-30571502 on Build GRCh38
rs1034120074 Chr.6: 30571626-30571626 on Build GRCh38
rs549451699 Chr.6: 30571520-30571520 on Build GRCh38
rs767297263 Chr.6: 30571563-30571563 on Build GRCh38
rs770916062 Chr.6: 30571550-30571550 on Build GRCh38
rs35008497 Chr.6: 30571437-30571438 on Build GRCh38
rs913456392 Chr.6: 30571348-30571348 on Build GRCh38
rs761595750 Chr.6: 30571509-30571509 on Build GRCh38
rs778697095 Chr.6: 30571533-30571533 on Build GRCh38
rs535954430 Chr.6: 30571773-30571773 on Build GRCh38
rs533438349 Chr.6: 30571355-30571355 on Build GRCh38
rs751410322 Chr.6: 30571507-30571507 on Build GRCh38
rs892846867 Chr.6: 30571627-30571627 on Build GRCh38
rs551480279 Chr.6: 30571646-30571646 on Build GRCh38
rs754620950 Chr.6: 30571522-30571522 on Build GRCh38
rs759683043 Chr.6: 30571501-30571501 on Build GRCh38
rs763833925 Chr.6: 30571446-30571446 on Build GRCh38
rs1023965439 Chr.6: 30571419-30571419 on Build GRCh38
rs1051063091 Chr.6: 30571352-30571352 on Build GRCh38
rs912547847 Chr.6: 30571487-30571487 on Build GRCh38
rs1009257426 Chr.6: 30571672-30571672 on Build GRCh38
rs922585558 Chr.6: 30571571-30571571 on Build GRCh38
rs751498234 Chr.6: 30571451-30571451 on Build GRCh38
rs769175168 Chr.6: 30571477-30571477 on Build GRCh38
rs776031365 Chr.6: 30571470-30571470 on Build GRCh38
rs779626964 Chr.6: 30571466-30571466 on Build GRCh38
rs941215977 Chr.6: 30571422-30571422 on Build GRCh38
rs759596698 Chr.6: 30571553-30571553 on Build GRCh38
rs906853752 Chr.6: 30571607-30571607 on Build GRCh38
rs969269707 Chr.6: 30571485-30571485 on Build GRCh38
rs1020409387 Chr.6: 30571687-30571687 on Build GRCh38
rs748905047 Chr.6: 30571468-30571468 on Build GRCh38
rs951359054 Chr.6: 30571782-30571782 on Build GRCh38
rs373820556 Chr.6: 30571560-30571560 on Build GRCh38
rs779355025 Chr.6: 30571481-30571481 on Build GRCh38
rs919986137 Chr.6: 30571536-30571536 on Build GRCh38
rs370826042 Chr.6: 30571455-30571455 on Build GRCh38
rs931377347 Chr.6: 30571746-30571746 on Build GRCh38
rs994644418 Chr.6: 30571377-30571377 on Build GRCh38
rs3132612 Chr.6: 30571661-30571661 on Build GRCh38
rs376635574 Chr.6: 30571458-30571458 on Build GRCh38
rs1004559246 Chr.6: 30571404-30571404 on Build GRCh38
rs528535182 Chr.6: 30571622-30571622 on Build GRCh38
rs1029714779 Chr.6: 30571768-30571768 on Build GRCh38
rs765820589 Chr.6: 30571591-30571591 on Build GRCh38
rs976791292 Chr.6: 30571752-30571752 on Build GRCh38
rs757126224 Chr.6: 30571452-30571452 on Build GRCh38
rs4148247 Chr.6: 30571428-30571428 on Build GRCh38
rs941149862 Chr.6: 30571612-30571612 on Build GRCh38
rs920017265 Chr.6: 30571742-30571742 on Build GRCh38
rs987246903 Chr.6: 30571486-30571486 on Build GRCh38
rs879395149 Chr.6: 30571739-30571739 on Build GRCh38
rs754532939 Chr.6: 30571593-30571593 on Build GRCh38
rs990251989 Chr.6: 30571462-30571462 on Build GRCh38
rs374538729 Chr.6: 30571456-30571456 on Build GRCh38
rs3130042 Chr.6: 30571350-30571350 on Build GRCh38
rs557131663 Chr.6: 30571693-30571693 on Build GRCh38
rs900321580 Chr.6: 30571376-30571376 on Build GRCh38
rs536928972 Chr.6: 30571691-30571691 on Build GRCh38
rs1056256496 Chr.6: 30571639-30571639 on Build GRCh38
rs191002614 Chr.6: 30571334-30571334 on Build GRCh38
rs968944368 Chr.6: 30571353-30571353 on Build GRCh38
rs753457440 Chr.6: 30571592-30571592 on Build GRCh38
rs762758549 Chr.6: 30571445-30571445 on Build GRCh38
rs879810641 Chr.6: 30571381-30571381 on Build GRCh38
rs1018442466 Chr.6: 30571719-30571719 on Build GRCh38
rs1039305278 Chr.6: 30571371-30571371 on Build GRCh38
rs1020258091 Chr.6: 30571721-30571721 on Build GRCh38
rs989104256 Chr.6: 30571351-30571351 on Build GRCh38
rs767397757 Chr.6: 30571513-30571513 on Build GRCh38
rs149350645 Chr.6: 30571500-30571500 on Build GRCh38
rs765030703 Chr.6: 30571598-30571598 on Build GRCh38
rs912674927 Chr.6: 30571365-30571365 on Build GRCh38
rs371238358 Chr.6: 30571539-30571539 on Build GRCh38
rs942798736 Chr.6: 30571366-30571366 on Build GRCh38
rs1020917112 Chr.6: 30571688-30571688 on Build GRCh38
rs754068128 Chr.6: 30571441-30571441 on Build GRCh38
rs770992423 Chr.6: 30571496-30571496 on Build GRCh38
rs975781426 Chr.6: 30571740-30571740 on Build GRCh38
rs905057975 Chr.6: 30571636-30571636 on Build GRCh38
rs942657026 Chr.6: 30571380-30571380 on Build GRCh38
rs183154838 Chr.6: 30571709-30571709 on Build GRCh38
rs990347389 Chr.6: 30571336-30571336 on Build GRCh38
rs1020494177 Chr.6: 30571467-30571467 on Build GRCh38
rs1018741162 Chr.6: 30571516-30571516 on Build GRCh38
rs753547024 Chr.6: 30571521-30571521 on Build GRCh38
rs987059084 Chr.6: 30571715-30571715 on Build GRCh38
rs760594771 Chr.6: 30571579-30571579 on Build GRCh38
rs529428649 Chr.6: 30571492-30571492 on Build GRCh38
rs1046990026 Chr.6: 30571419-30571420 on Build GRCh38
rs939066544 Chr.6: 30571637-30571637 on Build GRCh38
rs376093203 Chr.6: 30571549-30571549 on Build GRCh38
rs750356816 Chr.6: 30571515-30571515 on Build GRCh38
rs748680496 Chr.6: 30571484-30571484 on Build GRCh38
rs775661562 Chr.6: 30571559-30571559 on Build GRCh38
rs1045831442 Chr.6: 30571595-30571595 on Build GRCh38
rs1048893050 Chr.6: 30571546-30571546 on Build GRCh38

View Full Product Details