Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30580037-30580569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCF1
Entrez Gene ID
23
Gene Name
ATP binding cassette subfamily F member 1
Gene Aliases
ABC27, ABC50
UniGene
Hs.655285
Gene Chromosome Location
Chr.6: 30571393-30591532 on Build GRCh38
Transcripts
NM_001025091.1 NM_001090.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7353674 c.603G>T p.E201D Chr.6: 30580444-30580444 on Build GRCh38
1077348 c.579C>T p.F193F Chr.6: 30580420-30580420 on Build GRCh38
3410908 c.668A>G p.K223R Chr.6: 30580509-30580509 on Build GRCh38
6373911 c.665A>G p.K222R Chr.6: 30580506-30580506 on Build GRCh38
483823 c.677A>G p.Q226R Chr.6: 30580518-30580518 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs954240260 Chr.6: 30580411-30580411 on Build GRCh38
rs188349840 Chr.6: 30580484-30580484 on Build GRCh38
rs775620310 Chr.6: 30580361-30580362 on Build GRCh38
rs4148251 Chr.6: 30580361-30580361 on Build GRCh38
rs866934255 Chr.6: 30580374-30580374 on Build GRCh38
rs543266888 Chr.6: 30580084-30580084 on Build GRCh38
rs928077803 Chr.6: 30580263-30580263 on Build GRCh38
rs148910105 Chr.6: 30580454-30580454 on Build GRCh38
rs1055106942 Chr.6: 30580272-30580272 on Build GRCh38
rs575277265 Chr.6: 30580542-30580542 on Build GRCh38
rs768607015 Chr.6: 30580360-30580361 on Build GRCh38
rs141910056 Chr.6: 30580062-30580062 on Build GRCh38
rs374570578 Chr.6: 30580360-30580360 on Build GRCh38
rs552223658 Chr.6: 30580175-30580175 on Build GRCh38
rs780108913 Chr.6: 30580385-30580385 on Build GRCh38
rs138614350 Chr.6: 30580381-30580381 on Build GRCh38
rs1012851963 Chr.6: 30580137-30580137 on Build GRCh38
rs568821875 Chr.6: 30580204-30580204 on Build GRCh38
rs984557062 Chr.6: 30580072-30580072 on Build GRCh38
rs560843171 Chr.6: 30580161-30580161 on Build GRCh38
rs866445494 Chr.6: 30580271-30580271 on Build GRCh38
rs28687253 Chr.6: 30580362-30580362 on Build GRCh38
rs758935740 Chr.6: 30580441-30580441 on Build GRCh38
rs532141884 Chr.6: 30580109-30580109 on Build GRCh38
rs557726409 Chr.6: 30580220-30580220 on Build GRCh38
rs71533814 Chr.6: 30580360-30580360 on Build GRCh38
rs752239920 Chr.6: 30580448-30580448 on Build GRCh38
rs111709005 Chr.6: 30580116-30580116 on Build GRCh38
rs748025006 Chr.6: 30580388-30580388 on Build GRCh38
rs781188992 Chr.6: 30580375-30580375 on Build GRCh38
rs987489151 Chr.6: 30580233-30580233 on Build GRCh38
rs370131574 Chr.6: 30580113-30580113 on Build GRCh38
rs201353292 Chr.6: 30580338-30580345 on Build GRCh38
rs906620117 Chr.6: 30580112-30580112 on Build GRCh38
rs760865566 Chr.6: 30580494-30580496 on Build GRCh38
rs751167666 Chr.6: 30580199-30580199 on Build GRCh38
rs1021939212 Chr.6: 30580148-30580148 on Build GRCh38
rs948522321 Chr.6: 30580299-30580299 on Build GRCh38
rs920018768 Chr.6: 30580292-30580292 on Build GRCh38
rs773318274 Chr.6: 30580416-30580416 on Build GRCh38
rs1056875681 Chr.6: 30580121-30580121 on Build GRCh38
rs537701628 Chr.6: 30580329-30580330 on Build GRCh38
rs4148250 Chr.6: 30580347-30580347 on Build GRCh38
rs372349928 Chr.6: 30580375-30580375 on Build GRCh38
rs28561792 Chr.6: 30580360-30580360 on Build GRCh38
rs752967089 Chr.6: 30580448-30580450 on Build GRCh38
rs543194833 Chr.6: 30580090-30580090 on Build GRCh38
rs1042988604 Chr.6: 30580304-30580304 on Build GRCh38
rs6902544 Chr.6: 30580433-30580433 on Build GRCh38
rs894328855 Chr.6: 30580277-30580277 on Build GRCh38
rs567954927 Chr.6: 30580264-30580264 on Build GRCh38
rs550971825 Chr.6: 30580361-30580361 on Build GRCh38
rs529894783 Chr.6: 30580162-30580162 on Build GRCh38
rs553235913 Chr.6: 30580110-30580110 on Build GRCh38
rs781663400 Chr.6: 30580536-30580536 on Build GRCh38
rs1015743211 Chr.6: 30580245-30580245 on Build GRCh38
rs374454193 Chr.6: 30580170-30580170 on Build GRCh38
rs184057898 Chr.6: 30580342-30580342 on Build GRCh38
rs867279139 Chr.6: 30580500-30580500 on Build GRCh38
rs771719350 Chr.6: 30580401-30580401 on Build GRCh38
rs764985527 Chr.6: 30580445-30580445 on Build GRCh38
rs532199776 Chr.6: 30580173-30580173 on Build GRCh38
rs767563151 Chr.6: 30580374-30580375 on Build GRCh38
rs564909155 Chr.6: 30580128-30580128 on Build GRCh38
rs566482581 Chr.6: 30580172-30580172 on Build GRCh38
rs757818955 Chr.6: 30580526-30580526 on Build GRCh38
rs984406977 Chr.6: 30580216-30580216 on Build GRCh38
rs776344156 Chr.6: 30580436-30580436 on Build GRCh38
rs910085834 Chr.6: 30580219-30580219 on Build GRCh38
rs940071409 Chr.6: 30580234-30580234 on Build GRCh38
rs752020466 Chr.6: 30580365-30580365 on Build GRCh38
rs374854373 Chr.6: 30580361-30580361 on Build GRCh38
rs761950763 Chr.6: 30580361-30580365 on Build GRCh38
rs745936802 Chr.6: 30580376-30580376 on Build GRCh38
rs757334020 Chr.6: 30580370-30580370 on Build GRCh38
rs972706309 Chr.6: 30580240-30580240 on Build GRCh38
rs760626005 Chr.6: 30580420-30580420 on Build GRCh38
rs564759881 Chr.6: 30580085-30580085 on Build GRCh38
rs189740983 Chr.6: 30580155-30580155 on Build GRCh38
rs998486156 Chr.6: 30580163-30580163 on Build GRCh38
rs774663019 Chr.6: 30580361-30580363 on Build GRCh38
rs192870043 Chr.6: 30580206-30580206 on Build GRCh38
rs959758083 Chr.6: 30580366-30580366 on Build GRCh38
rs530719374 Chr.6: 30580142-30580142 on Build GRCh38
rs780954689 Chr.6: 30580394-30580394 on Build GRCh38
rs764298332 Chr.6: 30580470-30580470 on Build GRCh38
rs961589649 Chr.6: 30580434-30580434 on Build GRCh38
rs904428041 Chr.6: 30580347-30580347 on Build GRCh38

View Full Product Details