Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30729932-30730433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLOT1
Entrez Gene ID
10211
Gene Name
flotillin 1
Gene Aliases
-
UniGene
Hs.179986
Gene Chromosome Location
Chr.6: 30727709-30742851 on Build GRCh38
Transcripts
NM_001318875.1 NM_005803.3 XM_005248780.3 XM_006714947.3 XM_017010157.1 XM_017010158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3252084 c.1139T>C p.L380P Chr.6: 30730137-30730137 on Build GRCh38
6352160 c.1122A>G p.S374S Chr.6: 30730154-30730154 on Build GRCh38
3861533 c.1219A>C p.S407R Chr.6: 30730057-30730057 on Build GRCh38
3727412 c.1112C>T p.P371L Chr.6: 30730164-30730164 on Build GRCh38
4459140 c.1113C>T p.P371P Chr.6: 30730163-30730163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148562283 Chr.6: 30730319-30730319 on Build GRCh38
rs949020633 Chr.6: 30730316-30730316 on Build GRCh38
rs995907042 Chr.6: 30730006-30730006 on Build GRCh38
rs200728934 Chr.6: 30730115-30730115 on Build GRCh38
rs9968913 Chr.6: 30730408-30730408 on Build GRCh38
rs781454411 Chr.6: 30730395-30730395 on Build GRCh38
rs902397184 Chr.6: 30730358-30730358 on Build GRCh38
rs937829613 Chr.6: 30730243-30730243 on Build GRCh38
rs770279962 Chr.6: 30730077-30730077 on Build GRCh38
rs955353053 Chr.6: 30730069-30730069 on Build GRCh38
rs778572164 Chr.6: 30730223-30730223 on Build GRCh38
rs773750940 Chr.6: 30730149-30730149 on Build GRCh38
rs937536497 Chr.6: 30730193-30730193 on Build GRCh38
rs371329395 Chr.6: 30729986-30729986 on Build GRCh38
rs778465936 Chr.6: 30730234-30730234 on Build GRCh38
rs773173080 Chr.6: 30730033-30730033 on Build GRCh38
rs914695310 Chr.6: 30730219-30730219 on Build GRCh38
rs17189385 Chr.6: 30730339-30730339 on Build GRCh38
rs747496893 Chr.6: 30730381-30730381 on Build GRCh38
rs754638843 Chr.6: 30729995-30729995 on Build GRCh38
rs776564411 Chr.6: 30730390-30730390 on Build GRCh38
rs150262104 Chr.6: 30730127-30730127 on Build GRCh38
rs754593940 Chr.6: 30730216-30730216 on Build GRCh38
rs770019298 Chr.6: 30730133-30730133 on Build GRCh38
rs762881343 Chr.6: 30730037-30730037 on Build GRCh38
rs367715340 Chr.6: 30730038-30730038 on Build GRCh38
rs772594386 Chr.6: 30730017-30730017 on Build GRCh38
rs759614689 Chr.6: 30730407-30730407 on Build GRCh38
rs745341617 Chr.6: 30729981-30729981 on Build GRCh38
rs753293659 Chr.6: 30730208-30730208 on Build GRCh38
rs370922135 Chr.6: 30729975-30729975 on Build GRCh38
rs376833594 Chr.6: 30730406-30730406 on Build GRCh38
rs770784285 Chr.6: 30729995-30729997 on Build GRCh38
rs1055125964 Chr.6: 30730357-30730357 on Build GRCh38
rs1054857171 Chr.6: 30730244-30730244 on Build GRCh38
rs908434427 Chr.6: 30730091-30730091 on Build GRCh38
rs760214363 Chr.6: 30730203-30730203 on Build GRCh38
rs760783487 Chr.6: 30730035-30730035 on Build GRCh38
rs748480888 Chr.6: 30730092-30730092 on Build GRCh38
rs755381443 Chr.6: 30730078-30730078 on Build GRCh38
rs755765194 Chr.6: 30729983-30729983 on Build GRCh38
rs749948839 Chr.6: 30730045-30730045 on Build GRCh38
rs780590514 Chr.6: 30729978-30729978 on Build GRCh38
rs138527553 Chr.6: 30729994-30729995 on Build GRCh38
rs9500867 Chr.6: 30730154-30730154 on Build GRCh38
rs753065768 Chr.6: 30730232-30730232 on Build GRCh38
rs772472592 Chr.6: 30730093-30730093 on Build GRCh38
rs375201135 Chr.6: 30730000-30730000 on Build GRCh38
rs750355840 Chr.6: 30730198-30730198 on Build GRCh38
rs113030936 Chr.6: 30730049-30730049 on Build GRCh38
rs761443032 Chr.6: 30730058-30730058 on Build GRCh38
rs779744796 Chr.6: 30730075-30730075 on Build GRCh38
rs778334654 Chr.6: 30730112-30730112 on Build GRCh38
rs1042079304 Chr.6: 30730322-30730322 on Build GRCh38
rs759861713 Chr.6: 30730131-30730131 on Build GRCh38
rs917686339 Chr.6: 30730256-30730256 on Build GRCh38
rs201480751 Chr.6: 30730384-30730384 on Build GRCh38
rs1802471 Chr.6: 30730043-30730043 on Build GRCh38
rs372109926 Chr.6: 30730016-30730016 on Build GRCh38
rs774019896 Chr.6: 30730135-30730135 on Build GRCh38
rs9501029 Chr.6: 30729963-30729963 on Build GRCh38
rs751320927 Chr.6: 30730039-30730039 on Build GRCh38
rs537062839 Chr.6: 30730332-30730332 on Build GRCh38
rs140940537 Chr.6: 30730126-30730126 on Build GRCh38
rs3179198 Chr.6: 30730019-30730019 on Build GRCh38
rs58436052 Chr.6: 30729996-30729997 on Build GRCh38
rs779164631 Chr.6: 30730079-30730079 on Build GRCh38
rs745875039 Chr.6: 30730398-30730398 on Build GRCh38
rs538750081 Chr.6: 30729980-30729980 on Build GRCh38
rs201200201 Chr.6: 30730036-30730036 on Build GRCh38
rs919735765 Chr.6: 30730224-30730224 on Build GRCh38
rs774960591 Chr.6: 30730012-30730012 on Build GRCh38
rs560077348 Chr.6: 30730241-30730241 on Build GRCh38
rs554872476 Chr.6: 30730139-30730139 on Build GRCh38
rs540464383 Chr.6: 30730205-30730205 on Build GRCh38
rs767419945 Chr.6: 30730044-30730044 on Build GRCh38

View Full Product Details