Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30740597-30741109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLOT1
Entrez Gene ID
10211
Gene Name
flotillin 1
Gene Aliases
-
UniGene
Hs.179986
Gene Chromosome Location
Chr.6: 30727709-30742851 on Build GRCh38
Transcripts
NM_001318875.1 NM_005803.3 XM_005248780.3 XM_006714947.3 XM_017010157.1 XM_017010158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7102830 c.381C>G p.F127L Chr.6: 30740772-30740772 on Build GRCh38
4424929 c.428G>A p.G143D Chr.6: 30740725-30740725 on Build GRCh38
6349489 c.190-9C>G p.? Chr.6: 30740600-30740600 on Build GRCh38
4160576 c.377A>T p.K126I Chr.6: 30740776-30740776 on Build GRCh38
6349488 c.475-9C>G p.? Chr.6: 30740600-30740600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780076990 Chr.6: 30740844-30740845 on Build GRCh38
rs761010775 Chr.6: 30740820-30740820 on Build GRCh38
rs780865327 Chr.6: 30740709-30740709 on Build GRCh38
rs761072513 Chr.6: 30740809-30740811 on Build GRCh38
rs756789679 Chr.6: 30740694-30740694 on Build GRCh38
rs541425313 Chr.6: 30740856-30740856 on Build GRCh38
rs761359076 Chr.6: 30740847-30740847 on Build GRCh38
rs762799759 Chr.6: 30740837-30740837 on Build GRCh38
rs563176435 Chr.6: 30740908-30740908 on Build GRCh38
rs749526570 Chr.6: 30740846-30740847 on Build GRCh38
rs918680514 Chr.6: 30740835-30740835 on Build GRCh38
rs79585365 Chr.6: 30740928-30740928 on Build GRCh38
rs897496084 Chr.6: 30740988-30740988 on Build GRCh38
rs951353966 Chr.6: 30740836-30740836 on Build GRCh38
rs752416611 Chr.6: 30740833-30740834 on Build GRCh38
rs949983730 Chr.6: 30740880-30740880 on Build GRCh38
rs571256041 Chr.6: 30740947-30740947 on Build GRCh38
rs775165694 Chr.6: 30740826-30740826 on Build GRCh38
rs3130661 Chr.6: 30740918-30740918 on Build GRCh38
rs76989606 Chr.6: 30740937-30740937 on Build GRCh38
rs755711731 Chr.6: 30740725-30740725 on Build GRCh38
rs564092176 Chr.6: 30740860-30740860 on Build GRCh38
rs762804018 Chr.6: 30740666-30740666 on Build GRCh38
rs1018946071 Chr.6: 30741036-30741036 on Build GRCh38
rs917934075 Chr.6: 30740874-30740874 on Build GRCh38
rs764039022 Chr.6: 30740687-30740687 on Build GRCh38
rs541987555 Chr.6: 30740715-30740715 on Build GRCh38
rs548736019 Chr.6: 30740838-30740839 on Build GRCh38
rs772477281 Chr.6: 30740640-30740640 on Build GRCh38
rs149127648 Chr.6: 30740881-30740881 on Build GRCh38
rs1040100868 Chr.6: 30740953-30740953 on Build GRCh38
rs1050772531 Chr.6: 30740943-30740943 on Build GRCh38
rs1005604733 Chr.6: 30740835-30740843 on Build GRCh38
rs34375360 Chr.6: 30740834-30740835 on Build GRCh38
rs139305218 Chr.6: 30740741-30740741 on Build GRCh38
rs12111310 Chr.6: 30740651-30740651 on Build GRCh38
rs751092275 Chr.6: 30740688-30740688 on Build GRCh38
rs749596091 Chr.6: 30740745-30740745 on Build GRCh38
rs1006156299 Chr.6: 30741075-30741075 on Build GRCh38
rs555852170 Chr.6: 30740819-30740819 on Build GRCh38
rs766681940 Chr.6: 30740772-30740772 on Build GRCh38
rs556875495 Chr.6: 30741073-30741073 on Build GRCh38
rs757979530 Chr.6: 30741082-30741082 on Build GRCh38
rs994333324 Chr.6: 30740782-30740782 on Build GRCh38
rs1025772674 Chr.6: 30740817-30740817 on Build GRCh38
rs772245658 Chr.6: 30740768-30740768 on Build GRCh38
rs779461045 Chr.6: 30740748-30740748 on Build GRCh38
rs769056175 Chr.6: 30740747-30740747 on Build GRCh38
rs752498099 Chr.6: 30740653-30740653 on Build GRCh38
rs73727468 Chr.6: 30740920-30740920 on Build GRCh38
rs543301376 Chr.6: 30740885-30740885 on Build GRCh38
rs995645974 Chr.6: 30741064-30741064 on Build GRCh38
rs769166000 Chr.6: 30740634-30740634 on Build GRCh38
rs777343413 Chr.6: 30740835-30740837 on Build GRCh38
rs944344720 Chr.6: 30740946-30740946 on Build GRCh38
rs748240708 Chr.6: 30740754-30740754 on Build GRCh38
rs753781500 Chr.6: 30740987-30740987 on Build GRCh38
rs1041304126 Chr.6: 30740703-30740703 on Build GRCh38
rs536062519 Chr.6: 30741087-30741087 on Build GRCh38
rs1047775073 Chr.6: 30741041-30741041 on Build GRCh38
rs898634942 Chr.6: 30741037-30741037 on Build GRCh38
rs771211611 Chr.6: 30740822-30740822 on Build GRCh38
rs749780693 Chr.6: 30740626-30740626 on Build GRCh38
rs1025659167 Chr.6: 30741068-30741068 on Build GRCh38
rs761890901 Chr.6: 30740639-30740639 on Build GRCh38
rs764007535 Chr.6: 30740843-30740843 on Build GRCh38
rs962889608 Chr.6: 30740690-30740690 on Build GRCh38
rs369115088 Chr.6: 30741083-30741083 on Build GRCh38
rs527661312 Chr.6: 30740681-30740681 on Build GRCh38
rs143103020 Chr.6: 30740927-30740927 on Build GRCh38
rs1018437362 Chr.6: 30740665-30740665 on Build GRCh38
rs759456227 Chr.6: 30740644-30740644 on Build GRCh38
rs781317594 Chr.6: 30740835-30740836 on Build GRCh38
rs888597311 Chr.6: 30741062-30741062 on Build GRCh38
rs930177994 Chr.6: 30740999-30740999 on Build GRCh38
rs888619975 Chr.6: 30740827-30740827 on Build GRCh38
rs969830732 Chr.6: 30741086-30741086 on Build GRCh38
rs901457596 Chr.6: 30740731-30740731 on Build GRCh38
rs774295690 Chr.6: 30740637-30740637 on Build GRCh38
rs9281028 Chr.6: 30740856-30740857 on Build GRCh38
rs540172197 Chr.6: 30741006-30741006 on Build GRCh38
rs757789653 Chr.6: 30740835-30740838 on Build GRCh38
rs757294464 Chr.6: 30741019-30741019 on Build GRCh38
rs745617570 Chr.6: 30740835-30740835 on Build GRCh38
rs572415300 Chr.6: 30740846-30740846 on Build GRCh38
rs985882239 Chr.6: 30740929-30740929 on Build GRCh38
rs769732832 Chr.6: 30740843-30740844 on Build GRCh38
rs764761531 Chr.6: 30740830-30740831 on Build GRCh38

View Full Product Details