Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31144771-31145274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCHCR1
Entrez Gene ID
54535
Gene Name
coiled-coil alpha-helical rod protein 1
Gene Aliases
C6orf18, HCR, SBP
UniGene
Hs.485075
Gene Chromosome Location
Chr.6: 31142439-31158238 on Build GRCh38
Transcripts
NM_001105563.1 NM_001105564.1 NM_019052.3 XM_011514702.2 XM_011514703.1 XM_011514704.2 XM_011514705.1 XM_017010961.1 XM_017010962.1 XM_017010963.1 XM_017010964.1 XM_017010965.1 XM_017010966.1 XM_017010967.1 XM_017010968.1 XM_017010969.1 XM_017010970.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4435160 c.1637A>G p.K546R Chr.6: 31145046-31145046 on Build GRCh38
6502879 c.1792G>A p.G598R Chr.6: 31144891-31144891 on Build GRCh38
4532352 c.1877G>A p.G626E Chr.6: 31145073-31145073 on Build GRCh38
5625318 c.2032G>A p.E678K Chr.6: 31144918-31144918 on Build GRCh38
4389769 c.1666C>T p.Q556* Chr.6: 31145017-31145017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758053234 Chr.6: 31145253-31145255 on Build GRCh38
rs190832200 Chr.6: 31144808-31144808 on Build GRCh38
rs776217188 Chr.6: 31144931-31144931 on Build GRCh38
rs779730871 Chr.6: 31145062-31145062 on Build GRCh38
rs757520786 Chr.6: 31144864-31144864 on Build GRCh38
rs144588628 Chr.6: 31145175-31145175 on Build GRCh38
rs184553289 Chr.6: 31145125-31145125 on Build GRCh38
rs866756226 Chr.6: 31144828-31144828 on Build GRCh38
rs745552180 Chr.6: 31145047-31145047 on Build GRCh38
rs773444233 Chr.6: 31145105-31145105 on Build GRCh38
rs778801618 Chr.6: 31144897-31144897 on Build GRCh38
rs567133818 Chr.6: 31145138-31145138 on Build GRCh38
rs751470127 Chr.6: 31145203-31145203 on Build GRCh38
rs748123638 Chr.6: 31144900-31144900 on Build GRCh38
rs765837979 Chr.6: 31144834-31144834 on Build GRCh38
rs778321479 Chr.6: 31145240-31145240 on Build GRCh38
rs754470763 Chr.6: 31145233-31145233 on Build GRCh38
rs747364417 Chr.6: 31145241-31145241 on Build GRCh38
rs759682734 Chr.6: 31145116-31145116 on Build GRCh38
rs915584904 Chr.6: 31144880-31144880 on Build GRCh38
rs776807780 Chr.6: 31145250-31145250 on Build GRCh38
rs752582662 Chr.6: 31145007-31145007 on Build GRCh38
rs779638131 Chr.6: 31145144-31145144 on Build GRCh38
rs777366958 Chr.6: 31145033-31145033 on Build GRCh38
rs895519715 Chr.6: 31144915-31144915 on Build GRCh38
rs35679674 Chr.6: 31144984-31144985 on Build GRCh38
rs758152121 Chr.6: 31144911-31144911 on Build GRCh38
rs372842578 Chr.6: 31144807-31144807 on Build GRCh38
rs776220440 Chr.6: 31144814-31144814 on Build GRCh38
rs751689523 Chr.6: 31144851-31144851 on Build GRCh38
rs761674916 Chr.6: 31145204-31145204 on Build GRCh38
rs753921504 Chr.6: 31145114-31145114 on Build GRCh38
rs111504259 Chr.6: 31145031-31145031 on Build GRCh38
rs748632015 Chr.6: 31145234-31145234 on Build GRCh38
rs746819053 Chr.6: 31145035-31145035 on Build GRCh38
rs1265078 Chr.6: 31144825-31144825 on Build GRCh38
rs142986308 Chr.6: 31144924-31144924 on Build GRCh38
rs201470492 Chr.6: 31145085-31145085 on Build GRCh38
rs899284404 Chr.6: 31145146-31145146 on Build GRCh38
rs776900219 Chr.6: 31145180-31145180 on Build GRCh38
rs748634811 Chr.6: 31145171-31145171 on Build GRCh38
rs751505133 Chr.6: 31145123-31145123 on Build GRCh38
rs750779467 Chr.6: 31144845-31144851 on Build GRCh38
rs1022602600 Chr.6: 31145202-31145202 on Build GRCh38
rs148460661 Chr.6: 31145181-31145181 on Build GRCh38
rs762225280 Chr.6: 31145084-31145084 on Build GRCh38
rs771140403 Chr.6: 31145178-31145178 on Build GRCh38
rs777568339 Chr.6: 31144917-31144917 on Build GRCh38
rs2073720 Chr.6: 31145046-31145046 on Build GRCh38
rs879198967 Chr.6: 31144812-31144812 on Build GRCh38
rs757062392 Chr.6: 31145040-31145040 on Build GRCh38
rs140284482 Chr.6: 31144972-31144972 on Build GRCh38
rs2073719 Chr.6: 31145148-31145148 on Build GRCh38
rs769289623 Chr.6: 31145064-31145064 on Build GRCh38
rs557052806 Chr.6: 31145217-31145217 on Build GRCh38
rs765268981 Chr.6: 31145200-31145200 on Build GRCh38
rs778611988 Chr.6: 31144800-31144800 on Build GRCh38
rs766390211 Chr.6: 31145113-31145113 on Build GRCh38
rs750043921 Chr.6: 31145134-31145134 on Build GRCh38
rs769473592 Chr.6: 31144957-31144957 on Build GRCh38
rs764501660 Chr.6: 31144967-31144967 on Build GRCh38
rs767436631 Chr.6: 31144875-31144875 on Build GRCh38
rs773792118 Chr.6: 31144913-31144913 on Build GRCh38
rs551097618 Chr.6: 31145071-31145071 on Build GRCh38
rs778373022 Chr.6: 31145083-31145083 on Build GRCh38
rs3094226 Chr.6: 31145122-31145122 on Build GRCh38
rs555766439 Chr.6: 31145074-31145075 on Build GRCh38
rs756252094 Chr.6: 31144891-31144891 on Build GRCh38
rs532197348 Chr.6: 31144811-31144811 on Build GRCh38
rs754583644 Chr.6: 31145128-31145128 on Build GRCh38
rs751994930 Chr.6: 31144827-31144827 on Build GRCh38
rs767773705 Chr.6: 31144975-31144975 on Build GRCh38
rs552019115 Chr.6: 31145216-31145216 on Build GRCh38
rs748801113 Chr.6: 31145147-31145147 on Build GRCh38
rs201259956 Chr.6: 31145249-31145249 on Build GRCh38
rs373003491 Chr.6: 31145174-31145174 on Build GRCh38
rs769385738 Chr.6: 31145051-31145051 on Build GRCh38
rs775826272 Chr.6: 31145003-31145003 on Build GRCh38
rs745374329 Chr.6: 31144940-31144940 on Build GRCh38
rs377556857 Chr.6: 31144868-31144868 on Build GRCh38
rs766478370 Chr.6: 31145006-31145006 on Build GRCh38
rs758346684 Chr.6: 31145020-31145020 on Build GRCh38
rs770431424 Chr.6: 31144857-31144857 on Build GRCh38
rs3132539 Chr.6: 31145253-31145253 on Build GRCh38
rs199923626 Chr.6: 31145235-31145235 on Build GRCh38
rs746587026 Chr.6: 31144923-31144923 on Build GRCh38
rs772821439 Chr.6: 31144809-31144809 on Build GRCh38
rs868454605 Chr.6: 31145073-31145074 on Build GRCh38
rs779839053 Chr.6: 31145218-31145218 on Build GRCh38
rs933918624 Chr.6: 31145061-31145061 on Build GRCh38
rs193084124 Chr.6: 31145027-31145027 on Build GRCh38
rs367616484 Chr.6: 31145161-31145161 on Build GRCh38
rs369194135 Chr.6: 31144835-31144835 on Build GRCh38
rs969812007 Chr.6: 31144892-31144892 on Build GRCh38
rs746708455 Chr.6: 31144810-31144810 on Build GRCh38
rs764591247 Chr.6: 31144836-31144836 on Build GRCh38
rs934471154 Chr.6: 31145001-31145001 on Build GRCh38
rs752381617 Chr.6: 31145113-31145114 on Build GRCh38
rs750521371 Chr.6: 31144885-31144885 on Build GRCh38
rs130079 Chr.6: 31144960-31144960 on Build GRCh38
rs575266029 Chr.6: 31145228-31145228 on Build GRCh38
rs147152245 Chr.6: 31144966-31144966 on Build GRCh38
rs760911154 Chr.6: 31145107-31145107 on Build GRCh38
rs188955569 Chr.6: 31145205-31145205 on Build GRCh38
rs777231387 Chr.6: 31144965-31144965 on Build GRCh38
rs756155788 Chr.6: 31144991-31144991 on Build GRCh38
rs750668764 Chr.6: 31144978-31144978 on Build GRCh38

View Full Product Details