Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31536365-31536858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1G2-DDX39B
Entrez Gene ID
100532737
Gene Name
ATP6V1G2-DDX39B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31546848 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1 NR_003140.1

Gene Symbol
DDX39B
Entrez Gene ID
7919
Gene Name
DEAD-box helicase 39B
Gene Aliases
BAT1, D6S81E, UAP56
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31542475 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1 NR_003140.1

Gene Symbol
SNORD117
Entrez Gene ID
692233
Gene Name
small nucleolar RNA, C/D box 117
Gene Aliases
U83
UniGene
Gene Chromosome Location
Chr.6: 31536374-31536449 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1 NR_003140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7000207 c.523C>T p.R175C Chr.6: 31536593-31536593 on Build GRCh38
3253203 c.516_517insT p.G173fs*9 Chr.6: 31536668-31536669 on Build GRCh38
6641740 c.485T>G p.V162G Chr.6: 31536676-31536676 on Build GRCh38
2149592 c.448_449delGG p.G150fs*8 Chr.6: 31536667-31536668 on Build GRCh38
4781942 c.532G>T p.G178C Chr.6: 31536668-31536668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs10456396 Chr.6: 31536769-31536769 on Build GRCh38
rs551501760 Chr.6: 31536439-31536439 on Build GRCh38
rs369715555 Chr.6: 31536701-31536702 on Build GRCh38
rs773774748 Chr.6: 31536477-31536477 on Build GRCh38
rs1026886188 Chr.6: 31536618-31536618 on Build GRCh38
rs1005257156 Chr.6: 31536688-31536688 on Build GRCh38
rs750255910 Chr.6: 31536746-31536746 on Build GRCh38
rs752659011 Chr.6: 31536414-31536414 on Build GRCh38
rs190488922 Chr.6: 31536603-31536603 on Build GRCh38
rs759729035 Chr.6: 31536588-31536588 on Build GRCh38
rs370794080 Chr.6: 31536487-31536487 on Build GRCh38
rs777891843 Chr.6: 31536668-31536669 on Build GRCh38
rs375020328 Chr.6: 31536813-31536813 on Build GRCh38
rs566490834 Chr.6: 31536444-31536444 on Build GRCh38
rs778473688 Chr.6: 31536686-31536686 on Build GRCh38
rs760876299 Chr.6: 31536389-31536389 on Build GRCh38
rs757158055 Chr.6: 31536619-31536619 on Build GRCh38
rs778760179 Chr.6: 31536609-31536609 on Build GRCh38
rs758489522 Chr.6: 31536625-31536627 on Build GRCh38
rs9279337 Chr.6: 31536700-31536701 on Build GRCh38
rs1043589551 Chr.6: 31536803-31536803 on Build GRCh38
rs751496472 Chr.6: 31536412-31536412 on Build GRCh38
rs973082055 Chr.6: 31536816-31536816 on Build GRCh38
rs753546511 Chr.6: 31536479-31536479 on Build GRCh38
rs377444138 Chr.6: 31536689-31536689 on Build GRCh38
rs376725953 Chr.6: 31536771-31536772 on Build GRCh38
rs1050750044 Chr.6: 31536660-31536660 on Build GRCh38
rs755119073 Chr.6: 31536397-31536397 on Build GRCh38
rs777527702 Chr.6: 31536404-31536404 on Build GRCh38
rs766550277 Chr.6: 31536550-31536550 on Build GRCh38
rs771267299 Chr.6: 31536734-31536734 on Build GRCh38
rs754655043 Chr.6: 31536682-31536682 on Build GRCh38
rs527269585 Chr.6: 31536456-31536456 on Build GRCh38
rs768189104 Chr.6: 31536445-31536445 on Build GRCh38
rs748938626 Chr.6: 31536679-31536679 on Build GRCh38
rs75750725 Chr.6: 31536668-31536668 on Build GRCh38
rs747327894 Chr.6: 31536724-31536724 on Build GRCh38
rs140432746 Chr.6: 31536781-31536781 on Build GRCh38
rs753851723 Chr.6: 31536396-31536396 on Build GRCh38
rs776938788 Chr.6: 31536573-31536573 on Build GRCh38
rs755993810 Chr.6: 31536639-31536639 on Build GRCh38
rs960893063 Chr.6: 31536748-31536748 on Build GRCh38
rs373225565 Chr.6: 31536627-31536627 on Build GRCh38
rs913763708 Chr.6: 31536393-31536393 on Build GRCh38
rs1007941500 Chr.6: 31536553-31536553 on Build GRCh38
rs533016755 Chr.6: 31536438-31536438 on Build GRCh38
rs375972885 Chr.6: 31536717-31536717 on Build GRCh38
rs3130057 Chr.6: 31536810-31536810 on Build GRCh38
rs114751021 Chr.6: 31536417-31536417 on Build GRCh38
rs781117143 Chr.6: 31536427-31536427 on Build GRCh38
rs781707463 Chr.6: 31536802-31536802 on Build GRCh38
rs371213671 Chr.6: 31536714-31536714 on Build GRCh38
rs1040380322 Chr.6: 31536587-31536587 on Build GRCh38
rs773509777 Chr.6: 31536501-31536501 on Build GRCh38
rs373156900 Chr.6: 31536395-31536395 on Build GRCh38
rs779882199 Chr.6: 31536440-31536440 on Build GRCh38
rs969112454 Chr.6: 31536696-31536696 on Build GRCh38

View Full Product Details