Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 107028249-107028786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf203
Entrez Gene ID
51250
Gene Name
chromosome 6 open reading frame 203
Gene Aliases
HSPC230, PRED31
UniGene
Hs.486084
Gene Chromosome Location
Chr.6: 107028172-107051586 on Build GRCh38
Transcripts
NM_001142468.2 NM_001142470.2 NM_016487.4 XM_011535877.2 XM_017010920.1

SNP Detail

SNP ID Chromosome Location
rs1007403279 Chr.6: 107028522-107028522 on Build GRCh38
rs1007930142 Chr.6: 107028270-107028270 on Build GRCh38
rs570019290 Chr.6: 107028369-107028369 on Build GRCh38
rs989901056 Chr.6: 107028302-107028302 on Build GRCh38
rs919247428 Chr.6: 107028430-107028430 on Build GRCh38
rs928208475 Chr.6: 107028395-107028395 on Build GRCh38
rs534092207 Chr.6: 107028520-107028520 on Build GRCh38
rs939554206 Chr.6: 107028397-107028397 on Build GRCh38
rs781397037 Chr.6: 107028557-107028557 on Build GRCh38
rs6904668 Chr.6: 107028709-107028709 on Build GRCh38
rs56352510 Chr.6: 107028472-107028472 on Build GRCh38
rs751788234 Chr.6: 107028368-107028368 on Build GRCh38
rs185468237 Chr.6: 107028742-107028742 on Build GRCh38
rs570966177 Chr.6: 107028320-107028320 on Build GRCh38
rs547551615 Chr.6: 107028306-107028306 on Build GRCh38
rs980975256 Chr.6: 107028665-107028665 on Build GRCh38
rs533417117 Chr.6: 107028325-107028325 on Build GRCh38
rs139553925 Chr.6: 107028623-107028623 on Build GRCh38
rs782685561 Chr.6: 107028476-107028476 on Build GRCh38
rs550052568 Chr.6: 107028359-107028359 on Build GRCh38
rs568327181 Chr.6: 107028487-107028487 on Build GRCh38
rs6904173 Chr.6: 107028617-107028617 on Build GRCh38
rs1031325171 Chr.6: 107028572-107028572 on Build GRCh38
rs781804256 Chr.6: 107028279-107028279 on Build GRCh38
rs970177157 Chr.6: 107028637-107028637 on Build GRCh38
rs960977065 Chr.6: 107028699-107028699 on Build GRCh38
rs188609891 Chr.6: 107028693-107028693 on Build GRCh38
rs545875744 Chr.6: 107028661-107028661 on Build GRCh38
rs998493224 Chr.6: 107028282-107028282 on Build GRCh38
rs377252789 Chr.6: 107028717-107028717 on Build GRCh38
rs969579313 Chr.6: 107028314-107028314 on Build GRCh38
rs76852209 Chr.6: 107028304-107028304 on Build GRCh38
rs562070319 Chr.6: 107028299-107028299 on Build GRCh38
rs901789245 Chr.6: 107028559-107028559 on Build GRCh38
rs965902871 Chr.6: 107028280-107028280 on Build GRCh38
rs892794982 Chr.6: 107028604-107028604 on Build GRCh38
rs181460111 Chr.6: 107028695-107028695 on Build GRCh38
rs782179613 Chr.6: 107028530-107028530 on Build GRCh38
rs889161885 Chr.6: 107028490-107028490 on Build GRCh38
rs548455657 Chr.6: 107028278-107028278 on Build GRCh38
rs1036382652 Chr.6: 107028411-107028411 on Build GRCh38
rs535880549 Chr.6: 107028462-107028462 on Build GRCh38
rs972302243 Chr.6: 107028708-107028708 on Build GRCh38
rs998707760 Chr.6: 107028568-107028568 on Build GRCh38
rs782574214 Chr.6: 107028267-107028267 on Build GRCh38
rs1031277447 Chr.6: 107028296-107028296 on Build GRCh38

View Full Product Details