Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 107066953-107067462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND3
Entrez Gene ID
57673
Gene Name
BEN domain containing 3
Gene Aliases
KIAA1553
UniGene
Hs.418045
Gene Chromosome Location
Chr.6: 107065180-107115187 on Build GRCh38
Transcripts
NM_001080450.2 XM_005267079.3 XM_005267080.3 XM_011536005.2 XM_017011138.1

SNP Detail

SNP ID Chromosome Location
rs1053989667 Chr.6: 107067259-107067259 on Build GRCh38
rs1027588108 Chr.6: 107067407-107067407 on Build GRCh38
rs1003563696 Chr.6: 107066983-107066983 on Build GRCh38
rs935595700 Chr.6: 107067254-107067254 on Build GRCh38
rs60916888 Chr.6: 107067178-107067178 on Build GRCh38
rs924306631 Chr.6: 107067250-107067250 on Build GRCh38
rs1003919552 Chr.6: 107067360-107067360 on Build GRCh38
rs568644323 Chr.6: 107067095-107067095 on Build GRCh38
rs985967599 Chr.6: 107067439-107067439 on Build GRCh38
rs538108129 Chr.6: 107067395-107067395 on Build GRCh38
rs1015215972 Chr.6: 107066994-107066994 on Build GRCh38
rs1014916353 Chr.6: 107067388-107067388 on Build GRCh38
rs920421396 Chr.6: 107067140-107067140 on Build GRCh38
rs911665395 Chr.6: 107067185-107067185 on Build GRCh38
rs58643495 Chr.6: 107067205-107067205 on Build GRCh38
rs782809630 Chr.6: 107067368-107067368 on Build GRCh38
rs906629807 Chr.6: 107067357-107067357 on Build GRCh38
rs893964345 Chr.6: 107067278-107067278 on Build GRCh38
rs546646026 Chr.6: 107067199-107067199 on Build GRCh38
rs114085529 Chr.6: 107067332-107067332 on Build GRCh38
rs55817462 Chr.6: 107067392-107067392 on Build GRCh38
rs56261788 Chr.6: 107067246-107067246 on Build GRCh38
rs116772721 Chr.6: 107067124-107067124 on Build GRCh38
rs3749872 Chr.6: 107067300-107067300 on Build GRCh38
rs548477662 Chr.6: 107067038-107067038 on Build GRCh38
rs985851495 Chr.6: 107067181-107067181 on Build GRCh38
rs78361125 Chr.6: 107067419-107067419 on Build GRCh38
rs994744828 Chr.6: 107067397-107067397 on Build GRCh38

View Full Product Details