Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 108894037-108894585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC2
Entrez Gene ID
84071
Gene Name
armadillo repeat containing 2
Gene Aliases
bA787I22.1
UniGene
Hs.645481
Gene Chromosome Location
Chr.6: 108848410-108990738 on Build GRCh38
Transcripts
NM_001286609.1 NM_032131.5 XM_005267154.3 XM_005267155.2 XM_005267157.3 XM_006715573.2 XM_006715574.2 XM_011536166.1 XM_011536167.1 XM_011536168.2 XM_011536170.2 XM_011536172.2 XM_011536176.2 XM_017011350.1 XM_017011351.1 XM_017011352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604877 c.684G>T p.K228N Chr.6: 108894479-108894479 on Build GRCh38
3674513 c.672-1G>T p.? Chr.6: 108894466-108894466 on Build GRCh38
7157848 c.686G>T p.R229I Chr.6: 108894481-108894481 on Build GRCh38
1487099 c.692C>T p.A231V Chr.6: 108894487-108894487 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757674902 Chr.6: 108894470-108894470 on Build GRCh38
rs1055237739 Chr.6: 108894472-108894472 on Build GRCh38
rs555141689 Chr.6: 108894440-108894440 on Build GRCh38
rs1010952333 Chr.6: 108894488-108894488 on Build GRCh38
rs761877694 Chr.6: 108894500-108894500 on Build GRCh38
rs746816457 Chr.6: 108894489-108894489 on Build GRCh38
rs545135440 Chr.6: 108894388-108894388 on Build GRCh38
rs954589837 Chr.6: 108894354-108894354 on Build GRCh38
rs1034184621 Chr.6: 108894207-108894207 on Build GRCh38
rs765375919 Chr.6: 108894235-108894235 on Build GRCh38
rs530314127 Chr.6: 108894119-108894119 on Build GRCh38
rs1024116317 Chr.6: 108894264-108894264 on Build GRCh38
rs546062110 Chr.6: 108894504-108894504 on Build GRCh38
rs745314436 Chr.6: 108894485-108894485 on Build GRCh38
rs370951932 Chr.6: 108894536-108894536 on Build GRCh38
rs991746992 Chr.6: 108894253-108894253 on Build GRCh38
rs78851917 Chr.6: 108894363-108894363 on Build GRCh38
rs149415473 Chr.6: 108894077-108894077 on Build GRCh38
rs532698741 Chr.6: 108894269-108894269 on Build GRCh38
rs533794667 Chr.6: 108894459-108894459 on Build GRCh38
rs761754184 Chr.6: 108894206-108894206 on Build GRCh38
rs1010837966 Chr.6: 108894285-108894285 on Build GRCh38
rs757804284 Chr.6: 108894555-108894555 on Build GRCh38
rs1048006684 Chr.6: 108894442-108894445 on Build GRCh38
rs929542745 Chr.6: 108894396-108894396 on Build GRCh38
rs567056927 Chr.6: 108894228-108894228 on Build GRCh38
rs769592319 Chr.6: 108894506-108894506 on Build GRCh38
rs546855572 Chr.6: 108894141-108894141 on Build GRCh38
rs117631573 Chr.6: 108894356-108894356 on Build GRCh38
rs767785950 Chr.6: 108894521-108894521 on Build GRCh38
rs929578520 Chr.6: 108894169-108894169 on Build GRCh38
rs918935571 Chr.6: 108894378-108894378 on Build GRCh38
rs941703771 Chr.6: 108894128-108894128 on Build GRCh38
rs756403058 Chr.6: 108894458-108894458 on Build GRCh38
rs993733256 Chr.6: 108894165-108894165 on Build GRCh38
rs774508939 Chr.6: 108894507-108894507 on Build GRCh38
rs923136699 Chr.6: 108894081-108894081 on Build GRCh38
rs767583802 Chr.6: 108894447-108894447 on Build GRCh38
rs938222960 Chr.6: 108894463-108894463 on Build GRCh38
rs144832569 Chr.6: 108894086-108894086 on Build GRCh38
rs766633924 Chr.6: 108894381-108894381 on Build GRCh38
rs910272527 Chr.6: 108894357-108894357 on Build GRCh38
rs901997850 Chr.6: 108894415-108894415 on Build GRCh38
rs994362789 Chr.6: 108894367-108894367 on Build GRCh38
rs186239570 Chr.6: 108894439-108894439 on Build GRCh38
rs200206972 Chr.6: 108894487-108894487 on Build GRCh38
rs144172293 Chr.6: 108894361-108894361 on Build GRCh38
rs202205491 Chr.6: 108894542-108894542 on Build GRCh38
rs761832688 Chr.6: 108894420-108894420 on Build GRCh38
rs753089434 Chr.6: 108894452-108894452 on Build GRCh38
rs763145639 Chr.6: 108894315-108894315 on Build GRCh38
rs759795133 Chr.6: 108894558-108894559 on Build GRCh38
rs750530861 Chr.6: 108894259-108894259 on Build GRCh38
rs887771058 Chr.6: 108894179-108894179 on Build GRCh38
rs889329447 Chr.6: 108894085-108894085 on Build GRCh38
rs1040966032 Chr.6: 108894387-108894387 on Build GRCh38
rs575311737 Chr.6: 108894558-108894558 on Build GRCh38
rs112110455 Chr.6: 108894462-108894462 on Build GRCh38
rs776568087 Chr.6: 108894497-108894497 on Build GRCh38
rs1026348574 Chr.6: 108894178-108894178 on Build GRCh38
rs768633566 Chr.6: 108894494-108894494 on Build GRCh38
rs985936270 Chr.6: 108894110-108894110 on Build GRCh38
rs573307546 Chr.6: 108894156-108894156 on Build GRCh38
rs1024586337 Chr.6: 108894492-108894492 on Build GRCh38
rs906736833 Chr.6: 108894212-108894212 on Build GRCh38
rs766543042 Chr.6: 108894425-108894425 on Build GRCh38
rs140225526 Chr.6: 108894334-108894334 on Build GRCh38
rs967233019 Chr.6: 108894323-108894323 on Build GRCh38
rs752860322 Chr.6: 108894530-108894530 on Build GRCh38
rs977399143 Chr.6: 108894325-108894325 on Build GRCh38
rs778535144 Chr.6: 108894477-108894477 on Build GRCh38
rs753568161 Chr.6: 108894511-108894511 on Build GRCh38

View Full Product Details