Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 108904142-108904639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC2
Entrez Gene ID
84071
Gene Name
armadillo repeat containing 2
Gene Aliases
bA787I22.1
UniGene
Hs.645481
Gene Chromosome Location
Chr.6: 108848410-108990738 on Build GRCh38
Transcripts
NM_001286609.1 NM_032131.5 XM_005267154.3 XM_005267155.2 XM_005267157.3 XM_006715573.2 XM_006715574.2 XM_011536166.1 XM_011536167.1 XM_011536168.2 XM_011536170.2 XM_011536172.2 XM_011536176.2 XM_017011350.1 XM_017011351.1 XM_017011352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183617 c.929T>G p.F310C Chr.6: 108904311-108904311 on Build GRCh38
6869699 c.977T>C p.V326A Chr.6: 108904359-108904359 on Build GRCh38
3619336 c.934G>A p.G312R Chr.6: 108904316-108904316 on Build GRCh38
3619337 c.948C>T p.L316L Chr.6: 108904330-108904330 on Build GRCh38
739728 c.907G>A p.G303R Chr.6: 108904289-108904289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772610401 Chr.6: 108904373-108904373 on Build GRCh38
rs578078508 Chr.6: 108904243-108904243 on Build GRCh38
rs201982059 Chr.6: 108904428-108904428 on Build GRCh38
rs771481529 Chr.6: 108904359-108904359 on Build GRCh38
rs200362565 Chr.6: 108904244-108904244 on Build GRCh38
rs1039764529 Chr.6: 108904314-108904314 on Build GRCh38
rs753579426 Chr.6: 108904286-108904286 on Build GRCh38
rs761448249 Chr.6: 108904423-108904423 on Build GRCh38
rs748206573 Chr.6: 108904342-108904342 on Build GRCh38
rs951960569 Chr.6: 108904403-108904403 on Build GRCh38
rs573569816 Chr.6: 108904523-108904523 on Build GRCh38
rs370148088 Chr.6: 108904438-108904438 on Build GRCh38
rs1008305354 Chr.6: 108904274-108904274 on Build GRCh38
rs542991297 Chr.6: 108904506-108904506 on Build GRCh38
rs555801026 Chr.6: 108904211-108904211 on Build GRCh38
rs753545482 Chr.6: 108904421-108904421 on Build GRCh38
rs1029281428 Chr.6: 108904524-108904524 on Build GRCh38
rs774853314 Chr.6: 108904360-108904360 on Build GRCh38
rs774943473 Chr.6: 108904221-108904221 on Build GRCh38
rs760109055 Chr.6: 108904229-108904229 on Build GRCh38
rs375446603 Chr.6: 108904349-108904349 on Build GRCh38
rs756987265 Chr.6: 108904295-108904295 on Build GRCh38
rs752222007 Chr.6: 108904245-108904245 on Build GRCh38
rs142255179 Chr.6: 108904481-108904481 on Build GRCh38
rs542877021 Chr.6: 108904576-108904576 on Build GRCh38
rs773476059 Chr.6: 108904202-108904202 on Build GRCh38
rs750192654 Chr.6: 108904433-108904433 on Build GRCh38
rs749686178 Chr.6: 108904356-108904356 on Build GRCh38
rs892681486 Chr.6: 108904455-108904455 on Build GRCh38
rs189948384 Chr.6: 108904203-108904203 on Build GRCh38
rs771319031 Chr.6: 108904467-108904467 on Build GRCh38
rs939476320 Chr.6: 108904437-108904437 on Build GRCh38
rs781175381 Chr.6: 108904340-108904340 on Build GRCh38
rs954573623 Chr.6: 108904598-108904598 on Build GRCh38
rs758297619 Chr.6: 108904306-108904306 on Build GRCh38
rs1014696852 Chr.6: 108904464-108904464 on Build GRCh38
rs556571022 Chr.6: 108904510-108904510 on Build GRCh38
rs747017427 Chr.6: 108904369-108904369 on Build GRCh38
rs777987476 Chr.6: 108904351-108904351 on Build GRCh38
rs145604834 Chr.6: 108904400-108904400 on Build GRCh38
rs559388102 Chr.6: 108904590-108904590 on Build GRCh38
rs772593059 Chr.6: 108904207-108904208 on Build GRCh38
rs778664748 Chr.6: 108904297-108904297 on Build GRCh38
rs572573180 Chr.6: 108904233-108904233 on Build GRCh38
rs771573134 Chr.6: 108904214-108904214 on Build GRCh38
rs1052709305 Chr.6: 108904448-108904448 on Build GRCh38
rs760295248 Chr.6: 108904276-108904276 on Build GRCh38
rs771615568 Chr.6: 108904196-108904199 on Build GRCh38
rs770031498 Chr.6: 108904186-108904186 on Build GRCh38
rs750425400 Chr.6: 108904301-108904301 on Build GRCh38
rs760161977 Chr.6: 108904371-108904371 on Build GRCh38
rs765053879 Chr.6: 108904426-108904426 on Build GRCh38
rs368679434 Chr.6: 108904504-108904504 on Build GRCh38
rs570342634 Chr.6: 108904167-108904167 on Build GRCh38
rs758350463 Chr.6: 108904436-108904436 on Build GRCh38
rs774893696 Chr.6: 108904384-108904384 on Build GRCh38

View Full Product Details