Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109382420-109382961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD164
Entrez Gene ID
8763
Gene Name
CD164 molecule
Gene Aliases
DFNA66, MGC-24, MUC-24, endolyn
UniGene
Hs.520313
Gene Chromosome Location
Chr.6: 109366514-109382559 on Build GRCh38
Transcripts
NM_001142401.1 NM_001142402.1 NM_001142403.1 NM_001142404.1 NM_006016.4

SNP Detail

SNP ID Chromosome Location
rs918116734 Chr.6: 109382744-109382744 on Build GRCh38
rs568404449 Chr.6: 109382601-109382601 on Build GRCh38
rs763192498 Chr.6: 109382484-109382484 on Build GRCh38
rs556830431 Chr.6: 109382703-109382703 on Build GRCh38
rs953431787 Chr.6: 109382499-109382499 on Build GRCh38
rs1044723148 Chr.6: 109382467-109382467 on Build GRCh38
rs996765973 Chr.6: 109382846-109382846 on Build GRCh38
rs187922169 Chr.6: 109382582-109382582 on Build GRCh38
rs541603966 Chr.6: 109382773-109382773 on Build GRCh38
rs936306699 Chr.6: 109382783-109382783 on Build GRCh38
rs976463719 Chr.6: 109382493-109382493 on Build GRCh38
rs753246784 Chr.6: 109382669-109382669 on Build GRCh38
rs563385273 Chr.6: 109382897-109382897 on Build GRCh38
rs568008924 Chr.6: 109382777-109382777 on Build GRCh38
rs534208878 Chr.6: 109382607-109382607 on Build GRCh38
rs1032127383 Chr.6: 109382471-109382471 on Build GRCh38
rs562784646 Chr.6: 109382766-109382766 on Build GRCh38
rs901042508 Chr.6: 109382834-109382834 on Build GRCh38
rs949038324 Chr.6: 109382455-109382455 on Build GRCh38
rs909379725 Chr.6: 109382519-109382519 on Build GRCh38
rs554162795 Chr.6: 109382623-109382623 on Build GRCh38
rs1029020021 Chr.6: 109382494-109382494 on Build GRCh38
rs984908552 Chr.6: 109382518-109382518 on Build GRCh38
rs953399262 Chr.6: 109382696-109382696 on Build GRCh38
rs1020540695 Chr.6: 109382657-109382657 on Build GRCh38
rs1028991819 Chr.6: 109382673-109382673 on Build GRCh38
rs949681682 Chr.6: 109382571-109382571 on Build GRCh38
rs115705232 Chr.6: 109382888-109382888 on Build GRCh38
rs962165929 Chr.6: 109382738-109382738 on Build GRCh38
rs1006198060 Chr.6: 109382865-109382865 on Build GRCh38
rs940818222 Chr.6: 109382520-109382520 on Build GRCh38
rs114906276 Chr.6: 109382823-109382823 on Build GRCh38
rs3757227 Chr.6: 109382806-109382806 on Build GRCh38
rs981028226 Chr.6: 109382771-109382771 on Build GRCh38
rs1016284646 Chr.6: 109382867-109382867 on Build GRCh38
rs972267334 Chr.6: 109382522-109382522 on Build GRCh38
rs142381670 Chr.6: 109382924-109382924 on Build GRCh38
rs1048359680 Chr.6: 109382793-109382793 on Build GRCh38
rs960249115 Chr.6: 109382459-109382459 on Build GRCh38
rs549583447 Chr.6: 109382564-109382564 on Build GRCh38
rs2275652 Chr.6: 109382480-109382480 on Build GRCh38
rs879528061 Chr.6: 109382804-109382804 on Build GRCh38
rs546176364 Chr.6: 109382573-109382573 on Build GRCh38
rs751586433 Chr.6: 109382920-109382920 on Build GRCh38
rs17070543 Chr.6: 109382642-109382642 on Build GRCh38
rs945167448 Chr.6: 109382821-109382821 on Build GRCh38
rs897778321 Chr.6: 109382571-109382571 on Build GRCh38
rs1322816 Chr.6: 109382524-109382524 on Build GRCh38
rs989085177 Chr.6: 109382928-109382928 on Build GRCh38
rs1053396507 Chr.6: 109382797-109382797 on Build GRCh38
rs114769624 Chr.6: 109382583-109382583 on Build GRCh38
rs892331772 Chr.6: 109382475-109382475 on Build GRCh38
rs986304727 Chr.6: 109382739-109382739 on Build GRCh38
rs980913667 Chr.6: 109382903-109382903 on Build GRCh38
rs192277119 Chr.6: 109382838-109382838 on Build GRCh38
rs545636913 Chr.6: 109382797-109382798 on Build GRCh38
rs541892621 Chr.6: 109382441-109382441 on Build GRCh38
rs892222944 Chr.6: 109382625-109382625 on Build GRCh38
rs1006230907 Chr.6: 109382711-109382711 on Build GRCh38
rs1049636442 Chr.6: 109382847-109382847 on Build GRCh38
rs985867430 Chr.6: 109382915-109382915 on Build GRCh38
rs962420524 Chr.6: 109382872-109382872 on Build GRCh38
rs965789299 Chr.6: 109382932-109382932 on Build GRCh38
rs1009783936 Chr.6: 109382646-109382646 on Build GRCh38
rs577635666 Chr.6: 109382645-109382645 on Build GRCh38
rs970829746 Chr.6: 109382898-109382898 on Build GRCh38
rs958298113 Chr.6: 109382923-109382923 on Build GRCh38
rs1322815 Chr.6: 109382859-109382859 on Build GRCh38
rs375538983 Chr.6: 109382446-109382446 on Build GRCh38
rs3757229 Chr.6: 109382935-109382935 on Build GRCh38
rs1036242074 Chr.6: 109382548-109382548 on Build GRCh38
rs1016729567 Chr.6: 109382729-109382729 on Build GRCh38
rs115225098 Chr.6: 109382665-109382665 on Build GRCh38
rs1014429272 Chr.6: 109382829-109382829 on Build GRCh38
rs972618721 Chr.6: 109382740-109382740 on Build GRCh38
rs904929525 Chr.6: 109382468-109382468 on Build GRCh38
rs547325750 Chr.6: 109382476-109382476 on Build GRCh38
rs770659531 Chr.6: 109382473-109382473 on Build GRCh38
rs913594880 Chr.6: 109382933-109382933 on Build GRCh38
rs1034499956 Chr.6: 109382616-109382616 on Build GRCh38
rs183043432 Chr.6: 109382452-109382452 on Build GRCh38
rs1000594950 Chr.6: 109382470-109382470 on Build GRCh38
rs1025560612 Chr.6: 109382892-109382892 on Build GRCh38
rs1002037318 Chr.6: 109382586-109382586 on Build GRCh38
rs978870202 Chr.6: 109382477-109382477 on Build GRCh38
rs926848354 Chr.6: 109382908-109382908 on Build GRCh38
rs531605550 Chr.6: 109382810-109382810 on Build GRCh38
rs1019459273 Chr.6: 109382659-109382659 on Build GRCh38
rs373139537 Chr.6: 109382790-109382790 on Build GRCh38

View Full Product Details