Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 110404901-110405443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDO
Entrez Gene ID
8528
Gene Name
D-aspartate oxidase
Gene Aliases
DASOX, DDO-1, DDO-2
UniGene
Hs.591348
Gene Chromosome Location
Chr.6: 110392180-110415550 on Build GRCh38
Transcripts
NM_003649.2 NM_004032.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5996625 c.413C>T p.A138V Chr.6: 110404903-110404903 on Build GRCh38
32231 c.408C>G p.F136L Chr.6: 110404908-110404908 on Build GRCh38
6641363 c.393T>C p.T131T Chr.6: 110404923-110404923 on Build GRCh38
345787 c.387T>G p.T129T Chr.6: 110404929-110404929 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs903243912 Chr.6: 110405217-110405217 on Build GRCh38
rs1015337209 Chr.6: 110405375-110405375 on Build GRCh38
rs1035820324 Chr.6: 110405357-110405357 on Build GRCh38
rs553619500 Chr.6: 110405174-110405174 on Build GRCh38
rs145481637 Chr.6: 110405402-110405402 on Build GRCh38
rs112243388 Chr.6: 110405182-110405182 on Build GRCh38
rs973568035 Chr.6: 110405022-110405022 on Build GRCh38
rs146381499 Chr.6: 110405230-110405230 on Build GRCh38
rs938993665 Chr.6: 110405228-110405228 on Build GRCh38
rs4945858 Chr.6: 110405181-110405181 on Build GRCh38
rs973473657 Chr.6: 110405401-110405401 on Build GRCh38
rs761693931 Chr.6: 110404970-110404970 on Build GRCh38
rs9320329 Chr.6: 110405331-110405331 on Build GRCh38
rs954881443 Chr.6: 110405346-110405346 on Build GRCh38
rs1003719901 Chr.6: 110404951-110404951 on Build GRCh38
rs763495971 Chr.6: 110405244-110405244 on Build GRCh38
rs992160540 Chr.6: 110405407-110405407 on Build GRCh38
rs753761875 Chr.6: 110404971-110404972 on Build GRCh38
rs554626471 Chr.6: 110405279-110405279 on Build GRCh38
rs775410803 Chr.6: 110404927-110404927 on Build GRCh38
rs963193633 Chr.6: 110405013-110405013 on Build GRCh38
rs1008933955 Chr.6: 110405355-110405355 on Build GRCh38
rs552430235 Chr.6: 110404948-110404948 on Build GRCh38
rs560460465 Chr.6: 110405356-110405356 on Build GRCh38
rs191588399 Chr.6: 110405409-110405409 on Build GRCh38
rs536908648 Chr.6: 110405139-110405139 on Build GRCh38
rs962176392 Chr.6: 110405387-110405387 on Build GRCh38
rs199869693 Chr.6: 110405267-110405267 on Build GRCh38
rs142158644 Chr.6: 110404958-110404958 on Build GRCh38
rs4945859 Chr.6: 110405197-110405197 on Build GRCh38
rs974951819 Chr.6: 110405064-110405064 on Build GRCh38
rs768068000 Chr.6: 110404971-110404971 on Build GRCh38
rs968964622 Chr.6: 110405051-110405051 on Build GRCh38
rs558744426 Chr.6: 110405225-110405225 on Build GRCh38
rs746791254 Chr.6: 110404957-110404957 on Build GRCh38
rs1028962956 Chr.6: 110405102-110405102 on Build GRCh38
rs564881879 Chr.6: 110405397-110405397 on Build GRCh38
rs754816228 Chr.6: 110404972-110404972 on Build GRCh38
rs1051578215 Chr.6: 110405226-110405226 on Build GRCh38
rs548139973 Chr.6: 110405108-110405108 on Build GRCh38
rs777614368 Chr.6: 110405252-110405252 on Build GRCh38
rs760789679 Chr.6: 110405036-110405036 on Build GRCh38
rs554909782 Chr.6: 110405290-110405290 on Build GRCh38
rs17492667 Chr.6: 110405033-110405033 on Build GRCh38
rs376984169 Chr.6: 110404966-110404966 on Build GRCh38
rs890365033 Chr.6: 110404938-110404938 on Build GRCh38
rs149666172 Chr.6: 110405300-110405300 on Build GRCh38
rs745572196 Chr.6: 110404926-110404926 on Build GRCh38
rs982882943 Chr.6: 110405141-110405141 on Build GRCh38
rs766499120 Chr.6: 110404979-110404979 on Build GRCh38
rs1007473048 Chr.6: 110405250-110405250 on Build GRCh38
rs928921643 Chr.6: 110405159-110405159 on Build GRCh38
rs1043537954 Chr.6: 110405209-110405209 on Build GRCh38
rs534954949 Chr.6: 110405162-110405162 on Build GRCh38
rs368729360 Chr.6: 110404983-110404983 on Build GRCh38
rs116840869 Chr.6: 110405109-110405109 on Build GRCh38
rs576168948 Chr.6: 110405307-110405307 on Build GRCh38

View Full Product Details