Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 110888716-110889216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMD1
Entrez Gene ID
262
Gene Name
adenosylmethionine decarboxylase 1
Gene Aliases
ADOMETDC, AMD, SAMDC
UniGene
Hs.159118
Gene Chromosome Location
Chr.6: 110814621-110895713 on Build GRCh38
Transcripts
NM_001287214.1 NM_001287215.1 NM_001287216.1 NM_001634.5 NR_109768.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3857514 c.275T>C p.V92A Chr.6: 110888934-110888934 on Build GRCh38
4484345 c.279C>T p.P93P Chr.6: 110888938-110888938 on Build GRCh38
3374015 c.203G>T p.S68I Chr.6: 110888862-110888862 on Build GRCh38
5982143 c.261G>A p.L87L Chr.6: 110888920-110888920 on Build GRCh38
1167620 c.277delC p.L94fs*2 Chr.6: 110888936-110888936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377055532 Chr.6: 110889029-110889029 on Build GRCh38
rs906906106 Chr.6: 110889049-110889049 on Build GRCh38
rs1055315207 Chr.6: 110888963-110888963 on Build GRCh38
rs758290283 Chr.6: 110888817-110888817 on Build GRCh38
rs761384558 Chr.6: 110888999-110888999 on Build GRCh38
rs763691495 Chr.6: 110888993-110888993 on Build GRCh38
rs749478302 Chr.6: 110888893-110888893 on Build GRCh38
rs78693625 Chr.6: 110888847-110888847 on Build GRCh38
rs976082040 Chr.6: 110888746-110888746 on Build GRCh38
rs545422062 Chr.6: 110889052-110889052 on Build GRCh38
rs771129424 Chr.6: 110888908-110888908 on Build GRCh38
rs778575506 Chr.6: 110888806-110888806 on Build GRCh38
rs769987168 Chr.6: 110889057-110889062 on Build GRCh38
rs779004856 Chr.6: 110888948-110888948 on Build GRCh38
rs778037570 Chr.6: 110888885-110888885 on Build GRCh38
rs199938532 Chr.6: 110888816-110888816 on Build GRCh38
rs752350206 Chr.6: 110888770-110888770 on Build GRCh38
rs932053641 Chr.6: 110888786-110888786 on Build GRCh38
rs760317426 Chr.6: 110888983-110888983 on Build GRCh38
rs374684466 Chr.6: 110888917-110888917 on Build GRCh38
rs544535579 Chr.6: 110889117-110889117 on Build GRCh38
rs892930015 Chr.6: 110889089-110889089 on Build GRCh38
rs776213563 Chr.6: 110888997-110888997 on Build GRCh38
rs769852651 Chr.6: 110888851-110888851 on Build GRCh38
rs772742842 Chr.6: 110888953-110888953 on Build GRCh38
rs201052912 Chr.6: 110889012-110889012 on Build GRCh38
rs9400424 Chr.6: 110889073-110889073 on Build GRCh38
rs929625284 Chr.6: 110888766-110888766 on Build GRCh38
rs56171216 Chr.6: 110889048-110889048 on Build GRCh38
rs976033261 Chr.6: 110889163-110889163 on Build GRCh38
rs776115161 Chr.6: 110888977-110888977 on Build GRCh38
rs748276856 Chr.6: 110888839-110888839 on Build GRCh38
rs942245961 Chr.6: 110888826-110888826 on Build GRCh38
rs766363523 Chr.6: 110889028-110889028 on Build GRCh38
rs779936538 Chr.6: 110888830-110888830 on Build GRCh38
rs138120520 Chr.6: 110889128-110889128 on Build GRCh38
rs762803486 Chr.6: 110889035-110889035 on Build GRCh38
rs1032730127 Chr.6: 110889100-110889100 on Build GRCh38
rs375551851 Chr.6: 110889060-110889060 on Build GRCh38
rs113764512 Chr.6: 110888837-110888837 on Build GRCh38
rs568116120 Chr.6: 110888785-110888785 on Build GRCh38
rs758190870 Chr.6: 110889027-110889027 on Build GRCh38
rs530202399 Chr.6: 110889141-110889141 on Build GRCh38
rs750226507 Chr.6: 110889017-110889017 on Build GRCh38
rs909354044 Chr.6: 110888962-110888962 on Build GRCh38
rs746497106 Chr.6: 110889071-110889072 on Build GRCh38
rs746199629 Chr.6: 110888938-110888938 on Build GRCh38
rs923319129 Chr.6: 110888749-110888749 on Build GRCh38
rs1030747154 Chr.6: 110889186-110889186 on Build GRCh38
rs111749328 Chr.6: 110888815-110888815 on Build GRCh38
rs771566127 Chr.6: 110888830-110888831 on Build GRCh38
rs772097645 Chr.6: 110889119-110889119 on Build GRCh38
rs754846514 Chr.6: 110889032-110889032 on Build GRCh38

View Full Product Details