Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 110889982-110890489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMD1
Entrez Gene ID
262
Gene Name
adenosylmethionine decarboxylase 1
Gene Aliases
ADOMETDC, AMD, SAMDC
UniGene
Hs.159118
Gene Chromosome Location
Chr.6: 110814621-110895713 on Build GRCh38
Transcripts
NM_001287214.1 NM_001287215.1 NM_001287216.1 NM_001634.5 NR_109768.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3357932 c.340C>T p.R114C Chr.6: 110890269-110890269 on Build GRCh38
4488310 c.330C>T p.F110F Chr.6: 110890259-110890259 on Build GRCh38
3155929 c.402_403delAG p.E135fs*3 Chr.6: 110890331-110890332 on Build GRCh38
4847278 c.325-1G>C p.? Chr.6: 110890253-110890253 on Build GRCh38
6599065 c.427A>C p.N143H Chr.6: 110890356-110890356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757455993 Chr.6: 110890225-110890225 on Build GRCh38
rs1011512501 Chr.6: 110890409-110890409 on Build GRCh38
rs1022568103 Chr.6: 110890075-110890075 on Build GRCh38
rs1023933661 Chr.6: 110890012-110890015 on Build GRCh38
rs534455889 Chr.6: 110890231-110890231 on Build GRCh38
rs569645624 Chr.6: 110890095-110890095 on Build GRCh38
rs879023762 Chr.6: 110890325-110890325 on Build GRCh38
rs150093572 Chr.6: 110890293-110890293 on Build GRCh38
rs116158848 Chr.6: 110890221-110890221 on Build GRCh38
rs1031652791 Chr.6: 110890014-110890021 on Build GRCh38
rs765390945 Chr.6: 110890232-110890236 on Build GRCh38
rs1049701 Chr.6: 110890312-110890312 on Build GRCh38
rs779159812 Chr.6: 110890228-110890228 on Build GRCh38
rs749342824 Chr.6: 110890223-110890223 on Build GRCh38
rs757333872 Chr.6: 110890334-110890334 on Build GRCh38
rs148584379 Chr.6: 110890112-110890112 on Build GRCh38
rs541903808 Chr.6: 110890402-110890402 on Build GRCh38
rs554410125 Chr.6: 110890234-110890234 on Build GRCh38
rs770753047 Chr.6: 110890221-110890221 on Build GRCh38
rs933974930 Chr.6: 110890071-110890071 on Build GRCh38
rs781708932 Chr.6: 110890400-110890400 on Build GRCh38
rs570940378 Chr.6: 110890073-110890073 on Build GRCh38
rs369005588 Chr.6: 110890256-110890256 on Build GRCh38
rs959833296 Chr.6: 110890055-110890055 on Build GRCh38
rs1038616564 Chr.6: 110890162-110890162 on Build GRCh38
rs766168455 Chr.6: 110890309-110890309 on Build GRCh38
rs769318059 Chr.6: 110890393-110890393 on Build GRCh38
rs555342394 Chr.6: 110890152-110890152 on Build GRCh38
rs933076749 Chr.6: 110890161-110890161 on Build GRCh38
rs549434504 Chr.6: 110890088-110890088 on Build GRCh38
rs571692334 Chr.6: 110890015-110890016 on Build GRCh38
rs761345324 Chr.6: 110890247-110890247 on Build GRCh38
rs906580798 Chr.6: 110890343-110890343 on Build GRCh38
rs746263900 Chr.6: 110890230-110890230 on Build GRCh38
rs776555940 Chr.6: 110890108-110890108 on Build GRCh38
rs73534864 Chr.6: 110890040-110890040 on Build GRCh38
rs1031305182 Chr.6: 110890397-110890397 on Build GRCh38
rs975459162 Chr.6: 110890042-110890042 on Build GRCh38
rs562875601 Chr.6: 110890330-110890330 on Build GRCh38
rs772906873 Chr.6: 110890270-110890270 on Build GRCh38
rs746121282 Chr.6: 110890173-110890173 on Build GRCh38
rs115615189 Chr.6: 110890047-110890047 on Build GRCh38
rs777846535 Chr.6: 110890219-110890219 on Build GRCh38
rs746096718 Chr.6: 110890356-110890356 on Build GRCh38
rs769343821 Chr.6: 110890406-110890406 on Build GRCh38
rs143903348 Chr.6: 110890014-110890015 on Build GRCh38
rs187960765 Chr.6: 110890013-110890013 on Build GRCh38
rs779059721 Chr.6: 110890346-110890346 on Build GRCh38
rs759777544 Chr.6: 110890222-110890222 on Build GRCh38
rs943209778 Chr.6: 110890189-110890189 on Build GRCh38
rs775555558 Chr.6: 110890385-110890387 on Build GRCh38
rs993477097 Chr.6: 110890007-110890007 on Build GRCh38
rs759421502 Chr.6: 110890375-110890375 on Build GRCh38
rs776026537 Chr.6: 110890246-110890246 on Build GRCh38
rs751299857 Chr.6: 110890310-110890310 on Build GRCh38
rs1049699 Chr.6: 110890311-110890311 on Build GRCh38
rs1022516088 Chr.6: 110890461-110890461 on Build GRCh38
rs887039941 Chr.6: 110890096-110890096 on Build GRCh38
rs764880452 Chr.6: 110890443-110890443 on Build GRCh38
rs964196040 Chr.6: 110890458-110890458 on Build GRCh38
rs989709529 Chr.6: 110890431-110890431 on Build GRCh38
rs767406460 Chr.6: 110890329-110890329 on Build GRCh38
rs55804656 Chr.6: 110890165-110890165 on Build GRCh38
rs966960978 Chr.6: 110890462-110890462 on Build GRCh38
rs915388017 Chr.6: 110890036-110890036 on Build GRCh38
rs577487019 Chr.6: 110890235-110890235 on Build GRCh38
rs758473195 Chr.6: 110890379-110890379 on Build GRCh38
rs1034210942 Chr.6: 110890380-110890380 on Build GRCh38
rs201030223 Chr.6: 110890386-110890386 on Build GRCh38
rs527579017 Chr.6: 110890031-110890031 on Build GRCh38
rs1022891733 Chr.6: 110890442-110890442 on Build GRCh38
rs754933640 Chr.6: 110890316-110890316 on Build GRCh38
rs202160703 Chr.6: 110890240-110890240 on Build GRCh38
rs201408347 Chr.6: 110890221-110890222 on Build GRCh38

View Full Product Details