Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 876225-876761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GET4
Entrez Gene ID
51608
Gene Name
golgi to ER traffic protein 4
Gene Aliases
C7orf20, CEE, CGI-20, TRC35
UniGene
Hs.107387
Gene Chromosome Location
Chr.7: 876554-896434 on Build GRCh38
Transcripts
NM_015949.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5443347 c.25G>A p.E9K Chr.7: 876670-876670 on Build GRCh38
6334718 c.65G>A p.G22D Chr.7: 876710-876710 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs546300243 Chr.7: 876418-876418 on Build GRCh38
rs534242753 Chr.7: 876595-876595 on Build GRCh38
rs954410104 Chr.7: 876729-876729 on Build GRCh38
rs1033354347 Chr.7: 876507-876507 on Build GRCh38
rs941780049 Chr.7: 876428-876428 on Build GRCh38
rs950590644 Chr.7: 876669-876669 on Build GRCh38
rs956415588 Chr.7: 876246-876246 on Build GRCh38
rs922074505 Chr.7: 876496-876496 on Build GRCh38
rs866662575 Chr.7: 876338-876338 on Build GRCh38
rs371243966 Chr.7: 876601-876601 on Build GRCh38
rs574215696 Chr.7: 876642-876642 on Build GRCh38
rs543031316 Chr.7: 876653-876653 on Build GRCh38
rs1014864469 Chr.7: 876445-876445 on Build GRCh38
rs963339303 Chr.7: 876687-876687 on Build GRCh38
rs543891967 Chr.7: 876522-876537 on Build GRCh38
rs980869221 Chr.7: 876258-876258 on Build GRCh38
rs760179726 Chr.7: 876659-876659 on Build GRCh38
rs1012693401 Chr.7: 876651-876651 on Build GRCh38
rs922680961 Chr.7: 876585-876585 on Build GRCh38
rs1041128871 Chr.7: 876312-876312 on Build GRCh38
rs921651504 Chr.7: 876638-876638 on Build GRCh38
rs772047152 Chr.7: 876362-876391 on Build GRCh38
rs891954914 Chr.7: 876399-876399 on Build GRCh38
rs901285021 Chr.7: 876313-876313 on Build GRCh38
rs1024708521 Chr.7: 876448-876448 on Build GRCh38
rs970406875 Chr.7: 876253-876253 on Build GRCh38
rs765604974 Chr.7: 876672-876672 on Build GRCh38
rs939859202 Chr.7: 876524-876524 on Build GRCh38
rs1000288672 Chr.7: 876243-876243 on Build GRCh38
rs376978899 Chr.7: 876711-876711 on Build GRCh38
rs10265654 Chr.7: 876421-876421 on Build GRCh38
rs908378205 Chr.7: 876288-876288 on Build GRCh38
rs980405106 Chr.7: 876487-876487 on Build GRCh38
rs557926672 Chr.7: 876569-876569 on Build GRCh38
rs1051946828 Chr.7: 876506-876506 on Build GRCh38
rs754328990 Chr.7: 876666-876666 on Build GRCh38
rs145210818 Chr.7: 876738-876738 on Build GRCh38
rs778147329 Chr.7: 876699-876699 on Build GRCh38
rs939890503 Chr.7: 876290-876290 on Build GRCh38
rs749222913 Chr.7: 876727-876727 on Build GRCh38
rs756945817 Chr.7: 876279-876308 on Build GRCh38
rs987904555 Chr.7: 876249-876249 on Build GRCh38
rs1024162762 Chr.7: 876537-876537 on Build GRCh38
rs768512126 Chr.7: 876737-876737 on Build GRCh38
rs531949361 Chr.7: 876355-876355 on Build GRCh38
rs184893727 Chr.7: 876374-876374 on Build GRCh38
rs983116513 Chr.7: 876336-876336 on Build GRCh38
rs548596097 Chr.7: 876458-876458 on Build GRCh38
rs368942210 Chr.7: 876385-876385 on Build GRCh38
rs908494315 Chr.7: 876518-876518 on Build GRCh38
rs868405559 Chr.7: 876361-876361 on Build GRCh38
rs111793477 Chr.7: 876423-876423 on Build GRCh38
rs1035507594 Chr.7: 876598-876598 on Build GRCh38
rs963395248 Chr.7: 876623-876625 on Build GRCh38
rs908873780 Chr.7: 876613-876613 on Build GRCh38
rs969904009 Chr.7: 876280-876280 on Build GRCh38
rs569117511 Chr.7: 876464-876464 on Build GRCh38
rs1051893889 Chr.7: 876641-876641 on Build GRCh38
rs13310603 Chr.7: 876391-876391 on Build GRCh38
rs1022790648 Chr.7: 876272-876272 on Build GRCh38
rs952569178 Chr.7: 876508-876508 on Build GRCh38
rs917691830 Chr.7: 876250-876250 on Build GRCh38
rs10269286 Chr.7: 876368-876368 on Build GRCh38
rs991320475 Chr.7: 876265-876265 on Build GRCh38
rs10269500 Chr.7: 876504-876504 on Build GRCh38
rs747304707 Chr.7: 876702-876702 on Build GRCh38
rs904748080 Chr.7: 876363-876363 on Build GRCh38
rs879343845 Chr.7: 876513-876513 on Build GRCh38
rs563154856 Chr.7: 876264-876264 on Build GRCh38
rs1002635615 Chr.7: 876576-876576 on Build GRCh38
rs925957344 Chr.7: 876301-876301 on Build GRCh38
rs554332081 Chr.7: 876599-876599 on Build GRCh38
rs753777363 Chr.7: 876648-876650 on Build GRCh38
rs571354410 Chr.7: 876571-876571 on Build GRCh38
rs974503405 Chr.7: 876623-876623 on Build GRCh38
rs372402701 Chr.7: 876444-876444 on Build GRCh38
rs753026890 Chr.7: 876682-876682 on Build GRCh38
rs1045061620 Chr.7: 876662-876662 on Build GRCh38
rs931231726 Chr.7: 876285-876285 on Build GRCh38
rs111884158 Chr.7: 876505-876505 on Build GRCh38
rs977016541 Chr.7: 876565-876565 on Build GRCh38
rs781474745 Chr.7: 876717-876717 on Build GRCh38
rs111267126 Chr.7: 876453-876453 on Build GRCh38
rs1031807527 Chr.7: 876245-876245 on Build GRCh38
rs983806276 Chr.7: 876611-876611 on Build GRCh38
rs765938144 Chr.7: 876663-876663 on Build GRCh38
rs754977394 Chr.7: 876648-876653 on Build GRCh38
rs908876719 Chr.7: 876387-876387 on Build GRCh38
rs970901091 Chr.7: 876479-876479 on Build GRCh38
rs372879888 Chr.7: 876314-876314 on Build GRCh38
rs1039232331 Chr.7: 876456-876457 on Build GRCh38
rs758590114 Chr.7: 876698-876698 on Build GRCh38
rs565386205 Chr.7: 876367-876367 on Build GRCh38
rs545577161 Chr.7: 876366-876366 on Build GRCh38
rs935590620 Chr.7: 876647-876648 on Build GRCh38
rs1048224969 Chr.7: 876287-876287 on Build GRCh38
rs755486086 Chr.7: 876670-876670 on Build GRCh38

View Full Product Details