Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 964668-965180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX19
Entrez Gene ID
90639
Gene Name
COX19, cytochrome c oxidase assembly factor
Gene Aliases
-
UniGene
Hs.121593
Gene Chromosome Location
Chr.7: 964850-975599 on Build GRCh38
Transcripts
NM_001031617.2

SNP Detail

SNP ID Chromosome Location
rs573269638 Chr.7: 965045-965045 on Build GRCh38
rs919124213 Chr.7: 964721-964721 on Build GRCh38
rs1018361386 Chr.7: 964689-964689 on Build GRCh38
rs929496093 Chr.7: 964842-964842 on Build GRCh38
rs2397 Chr.7: 964887-964887 on Build GRCh38
rs532077832 Chr.7: 964696-964696 on Build GRCh38
rs915893322 Chr.7: 964992-964992 on Build GRCh38
rs890842752 Chr.7: 964892-964892 on Build GRCh38
rs947018768 Chr.7: 965113-965113 on Build GRCh38
rs555805945 Chr.7: 965096-965096 on Build GRCh38
rs778765336 Chr.7: 964805-964805 on Build GRCh38
rs930424953 Chr.7: 964722-964722 on Build GRCh38
rs1039343146 Chr.7: 964901-964901 on Build GRCh38
rs575034484 Chr.7: 965027-965027 on Build GRCh38
rs899525358 Chr.7: 964907-964907 on Build GRCh38
rs553532344 Chr.7: 965030-965030 on Build GRCh38
rs963415389 Chr.7: 965006-965006 on Build GRCh38
rs150546833 Chr.7: 964791-964791 on Build GRCh38
rs776604120 Chr.7: 964777-964777 on Build GRCh38
rs889008523 Chr.7: 964731-964731 on Build GRCh38
rs11769026 Chr.7: 964687-964687 on Build GRCh38
rs1042542767 Chr.7: 965019-965019 on Build GRCh38
rs929548268 Chr.7: 965062-965062 on Build GRCh38
rs751171789 Chr.7: 965081-965081 on Build GRCh38
rs775744591 Chr.7: 965112-965112 on Build GRCh38
rs1049016349 Chr.7: 964724-964724 on Build GRCh38
rs1037113585 Chr.7: 964695-964695 on Build GRCh38
rs780988785 Chr.7: 964778-964778 on Build GRCh38
rs184220847 Chr.7: 964977-964977 on Build GRCh38
rs75881071 Chr.7: 964960-964960 on Build GRCh38
rs758367502 Chr.7: 964816-964816 on Build GRCh38
rs1042706116 Chr.7: 965146-965146 on Build GRCh38
rs1027091009 Chr.7: 965070-965070 on Build GRCh38
rs915908566 Chr.7: 964809-964809 on Build GRCh38
rs1033910229 Chr.7: 964799-964799 on Build GRCh38
rs967223275 Chr.7: 964776-964776 on Build GRCh38
rs1040472586 Chr.7: 964734-964734 on Build GRCh38
rs1014295723 Chr.7: 965048-965048 on Build GRCh38
rs552859331 Chr.7: 964797-964797 on Build GRCh38
rs765145941 Chr.7: 964831-964831 on Build GRCh38
rs1025185697 Chr.7: 965058-965058 on Build GRCh38
rs895313000 Chr.7: 965046-965046 on Build GRCh38
rs551890281 Chr.7: 964754-964754 on Build GRCh38
rs754770567 Chr.7: 964771-964771 on Build GRCh38
rs575799648 Chr.7: 965149-965149 on Build GRCh38
rs928701649 Chr.7: 964796-964796 on Build GRCh38
rs1008412441 Chr.7: 964899-964899 on Build GRCh38
rs17227152 Chr.7: 964950-964950 on Build GRCh38
rs1021530093 Chr.7: 964923-964923 on Build GRCh38
rs1039898790 Chr.7: 965101-965101 on Build GRCh38
rs984600064 Chr.7: 964936-964936 on Build GRCh38
rs903083147 Chr.7: 965147-965147 on Build GRCh38
rs1011445831 Chr.7: 964917-964917 on Build GRCh38
rs565317338 Chr.7: 964760-964760 on Build GRCh38
rs542282863 Chr.7: 965064-965064 on Build GRCh38
rs943300207 Chr.7: 964959-964959 on Build GRCh38
rs919477076 Chr.7: 965051-965051 on Build GRCh38
rs1000553979 Chr.7: 965040-965040 on Build GRCh38
rs976410975 Chr.7: 965128-965128 on Build GRCh38
rs570642638 Chr.7: 964968-964968 on Build GRCh38
rs548072151 Chr.7: 964814-964814 on Build GRCh38
rs181033979 Chr.7: 964806-964806 on Build GRCh38
rs769092159 Chr.7: 965132-965132 on Build GRCh38
rs781423663 Chr.7: 964841-964841 on Build GRCh38
rs780294617 Chr.7: 964953-964953 on Build GRCh38
rs919988712 Chr.7: 964904-964904 on Build GRCh38
rs943790328 Chr.7: 965097-965097 on Build GRCh38
rs1007463269 Chr.7: 964733-964733 on Build GRCh38
rs943178002 Chr.7: 965116-965116 on Build GRCh38
rs1035949898 Chr.7: 964957-964957 on Build GRCh38

View Full Product Details