Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 968890-969403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX19
Entrez Gene ID
90639
Gene Name
COX19, cytochrome c oxidase assembly factor
Gene Aliases
-
UniGene
Hs.121593
Gene Chromosome Location
Chr.7: 964850-975599 on Build GRCh38
Transcripts
NM_001031617.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
743506 c.251G>C p.G84A Chr.7: 969400-969400 on Build GRCh38
3083122 c.270_271insA p.*91fs? Chr.7: 969380-969381 on Build GRCh38
327402 c.257C>G p.S86* Chr.7: 969394-969394 on Build GRCh38
3729941 c.270delA p.K90fs? Chr.7: 969381-969381 on Build GRCh38
743505 c.250G>T p.G84* Chr.7: 969401-969401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370034495 Chr.7: 969352-969352 on Build GRCh38
rs1039631903 Chr.7: 969021-969021 on Build GRCh38
rs546180203 Chr.7: 969078-969078 on Build GRCh38
rs746871444 Chr.7: 969374-969374 on Build GRCh38
rs762008789 Chr.7: 969347-969347 on Build GRCh38
rs13227492 Chr.7: 969062-969062 on Build GRCh38
rs543146354 Chr.7: 969137-969137 on Build GRCh38
rs963953162 Chr.7: 969331-969331 on Build GRCh38
rs138508539 Chr.7: 968954-968954 on Build GRCh38
rs557536956 Chr.7: 969034-969034 on Build GRCh38
rs2293521 Chr.7: 969330-969330 on Build GRCh38
rs761954928 Chr.7: 968950-968950 on Build GRCh38
rs903053924 Chr.7: 969259-969259 on Build GRCh38
rs910224691 Chr.7: 968955-968955 on Build GRCh38
rs951187512 Chr.7: 969179-969179 on Build GRCh38
rs1017914204 Chr.7: 969276-969276 on Build GRCh38
rs575820735 Chr.7: 969132-969132 on Build GRCh38
rs866652408 Chr.7: 969243-969243 on Build GRCh38
rs952033640 Chr.7: 968930-968930 on Build GRCh38
rs553405739 Chr.7: 969083-969083 on Build GRCh38
rs922931795 Chr.7: 969314-969314 on Build GRCh38
rs141657465 Chr.7: 969056-969056 on Build GRCh38
rs998932578 Chr.7: 969264-969264 on Build GRCh38
rs879150161 Chr.7: 969254-969254 on Build GRCh38
rs947353077 Chr.7: 969237-969237 on Build GRCh38
rs963681067 Chr.7: 969157-969157 on Build GRCh38
rs551729335 Chr.7: 969272-969272 on Build GRCh38
rs960061483 Chr.7: 969107-969107 on Build GRCh38
rs1039349133 Chr.7: 969212-969212 on Build GRCh38
rs758937726 Chr.7: 969217-969217 on Build GRCh38
rs912255091 Chr.7: 969001-969001 on Build GRCh38
rs1043261004 Chr.7: 969238-969238 on Build GRCh38
rs1055400313 Chr.7: 969357-969357 on Build GRCh38
rs113867737 Chr.7: 969358-969359 on Build GRCh38
rs532084830 Chr.7: 969211-969211 on Build GRCh38
rs756313489 Chr.7: 969349-969350 on Build GRCh38
rs752583609 Chr.7: 969353-969353 on Build GRCh38
rs1046701034 Chr.7: 968990-968990 on Build GRCh38
rs982538136 Chr.7: 969187-969187 on Build GRCh38
rs56227150 Chr.7: 969290-969290 on Build GRCh38
rs542525195 Chr.7: 969085-969085 on Build GRCh38
rs975749123 Chr.7: 969291-969291 on Build GRCh38
rs942998566 Chr.7: 969025-969025 on Build GRCh38
rs1011800191 Chr.7: 969051-969051 on Build GRCh38
rs373842534 Chr.7: 969366-969366 on Build GRCh38
rs537129627 Chr.7: 969006-969006 on Build GRCh38
rs943957107 Chr.7: 969016-969016 on Build GRCh38
rs943675083 Chr.7: 969208-969208 on Build GRCh38
rs1024962400 Chr.7: 969156-969156 on Build GRCh38
rs919463385 Chr.7: 969165-969165 on Build GRCh38
rs777702459 Chr.7: 969367-969367 on Build GRCh38
rs548956527 Chr.7: 969105-969105 on Build GRCh38
rs995124029 Chr.7: 969340-969340 on Build GRCh38
rs554435029 Chr.7: 968935-968935 on Build GRCh38
rs751723244 Chr.7: 969343-969343 on Build GRCh38
rs912223876 Chr.7: 969190-969190 on Build GRCh38
rs903251178 Chr.7: 969077-969077 on Build GRCh38
rs994247757 Chr.7: 969166-969166 on Build GRCh38
rs761092683 Chr.7: 969110-969110 on Build GRCh38
rs75405961 Chr.7: 969163-969163 on Build GRCh38
rs376965711 Chr.7: 969350-969350 on Build GRCh38
rs72201515 Chr.7: 968918-968918 on Build GRCh38
rs573548382 Chr.7: 969084-969084 on Build GRCh38
rs895906226 Chr.7: 969281-969281 on Build GRCh38

View Full Product Details