Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 975258-975758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX19
Entrez Gene ID
90639
Gene Name
COX19, cytochrome c oxidase assembly factor
Gene Aliases
-
UniGene
Hs.121593
Gene Chromosome Location
Chr.7: 964850-975599 on Build GRCh38
Transcripts
NM_001031617.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3431008 c.15G>T p.M5I Chr.7: 975495-975495 on Build GRCh38
6175364 c.48C>T p.P16P Chr.7: 975462-975462 on Build GRCh38
6245034 c.54C>A p.D18E Chr.7: 975456-975456 on Build GRCh38
1202108 c.11C>A p.A4D Chr.7: 975499-975499 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779055980 Chr.7: 975542-975542 on Build GRCh38
rs891728177 Chr.7: 975306-975306 on Build GRCh38
rs1029224613 Chr.7: 975721-975721 on Build GRCh38
rs543109728 Chr.7: 975589-975589 on Build GRCh38
rs971044282 Chr.7: 975405-975405 on Build GRCh38
rs1004197599 Chr.7: 975421-975421 on Build GRCh38
rs1024168849 Chr.7: 975318-975318 on Build GRCh38
rs988873321 Chr.7: 975563-975563 on Build GRCh38
rs770837687 Chr.7: 975513-975513 on Build GRCh38
rs369873504 Chr.7: 975488-975488 on Build GRCh38
rs200879257 Chr.7: 975483-975483 on Build GRCh38
rs760322069 Chr.7: 975440-975440 on Build GRCh38
rs749287209 Chr.7: 975477-975477 on Build GRCh38
rs891692712 Chr.7: 975461-975461 on Build GRCh38
rs912935360 Chr.7: 975585-975585 on Build GRCh38
rs1027695352 Chr.7: 975324-975324 on Build GRCh38
rs376293160 Chr.7: 975524-975524 on Build GRCh38
rs769577496 Chr.7: 975521-975521 on Build GRCh38
rs1028853960 Chr.7: 975715-975715 on Build GRCh38
rs568298546 Chr.7: 975366-975366 on Build GRCh38
rs983492455 Chr.7: 975356-975356 on Build GRCh38
rs1001610164 Chr.7: 975320-975320 on Build GRCh38
rs919194417 Chr.7: 975296-975296 on Build GRCh38
rs199952580 Chr.7: 975470-975470 on Build GRCh38
rs778384802 Chr.7: 975381-975381 on Build GRCh38
rs759188085 Chr.7: 975449-975449 on Build GRCh38
rs778396345 Chr.7: 975395-975395 on Build GRCh38
rs923625121 Chr.7: 975512-975512 on Build GRCh38
rs1049420901 Chr.7: 975290-975290 on Build GRCh38
rs567123064 Chr.7: 975508-975508 on Build GRCh38
rs138613071 Chr.7: 975474-975474 on Build GRCh38
rs762299988 Chr.7: 975500-975500 on Build GRCh38
rs755315888 Chr.7: 975506-975506 on Build GRCh38
rs771354041 Chr.7: 975402-975402 on Build GRCh38
rs763164727 Chr.7: 975423-975423 on Build GRCh38
rs766211888 Chr.7: 975446-975446 on Build GRCh38
rs755198910 Chr.7: 975539-975539 on Build GRCh38
rs866472349 Chr.7: 975653-975653 on Build GRCh38
rs750216092 Chr.7: 975522-975522 on Build GRCh38
rs768787562 Chr.7: 975479-975479 on Build GRCh38
rs535786468 Chr.7: 975518-975518 on Build GRCh38
rs1039251794 Chr.7: 975280-975280 on Build GRCh38
rs112587865 Chr.7: 975635-975635 on Build GRCh38
rs997938345 Chr.7: 975712-975712 on Build GRCh38
rs752107932 Chr.7: 975463-975463 on Build GRCh38
rs746099658 Chr.7: 975410-975410 on Build GRCh38
rs772966683 Chr.7: 975482-975482 on Build GRCh38
rs999869417 Chr.7: 975301-975301 on Build GRCh38
rs901809842 Chr.7: 975693-975693 on Build GRCh38
rs376380079 Chr.7: 975422-975422 on Build GRCh38
rs758805227 Chr.7: 975394-975394 on Build GRCh38
rs147438710 Chr.7: 975501-975501 on Build GRCh38
rs781037069 Chr.7: 975556-975556 on Build GRCh38
rs773233226 Chr.7: 975427-975427 on Build GRCh38
rs904150834 Chr.7: 975298-975298 on Build GRCh38
rs964134584 Chr.7: 975694-975694 on Build GRCh38
rs531007457 Chr.7: 975371-975371 on Build GRCh38
rs1053416422 Chr.7: 975305-975305 on Build GRCh38
rs906683178 Chr.7: 975558-975558 on Build GRCh38
rs1015427138 Chr.7: 975685-975685 on Build GRCh38
rs932424471 Chr.7: 975289-975289 on Build GRCh38
rs747283944 Chr.7: 975399-975399 on Build GRCh38
rs1052810536 Chr.7: 975452-975452 on Build GRCh38
rs952021086 Chr.7: 975341-975341 on Build GRCh38
rs905808703 Chr.7: 975616-975616 on Build GRCh38
rs749540793 Chr.7: 975509-975509 on Build GRCh38
rs781202136 Chr.7: 975405-975405 on Build GRCh38
rs776576886 Chr.7: 975514-975514 on Build GRCh38
rs754429069 Chr.7: 975379-975379 on Build GRCh38
rs750971043 Chr.7: 975536-975536 on Build GRCh38
rs773879377 Chr.7: 975526-975526 on Build GRCh38
rs557532718 Chr.7: 975621-975643 on Build GRCh38
rs149293639 Chr.7: 975493-975493 on Build GRCh38
rs932141316 Chr.7: 975401-975401 on Build GRCh38
rs764913858 Chr.7: 975450-975450 on Build GRCh38
rs1005008646 Chr.7: 975671-975671 on Build GRCh38
rs757775856 Chr.7: 975503-975503 on Build GRCh38
rs3735678 Chr.7: 975385-975385 on Build GRCh38
rs757503087 Chr.7: 975469-975469 on Build GRCh38
rs555525058 Chr.7: 975533-975533 on Build GRCh38
rs753694219 Chr.7: 975448-975448 on Build GRCh38
rs757943601 Chr.7: 975462-975462 on Build GRCh38
rs1046508976 Chr.7: 975321-975321 on Build GRCh38
rs370650176 Chr.7: 975505-975505 on Build GRCh38
rs775150357 Chr.7: 975498-975498 on Build GRCh38
rs1016852151 Chr.7: 975677-975677 on Build GRCh38
rs748214719 Chr.7: 975554-975554 on Build GRCh38
rs758508610 Chr.7: 975555-975555 on Build GRCh38
rs764824177 Chr.7: 975497-975497 on Build GRCh38
rs769778907 Chr.7: 975561-975561 on Build GRCh38
rs150428640 Chr.7: 975473-975473 on Build GRCh38
rs937771993 Chr.7: 975458-975458 on Build GRCh38
rs887857139 Chr.7: 975670-975670 on Build GRCh38
rs756489750 Chr.7: 975537-975537 on Build GRCh38
rs1005517205 Chr.7: 975662-975662 on Build GRCh38
rs570851493 Chr.7: 975455-975455 on Build GRCh38
rs771856290 Chr.7: 975426-975426 on Build GRCh38
rs887331734 Chr.7: 975657-975657 on Build GRCh38
rs867043038 Chr.7: 975612-975612 on Build GRCh38
rs770197471 Chr.7: 975411-975411 on Build GRCh38
rs575459758 Chr.7: 975538-975538 on Build GRCh38
rs954682520 Chr.7: 975731-975731 on Build GRCh38
rs374651385 Chr.7: 975510-975510 on Build GRCh38
rs762650817 Chr.7: 975523-975523 on Build GRCh38
rs548331157 Chr.7: 975319-975319 on Build GRCh38
rs745816054 Chr.7: 975559-975559 on Build GRCh38
rs377326731 Chr.7: 975504-975504 on Build GRCh38
rs3735677 Chr.7: 975307-975307 on Build GRCh38
rs148763453 Chr.7: 975735-975735 on Build GRCh38
rs576534626 Chr.7: 975646-975646 on Build GRCh38
rs770842621 Chr.7: 975485-975485 on Build GRCh38

View Full Product Details