Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 988978-989502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012719.1 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

Gene Symbol
CYP2W1
Entrez Gene ID
54905
Gene Name
cytochrome P450 family 2 subfamily W member 1
Gene Aliases
-
UniGene
Hs.272795
Gene Chromosome Location
Chr.7: 983157-989640 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012719.1 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

SNP Detail

SNP ID Chromosome Location
rs549815123 Chr.7: 989225-989225 on Build GRCh38
rs371837394 Chr.7: 989363-989363 on Build GRCh38
rs193101956 Chr.7: 989174-989174 on Build GRCh38
rs761018416 Chr.7: 989416-989416 on Build GRCh38
rs772514228 Chr.7: 989241-989241 on Build GRCh38
rs564406715 Chr.7: 989107-989107 on Build GRCh38
rs1056278662 Chr.7: 989166-989166 on Build GRCh38
rs562047821 Chr.7: 989022-989022 on Build GRCh38
rs768995114 Chr.7: 989079-989079 on Build GRCh38
rs964049805 Chr.7: 989077-989077 on Build GRCh38
rs1040625761 Chr.7: 989266-989266 on Build GRCh38
rs928001218 Chr.7: 989443-989443 on Build GRCh38
rs1049842106 Chr.7: 989236-989236 on Build GRCh38
rs1025580271 Chr.7: 989356-989356 on Build GRCh38
rs929270125 Chr.7: 989083-989083 on Build GRCh38
rs986944379 Chr.7: 989069-989069 on Build GRCh38
rs748498766 Chr.7: 989230-989230 on Build GRCh38
rs991303260 Chr.7: 989436-989436 on Build GRCh38
rs1009459818 Chr.7: 989087-989087 on Build GRCh38
rs1000530616 Chr.7: 989336-989336 on Build GRCh38
rs1030435277 Chr.7: 989031-989031 on Build GRCh38
rs919921970 Chr.7: 989218-989218 on Build GRCh38
rs945222720 Chr.7: 989439-989439 on Build GRCh38
rs920168187 Chr.7: 989369-989369 on Build GRCh38
rs977328004 Chr.7: 989361-989361 on Build GRCh38
rs993693544 Chr.7: 989192-989192 on Build GRCh38
rs577289586 Chr.7: 989235-989235 on Build GRCh38
rs551395751 Chr.7: 989437-989437 on Build GRCh38
rs967666815 Chr.7: 989157-989157 on Build GRCh38
rs928443784 Chr.7: 989150-989150 on Build GRCh38
rs1033452623 Chr.7: 989338-989338 on Build GRCh38
rs991893388 Chr.7: 989340-989340 on Build GRCh38
rs183944138 Chr.7: 989417-989417 on Build GRCh38
rs542347265 Chr.7: 989003-989003 on Build GRCh38
rs915958790 Chr.7: 989167-989167 on Build GRCh38
rs544218997 Chr.7: 989082-989082 on Build GRCh38
rs1052053756 Chr.7: 989200-989200 on Build GRCh38
rs1042736283 Chr.7: 989450-989450 on Build GRCh38
rs183047053 Chr.7: 989062-989062 on Build GRCh38
rs1018058763 Chr.7: 989076-989076 on Build GRCh38
rs901654407 Chr.7: 989278-989278 on Build GRCh38
rs1037775660 Chr.7: 989457-989457 on Build GRCh38
rs890612710 Chr.7: 989203-989203 on Build GRCh38
rs1011346677 Chr.7: 989260-989260 on Build GRCh38
rs773797499 Chr.7: 989259-989259 on Build GRCh38
rs533230757 Chr.7: 989236-989236 on Build GRCh38
rs1021059431 Chr.7: 989144-989144 on Build GRCh38
rs931339305 Chr.7: 989376-989376 on Build GRCh38
rs985416087 Chr.7: 989379-989379 on Build GRCh38
rs966343308 Chr.7: 989084-989084 on Build GRCh38
rs961527464 Chr.7: 989358-989358 on Build GRCh38
rs903555881 Chr.7: 989318-989318 on Build GRCh38
rs528164657 Chr.7: 989232-989232 on Build GRCh38
rs967062303 Chr.7: 989317-989317 on Build GRCh38
rs775987707 Chr.7: 989465-989465 on Build GRCh38
rs187651886 Chr.7: 989438-989438 on Build GRCh38
rs187334159 Chr.7: 989159-989159 on Build GRCh38
rs1021280171 Chr.7: 989291-989291 on Build GRCh38
rs868301995 Chr.7: 989127-989127 on Build GRCh38
rs897848361 Chr.7: 989175-989175 on Build GRCh38
rs960232102 Chr.7: 989402-989402 on Build GRCh38
rs371134564 Chr.7: 989170-989170 on Build GRCh38
rs993724345 Chr.7: 989004-989004 on Build GRCh38

View Full Product Details