Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1744554-1745095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELFN1
Entrez Gene ID
392617
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 1
Gene Aliases
PPP1R28
UniGene
Hs.42896
Gene Chromosome Location
Chr.7: 1666075-1747954 on Build GRCh38
Transcripts
NM_001128636.2 XM_006715725.3 XM_006715726.2 XM_006715727.3 XM_011515397.2 XM_011515398.2 XM_011515401.2 XM_017012203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6247951 c.73G>T p.A25S Chr.7: 1744669-1744669 on Build GRCh38
4139342 c.124G>A p.A42T Chr.7: 1744720-1744720 on Build GRCh38
3306781 c.356G>A p.R119Q Chr.7: 1744952-1744952 on Build GRCh38
3306779 c.340C>A p.Q114K Chr.7: 1744936-1744936 on Build GRCh38
6996473 c.205G>A p.E69K Chr.7: 1744801-1744801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774091154 Chr.7: 1744711-1744711 on Build GRCh38
rs746584397 Chr.7: 1744642-1744642 on Build GRCh38
rs370472465 Chr.7: 1744818-1744818 on Build GRCh38
rs1041507240 Chr.7: 1744764-1744764 on Build GRCh38
rs1045671838 Chr.7: 1745038-1745038 on Build GRCh38
rs759357234 Chr.7: 1744879-1744879 on Build GRCh38
rs534524610 Chr.7: 1744590-1744590 on Build GRCh38
rs1003592158 Chr.7: 1744663-1744663 on Build GRCh38
rs1005546401 Chr.7: 1744900-1744900 on Build GRCh38
rs1000090724 Chr.7: 1744631-1744631 on Build GRCh38
rs780207326 Chr.7: 1744618-1744618 on Build GRCh38
rs763630960 Chr.7: 1744698-1744698 on Build GRCh38
rs771757749 Chr.7: 1744633-1744633 on Build GRCh38
rs754663020 Chr.7: 1744620-1744620 on Build GRCh38
rs770198560 Chr.7: 1744926-1744926 on Build GRCh38
rs1043696015 Chr.7: 1744782-1744782 on Build GRCh38
rs577726835 Chr.7: 1744878-1744878 on Build GRCh38
rs977157332 Chr.7: 1744970-1744970 on Build GRCh38
rs767707781 Chr.7: 1744750-1744750 on Build GRCh38
rs765281801 Chr.7: 1744884-1744884 on Build GRCh38
rs749827298 Chr.7: 1744952-1744952 on Build GRCh38
rs774875966 Chr.7: 1744978-1744978 on Build GRCh38
rs965160942 Chr.7: 1744958-1744958 on Build GRCh38
rs148419230 Chr.7: 1744749-1744749 on Build GRCh38
rs777068156 Chr.7: 1744662-1744662 on Build GRCh38
rs925300037 Chr.7: 1745028-1745028 on Build GRCh38
rs868240549 Chr.7: 1744917-1744917 on Build GRCh38
rs935653302 Chr.7: 1745004-1745004 on Build GRCh38
rs1000045716 Chr.7: 1744603-1744603 on Build GRCh38
rs933698638 Chr.7: 1744761-1744761 on Build GRCh38
rs778549064 Chr.7: 1744785-1744785 on Build GRCh38
rs768988884 Chr.7: 1744832-1744832 on Build GRCh38
rs751739457 Chr.7: 1744613-1744613 on Build GRCh38
rs935336609 Chr.7: 1745031-1745031 on Build GRCh38
rs371567558 Chr.7: 1744899-1744899 on Build GRCh38
rs368018088 Chr.7: 1744776-1744776 on Build GRCh38
rs202117598 Chr.7: 1744893-1744893 on Build GRCh38
rs896616396 Chr.7: 1744856-1744856 on Build GRCh38
rs770385203 Chr.7: 1744819-1744819 on Build GRCh38
rs900958186 Chr.7: 1744595-1744595 on Build GRCh38
rs114295521 Chr.7: 1744656-1744656 on Build GRCh38
rs567598793 Chr.7: 1744619-1744619 on Build GRCh38
rs571712654 Chr.7: 1744589-1744589 on Build GRCh38
rs531447239 Chr.7: 1745058-1745058 on Build GRCh38
rs1039631982 Chr.7: 1744585-1744585 on Build GRCh38
rs975230706 Chr.7: 1744968-1744968 on Build GRCh38
rs910156716 Chr.7: 1744679-1744679 on Build GRCh38
rs774647028 Chr.7: 1744838-1744838 on Build GRCh38
rs775349893 Chr.7: 1745071-1745071 on Build GRCh38
rs979125513 Chr.7: 1745013-1745013 on Build GRCh38
rs375131305 Chr.7: 1744674-1744674 on Build GRCh38
rs770363824 Chr.7: 1744814-1744814 on Build GRCh38
rs756087137 Chr.7: 1744617-1744617 on Build GRCh38
rs77210571 Chr.7: 1744911-1744911 on Build GRCh38
rs943968930 Chr.7: 1744579-1744579 on Build GRCh38
rs533079342 Chr.7: 1744827-1744827 on Build GRCh38
rs773156974 Chr.7: 1745015-1745015 on Build GRCh38
rs765480745 Chr.7: 1744673-1744673 on Build GRCh38
rs752573685 Chr.7: 1744792-1744792 on Build GRCh38
rs767656349 Chr.7: 1744866-1744866 on Build GRCh38
rs777457873 Chr.7: 1744800-1744800 on Build GRCh38
rs554016580 Chr.7: 1744607-1744607 on Build GRCh38
rs764452221 Chr.7: 1744699-1744699 on Build GRCh38
rs762104391 Chr.7: 1744982-1744982 on Build GRCh38
rs369219960 Chr.7: 1745070-1745070 on Build GRCh38
rs370941771 Chr.7: 1744672-1744672 on Build GRCh38
rs1045941152 Chr.7: 1744839-1744839 on Build GRCh38
rs780787281 Chr.7: 1744817-1744817 on Build GRCh38
rs747704550 Chr.7: 1744632-1744632 on Build GRCh38
rs758680396 Chr.7: 1744602-1744602 on Build GRCh38
rs373314329 Chr.7: 1744902-1744902 on Build GRCh38
rs140278883 Chr.7: 1744983-1744983 on Build GRCh38
rs922801804 Chr.7: 1744710-1744710 on Build GRCh38
rs756930012 Chr.7: 1744908-1744908 on Build GRCh38
rs763064950 Chr.7: 1744692-1744692 on Build GRCh38
rs931956034 Chr.7: 1744780-1744780 on Build GRCh38
rs982521348 Chr.7: 1744919-1744919 on Build GRCh38
rs1031149882 Chr.7: 1744606-1744606 on Build GRCh38
rs757471650 Chr.7: 1744746-1744746 on Build GRCh38
rs769101353 Chr.7: 1744957-1744957 on Build GRCh38
rs762155636 Chr.7: 1744857-1744857 on Build GRCh38
rs545520580 Chr.7: 1744638-1744638 on Build GRCh38
rs745425168 Chr.7: 1744920-1744920 on Build GRCh38
rs763479740 Chr.7: 1744831-1744831 on Build GRCh38
rs746674332 Chr.7: 1744808-1744808 on Build GRCh38
rs758037742 Chr.7: 1744797-1744797 on Build GRCh38
rs1013837355 Chr.7: 1744616-1744616 on Build GRCh38
rs182090780 Chr.7: 1744885-1744885 on Build GRCh38
rs1013202524 Chr.7: 1744641-1744641 on Build GRCh38
rs947286942 Chr.7: 1744994-1744994 on Build GRCh38
rs752800060 Chr.7: 1744596-1744596 on Build GRCh38
rs780670586 Chr.7: 1744929-1744929 on Build GRCh38

View Full Product Details