Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1746865-1747364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELFN1
Entrez Gene ID
392617
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 1
Gene Aliases
PPP1R28
UniGene
Hs.42896
Gene Chromosome Location
Chr.7: 1666075-1747954 on Build GRCh38
Transcripts
NM_001128636.2 XM_006715725.3 XM_006715726.2 XM_006715727.3 XM_011515397.2 XM_011515398.2 XM_011515401.2 XM_017012203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4565659 c.2364_2365GG>AA p.E789K Chr.7: 1746960-1746961 on Build GRCh38
5544898 c.2365G>A p.E789K Chr.7: 1746961-1746961 on Build GRCh38
3411880 c.2416T>C p.F806L Chr.7: 1747012-1747012 on Build GRCh38
3394652 c.2353C>T p.R785W Chr.7: 1746949-1746949 on Build GRCh38
5444328 c.2295C>T p.S765S Chr.7: 1746891-1746891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs57908849 Chr.7: 1747335-1747336 on Build GRCh38
rs897812110 Chr.7: 1746946-1746946 on Build GRCh38
rs868205051 Chr.7: 1746961-1746961 on Build GRCh38
rs748145478 Chr.7: 1746901-1746901 on Build GRCh38
rs777702721 Chr.7: 1747034-1747034 on Build GRCh38
rs568015707 Chr.7: 1747219-1747219 on Build GRCh38
rs926291905 Chr.7: 1747164-1747164 on Build GRCh38
rs959058757 Chr.7: 1747336-1747336 on Build GRCh38
rs1041887956 Chr.7: 1747159-1747159 on Build GRCh38
rs543465852 Chr.7: 1746895-1746895 on Build GRCh38
rs919308533 Chr.7: 1747231-1747231 on Build GRCh38
rs1020517722 Chr.7: 1746908-1746908 on Build GRCh38
rs148122916 Chr.7: 1747099-1747099 on Build GRCh38
rs746290433 Chr.7: 1747315-1747316 on Build GRCh38
rs758397436 Chr.7: 1747023-1747023 on Build GRCh38
rs770773878 Chr.7: 1746948-1746948 on Build GRCh38
rs1033285887 Chr.7: 1747323-1747323 on Build GRCh38
rs946108349 Chr.7: 1747297-1747297 on Build GRCh38
rs769139773 Chr.7: 1747098-1747098 on Build GRCh38
rs750674255 Chr.7: 1746990-1746990 on Build GRCh38
rs763339339 Chr.7: 1747100-1747100 on Build GRCh38
rs3077247 Chr.7: 1747313-1747314 on Build GRCh38
rs76371074 Chr.7: 1747182-1747182 on Build GRCh38
rs781059252 Chr.7: 1746965-1746966 on Build GRCh38
rs377601393 Chr.7: 1747177-1747177 on Build GRCh38
rs200235323 Chr.7: 1746987-1746987 on Build GRCh38
rs143376319 Chr.7: 1747302-1747303 on Build GRCh38
rs954555192 Chr.7: 1747244-1747244 on Build GRCh38
rs557288458 Chr.7: 1746952-1746952 on Build GRCh38
rs142744207 Chr.7: 1747132-1747132 on Build GRCh38
rs370334516 Chr.7: 1746981-1746981 on Build GRCh38
rs946179754 Chr.7: 1747142-1747142 on Build GRCh38
rs776267027 Chr.7: 1746949-1746949 on Build GRCh38
rs958649118 Chr.7: 1746967-1746967 on Build GRCh38
rs780202361 Chr.7: 1747010-1747010 on Build GRCh38
rs570881196 Chr.7: 1747316-1747316 on Build GRCh38
rs372692511 Chr.7: 1747088-1747088 on Build GRCh38
rs757469332 Chr.7: 1746985-1746985 on Build GRCh38
rs79870618 Chr.7: 1747283-1747283 on Build GRCh38
rs894101510 Chr.7: 1747172-1747172 on Build GRCh38
rs773186780 Chr.7: 1746931-1746931 on Build GRCh38
rs530916643 Chr.7: 1747104-1747104 on Build GRCh38
rs554145292 Chr.7: 1747211-1747211 on Build GRCh38
rs907438302 Chr.7: 1746920-1746920 on Build GRCh38
rs757431354 Chr.7: 1747263-1747263 on Build GRCh38
rs902005739 Chr.7: 1747264-1747264 on Build GRCh38
rs565678260 Chr.7: 1747208-1747208 on Build GRCh38
rs940310999 Chr.7: 1747285-1747306 on Build GRCh38
rs574238696 Chr.7: 1746894-1746894 on Build GRCh38
rs910480167 Chr.7: 1747276-1747276 on Build GRCh38
rs1534789 Chr.7: 1747209-1747209 on Build GRCh38
rs1010838225 Chr.7: 1746902-1746902 on Build GRCh38
rs923131587 Chr.7: 1747018-1747018 on Build GRCh38
rs878915853 Chr.7: 1747060-1747060 on Build GRCh38
rs756084251 Chr.7: 1747001-1747001 on Build GRCh38
rs758849960 Chr.7: 1747167-1747167 on Build GRCh38
rs192078216 Chr.7: 1747163-1747163 on Build GRCh38
rs551937318 Chr.7: 1747196-1747196 on Build GRCh38
rs907407036 Chr.7: 1747161-1747161 on Build GRCh38
rs771713224 Chr.7: 1746924-1746924 on Build GRCh38
rs114713154 Chr.7: 1747065-1747065 on Build GRCh38
rs577340198 Chr.7: 1746984-1746984 on Build GRCh38
rs914599487 Chr.7: 1747284-1747284 on Build GRCh38
rs111457141 Chr.7: 1746953-1746953 on Build GRCh38
rs536770487 Chr.7: 1747249-1747249 on Build GRCh38
rs5019477 Chr.7: 1747317-1747317 on Build GRCh38
rs542293020 Chr.7: 1747097-1747097 on Build GRCh38
rs775822166 Chr.7: 1746970-1746970 on Build GRCh38
rs61188936 Chr.7: 1747316-1747317 on Build GRCh38
rs1030116357 Chr.7: 1747227-1747227 on Build GRCh38
rs374165762 Chr.7: 1747059-1747059 on Build GRCh38
rs746626090 Chr.7: 1746947-1746947 on Build GRCh38
rs748011357 Chr.7: 1747014-1747014 on Build GRCh38
rs993246490 Chr.7: 1747216-1747216 on Build GRCh38
rs889139410 Chr.7: 1747250-1747250 on Build GRCh38
rs776355461 Chr.7: 1747096-1747096 on Build GRCh38
rs74671825 Chr.7: 1747313-1747313 on Build GRCh38
rs1036475500 Chr.7: 1746932-1746932 on Build GRCh38
rs1007796632 Chr.7: 1747252-1747252 on Build GRCh38
rs764807037 Chr.7: 1746968-1746968 on Build GRCh38
rs964047812 Chr.7: 1747277-1747277 on Build GRCh38
rs1040677830 Chr.7: 1747255-1747255 on Build GRCh38
rs1056066077 Chr.7: 1747168-1747168 on Build GRCh38

View Full Product Details