Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4762077-4762565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXK1
Entrez Gene ID
221937
Gene Name
forkhead box K1
Gene Aliases
FOXK1L
UniGene
Hs.487393
Gene Chromosome Location
Chr.7: 4644691-4771443 on Build GRCh38
Transcripts
NM_001037165.1 XM_011515191.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3950559 c.2003G>T p.G668V Chr.7: 4762265-4762265 on Build GRCh38
5062498 c.1883C>T p.A628V Chr.7: 4762211-4762211 on Build GRCh38
4828822 c.2011G>A p.E671K Chr.7: 4762273-4762273 on Build GRCh38
7206668 c.1967C>G p.S656C Chr.7: 4762229-4762229 on Build GRCh38
6547960 c.2026G>T p.V676F Chr.7: 4762288-4762288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886286216 Chr.7: 4762233-4762233 on Build GRCh38
rs111467325 Chr.7: 4762478-4762478 on Build GRCh38
rs989317903 Chr.7: 4762188-4762188 on Build GRCh38
rs755913952 Chr.7: 4762327-4762327 on Build GRCh38
rs778049352 Chr.7: 4762195-4762195 on Build GRCh38
rs368673072 Chr.7: 4762341-4762341 on Build GRCh38
rs986132321 Chr.7: 4762540-4762540 on Build GRCh38
rs13242966 Chr.7: 4762511-4762511 on Build GRCh38
rs763448164 Chr.7: 4762295-4762295 on Build GRCh38
rs201818414 Chr.7: 4762139-4762139 on Build GRCh38
rs192821100 Chr.7: 4762221-4762221 on Build GRCh38
rs112910907 Chr.7: 4762230-4762230 on Build GRCh38
rs113624299 Chr.7: 4762502-4762502 on Build GRCh38
rs748606326 Chr.7: 4762477-4762477 on Build GRCh38
rs777036898 Chr.7: 4762176-4762176 on Build GRCh38
rs747112105 Chr.7: 4762287-4762287 on Build GRCh38
rs528417588 Chr.7: 4762293-4762293 on Build GRCh38
rs187866118 Chr.7: 4762538-4762538 on Build GRCh38
rs771976720 Chr.7: 4762262-4762262 on Build GRCh38
rs776449631 Chr.7: 4762510-4762510 on Build GRCh38
rs371407158 Chr.7: 4762435-4762435 on Build GRCh38
rs956842327 Chr.7: 4762379-4762379 on Build GRCh38
rs71529310 Chr.7: 4762301-4762301 on Build GRCh38
rs767379360 Chr.7: 4762154-4762154 on Build GRCh38
rs771157780 Chr.7: 4762290-4762290 on Build GRCh38
rs143086698 Chr.7: 4762254-4762254 on Build GRCh38
rs762393895 Chr.7: 4762220-4762220 on Build GRCh38
rs1000865739 Chr.7: 4762288-4762288 on Build GRCh38
rs761269819 Chr.7: 4762303-4762303 on Build GRCh38
rs976522619 Chr.7: 4762245-4762245 on Build GRCh38
rs1053753 Chr.7: 4762404-4762404 on Build GRCh38
rs201501146 Chr.7: 4762257-4762257 on Build GRCh38
rs950037990 Chr.7: 4762197-4762197 on Build GRCh38
rs1004614271 Chr.7: 4762235-4762235 on Build GRCh38
rs15715 Chr.7: 4762342-4762342 on Build GRCh38
rs373887374 Chr.7: 4762219-4762219 on Build GRCh38
rs771122446 Chr.7: 4762157-4762157 on Build GRCh38
rs890021052 Chr.7: 4762275-4762275 on Build GRCh38
rs111856572 Chr.7: 4762299-4762299 on Build GRCh38
rs781336946 Chr.7: 4762392-4762392 on Build GRCh38
rs985131363 Chr.7: 4762366-4762366 on Build GRCh38
rs940659256 Chr.7: 4762535-4762535 on Build GRCh38
rs750121455 Chr.7: 4762309-4762309 on Build GRCh38
rs534515942 Chr.7: 4762409-4762409 on Build GRCh38
rs538698881 Chr.7: 4762329-4762329 on Build GRCh38
rs148135152 Chr.7: 4762231-4762231 on Build GRCh38
rs909395379 Chr.7: 4762282-4762282 on Build GRCh38
rs199826676 Chr.7: 4762426-4762426 on Build GRCh38
rs770942503 Chr.7: 4762390-4762390 on Build GRCh38
rs997853367 Chr.7: 4762517-4762517 on Build GRCh38
rs570756478 Chr.7: 4762236-4762236 on Build GRCh38
rs931759576 Chr.7: 4762406-4762406 on Build GRCh38
rs994260019 Chr.7: 4762155-4762155 on Build GRCh38
rs573528476 Chr.7: 4762134-4762134 on Build GRCh38
rs559428025 Chr.7: 4762151-4762151 on Build GRCh38
rs770069495 Chr.7: 4762396-4762396 on Build GRCh38
rs1012021527 Chr.7: 4762302-4762302 on Build GRCh38
rs988877252 Chr.7: 4762297-4762297 on Build GRCh38
rs777901288 Chr.7: 4762279-4762279 on Build GRCh38
rs552635478 Chr.7: 4762163-4762163 on Build GRCh38
rs774750653 Chr.7: 4762400-4762400 on Build GRCh38
rs1033687239 Chr.7: 4762458-4762458 on Build GRCh38
rs370322820 Chr.7: 4762250-4762250 on Build GRCh38
rs867731857 Chr.7: 4762334-4762334 on Build GRCh38
rs765696483 Chr.7: 4762211-4762211 on Build GRCh38
rs62453207 Chr.7: 4762434-4762434 on Build GRCh38
rs746239626 Chr.7: 4762402-4762402 on Build GRCh38
rs778761314 Chr.7: 4762343-4762343 on Build GRCh38
rs759848287 Chr.7: 4762292-4762292 on Build GRCh38
rs768925063 Chr.7: 4762461-4762461 on Build GRCh38
rs772832879 Chr.7: 4762480-4762480 on Build GRCh38
rs575323176 Chr.7: 4762468-4762468 on Build GRCh38
rs555172890 Chr.7: 4762401-4762401 on Build GRCh38
rs746097478 Chr.7: 4762394-4762394 on Build GRCh38
rs374087096 Chr.7: 4762415-4762415 on Build GRCh38
rs569375484 Chr.7: 4762381-4762381 on Build GRCh38
rs752785227 Chr.7: 4762181-4762181 on Build GRCh38
rs749664106 Chr.7: 4762399-4762399 on Build GRCh38
rs546492376 Chr.7: 4762521-4762521 on Build GRCh38
rs767157990 Chr.7: 4762306-4762306 on Build GRCh38
rs923262748 Chr.7: 4762249-4762249 on Build GRCh38
rs6976790 Chr.7: 4762398-4762398 on Build GRCh38
rs201914134 Chr.7: 4762232-4762232 on Build GRCh38
rs776753169 Chr.7: 4762291-4762291 on Build GRCh38
rs756307100 Chr.7: 4762437-4762437 on Build GRCh38
rs546555728 Chr.7: 4762294-4762294 on Build GRCh38
rs765393957 Chr.7: 4762456-4762456 on Build GRCh38
rs750211580 Chr.7: 4762240-4762240 on Build GRCh38
rs561152075 Chr.7: 4762505-4762505 on Build GRCh38
rs1044735006 Chr.7: 4762484-4762484 on Build GRCh38
rs371863185 Chr.7: 4762413-4762413 on Build GRCh38
rs978879673 Chr.7: 4762106-4762106 on Build GRCh38
rs61733617 Chr.7: 4762194-4762194 on Build GRCh38
rs980081239 Chr.7: 4762330-4762330 on Build GRCh38
rs542439662 Chr.7: 4762481-4762481 on Build GRCh38
rs766095602 Chr.7: 4762504-4762504 on Build GRCh38
rs907777583 Chr.7: 4762534-4762534 on Build GRCh38
rs747075130 Chr.7: 4762363-4762363 on Build GRCh38
rs760953599 Chr.7: 4762153-4762153 on Build GRCh38
rs757859290 Chr.7: 4762309-4762310 on Build GRCh38

View Full Product Details