Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4763254-4763761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXK1
Entrez Gene ID
221937
Gene Name
forkhead box K1
Gene Aliases
FOXK1L
UniGene
Hs.487393
Gene Chromosome Location
Chr.7: 4644691-4771443 on Build GRCh38
Transcripts
NM_001037165.1 XM_011515191.2

SNP Detail

SNP ID Chromosome Location
rs927294052 Chr.7: 4763541-4763541 on Build GRCh38
rs190263181 Chr.7: 4763497-4763497 on Build GRCh38
rs537710471 Chr.7: 4763394-4763394 on Build GRCh38
rs531863599 Chr.7: 4763308-4763308 on Build GRCh38
rs116550350 Chr.7: 4763606-4763606 on Build GRCh38
rs546321524 Chr.7: 4763526-4763526 on Build GRCh38
rs138618206 Chr.7: 4763388-4763388 on Build GRCh38
rs1005216621 Chr.7: 4763336-4763336 on Build GRCh38
rs966860796 Chr.7: 4763651-4763651 on Build GRCh38
rs964316992 Chr.7: 4763426-4763426 on Build GRCh38
rs963630509 Chr.7: 4763356-4763356 on Build GRCh38
rs890699000 Chr.7: 4763627-4763627 on Build GRCh38
rs79572472 Chr.7: 4763322-4763322 on Build GRCh38
rs906480504 Chr.7: 4763400-4763400 on Build GRCh38
rs533658470 Chr.7: 4763379-4763379 on Build GRCh38
rs775428351 Chr.7: 4763605-4763605 on Build GRCh38
rs946145334 Chr.7: 4763595-4763595 on Build GRCh38
rs964789963 Chr.7: 4763505-4763505 on Build GRCh38
rs927220638 Chr.7: 4763672-4763672 on Build GRCh38
rs978113614 Chr.7: 4763565-4763565 on Build GRCh38
rs943487642 Chr.7: 4763381-4763381 on Build GRCh38
rs913348936 Chr.7: 4763570-4763571 on Build GRCh38
rs564689177 Chr.7: 4763306-4763306 on Build GRCh38
rs987595981 Chr.7: 4763491-4763491 on Build GRCh38
rs574669573 Chr.7: 4763441-4763441 on Build GRCh38
rs2175680 Chr.7: 4763283-4763283 on Build GRCh38
rs369427435 Chr.7: 4763478-4763478 on Build GRCh38
rs939212139 Chr.7: 4763713-4763713 on Build GRCh38
rs915219418 Chr.7: 4763534-4763534 on Build GRCh38
rs4996104 Chr.7: 4763355-4763355 on Build GRCh38
rs894888470 Chr.7: 4763421-4763421 on Build GRCh38
rs560545878 Chr.7: 4763282-4763282 on Build GRCh38
rs987138161 Chr.7: 4763293-4763293 on Build GRCh38
rs1003361633 Chr.7: 4763417-4763417 on Build GRCh38
rs983286322 Chr.7: 4763346-4763346 on Build GRCh38
rs911583846 Chr.7: 4763290-4763290 on Build GRCh38
rs547906387 Chr.7: 4763364-4763364 on Build GRCh38
rs566153256 Chr.7: 4763725-4763725 on Build GRCh38
rs556415312 Chr.7: 4763430-4763430 on Build GRCh38
rs964438912 Chr.7: 4763305-4763305 on Build GRCh38
rs568623550 Chr.7: 4763639-4763639 on Build GRCh38
rs1054444757 Chr.7: 4763570-4763570 on Build GRCh38
rs993776192 Chr.7: 4763612-4763612 on Build GRCh38
rs530831657 Chr.7: 4763469-4763469 on Build GRCh38
rs144645794 Chr.7: 4763697-4763697 on Build GRCh38
rs927959776 Chr.7: 4763705-4763705 on Build GRCh38
rs910617310 Chr.7: 4763380-4763380 on Build GRCh38
rs7810183 Chr.7: 4763454-4763454 on Build GRCh38
rs1029322938 Chr.7: 4763614-4763614 on Build GRCh38
rs891406819 Chr.7: 4763392-4763392 on Build GRCh38
rs150142708 Chr.7: 4763363-4763363 on Build GRCh38
rs113459374 Chr.7: 4763372-4763372 on Build GRCh38
rs771981838 Chr.7: 4763587-4763587 on Build GRCh38
rs1017731846 Chr.7: 4763635-4763635 on Build GRCh38
rs192676074 Chr.7: 4763277-4763277 on Build GRCh38
rs979920448 Chr.7: 4763660-4763660 on Build GRCh38
rs954981043 Chr.7: 4763473-4763473 on Build GRCh38
rs1017046210 Chr.7: 4763351-4763351 on Build GRCh38
rs531922825 Chr.7: 4763594-4763594 on Build GRCh38
rs529594472 Chr.7: 4763634-4763634 on Build GRCh38
rs1022785644 Chr.7: 4763645-4763645 on Build GRCh38
rs77329503 Chr.7: 4763644-4763644 on Build GRCh38
rs1055005453 Chr.7: 4763418-4763418 on Build GRCh38
rs149322726 Chr.7: 4763592-4763592 on Build GRCh38
rs550470458 Chr.7: 4763542-4763542 on Build GRCh38
rs527819519 Chr.7: 4763292-4763292 on Build GRCh38
rs112649083 Chr.7: 4763389-4763389 on Build GRCh38
rs184763840 Chr.7: 4763444-4763444 on Build GRCh38
rs562385170 Chr.7: 4763609-4763609 on Build GRCh38
rs1003268366 Chr.7: 4763319-4763319 on Build GRCh38
rs957274283 Chr.7: 4763709-4763709 on Build GRCh38
rs1026510010 Chr.7: 4763463-4763463 on Build GRCh38
rs1042344554 Chr.7: 4763399-4763399 on Build GRCh38
rs978080902 Chr.7: 4763691-4763691 on Build GRCh38
rs559566823 Chr.7: 4763296-4763296 on Build GRCh38
rs527836537 Chr.7: 4763307-4763307 on Build GRCh38
rs568699004 Chr.7: 4763329-4763329 on Build GRCh38
rs894975794 Chr.7: 4763602-4763602 on Build GRCh38
rs954888573 Chr.7: 4763371-4763371 on Build GRCh38

View Full Product Details