Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5479715-5480203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL18
Entrez Gene ID
80028
Gene Name
F-box and leucine rich repeat protein 18
Gene Aliases
Fbl18
UniGene
Hs.623974
Gene Chromosome Location
Chr.7: 5454448-5513813 on Build GRCh38
Transcripts
NM_024963.5 XM_011515531.2

SNP Detail

SNP ID Chromosome Location
rs910407557 Chr.7: 5480161-5480161 on Build GRCh38
rs76652583 Chr.7: 5479796-5479796 on Build GRCh38
rs529069291 Chr.7: 5479934-5479934 on Build GRCh38
rs368649776 Chr.7: 5479936-5479936 on Build GRCh38
rs1022798584 Chr.7: 5479741-5479741 on Build GRCh38
rs916401367 Chr.7: 5480066-5480066 on Build GRCh38
rs901994269 Chr.7: 5479829-5479829 on Build GRCh38
rs920240168 Chr.7: 5480136-5480136 on Build GRCh38
rs566018223 Chr.7: 5480008-5480008 on Build GRCh38
rs4416742 Chr.7: 5479911-5479911 on Build GRCh38
rs757157504 Chr.7: 5479945-5479945 on Build GRCh38
rs980943657 Chr.7: 5479762-5479762 on Build GRCh38
rs539577091 Chr.7: 5480031-5480031 on Build GRCh38
rs951809264 Chr.7: 5480143-5480143 on Build GRCh38
rs923588949 Chr.7: 5480091-5480091 on Build GRCh38
rs1050578416 Chr.7: 5480169-5480169 on Build GRCh38
rs1009974502 Chr.7: 5480019-5480019 on Build GRCh38
rs545795342 Chr.7: 5479842-5479842 on Build GRCh38
rs766760218 Chr.7: 5479928-5479928 on Build GRCh38
rs750700019 Chr.7: 5479995-5479995 on Build GRCh38
rs952130476 Chr.7: 5479910-5479910 on Build GRCh38
rs549780606 Chr.7: 5479878-5479878 on Build GRCh38
rs996336352 Chr.7: 5479850-5479850 on Build GRCh38
rs774605155 Chr.7: 5479946-5479946 on Build GRCh38
rs944652568 Chr.7: 5479996-5479996 on Build GRCh38
rs927858860 Chr.7: 5480020-5480020 on Build GRCh38
rs773488021 Chr.7: 5479867-5479867 on Build GRCh38
rs972145221 Chr.7: 5480133-5480133 on Build GRCh38
rs916946200 Chr.7: 5479862-5479862 on Build GRCh38
rs1040674901 Chr.7: 5480005-5480005 on Build GRCh38
rs1016253917 Chr.7: 5480119-5480119 on Build GRCh38
rs1043900209 Chr.7: 5480087-5480087 on Build GRCh38
rs1040677633 Chr.7: 5479825-5479825 on Build GRCh38
rs930358534 Chr.7: 5479952-5479952 on Build GRCh38
rs999771327 Chr.7: 5479772-5479772 on Build GRCh38
rs990633995 Chr.7: 5480041-5480042 on Build GRCh38
rs763931201 Chr.7: 5479899-5479901 on Build GRCh38
rs569683390 Chr.7: 5480072-5480072 on Build GRCh38
rs1033809126 Chr.7: 5479769-5479769 on Build GRCh38
rs759639438 Chr.7: 5480042-5480042 on Build GRCh38
rs968241110 Chr.7: 5479750-5479750 on Build GRCh38
rs1050241821 Chr.7: 5479958-5479958 on Build GRCh38
rs537048606 Chr.7: 5480113-5480113 on Build GRCh38
rs555745762 Chr.7: 5480114-5480114 on Build GRCh38
rs1013877692 Chr.7: 5480102-5480102 on Build GRCh38
rs1002265133 Chr.7: 5480067-5480067 on Build GRCh38
rs985828292 Chr.7: 5480150-5480150 on Build GRCh38
rs564106967 Chr.7: 5479859-5479859 on Build GRCh38
rs531349879 Chr.7: 5479865-5479865 on Build GRCh38
rs557843442 Chr.7: 5480068-5480068 on Build GRCh38
rs903941842 Chr.7: 5480064-5480064 on Build GRCh38
rs767949800 Chr.7: 5480090-5480090 on Build GRCh38
rs547615601 Chr.7: 5479987-5479987 on Build GRCh38
rs961618579 Chr.7: 5480131-5480131 on Build GRCh38
rs762085266 Chr.7: 5479742-5479742 on Build GRCh38

View Full Product Details