Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6023270-6023766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIMP2
Entrez Gene ID
7965
Gene Name
aminoacyl tRNA synthetase complex interacting multifunctional protein 2
Gene Aliases
JTV-1, JTV1, P38
UniGene
Hs.301613
Gene Chromosome Location
Chr.7: 6009241-6023834 on Build GRCh38
Transcripts
NM_001326606.1 NM_001326607.1 NM_001326609.1 NM_001326611.1 NM_006303.3 XM_005249847.3 XM_005249848.2 XM_005249850.3 XM_006715772.2 XM_011515519.1 NM_001134335.1 NM_014413.3

Gene Symbol
EIF2AK1
Entrez Gene ID
27102
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 1
Gene Aliases
HCR, HRI
UniGene
Hs.520205
Gene Chromosome Location
Chr.7: 6022247-6059229 on Build GRCh38
Transcripts
NM_001326606.1 NM_001326607.1 NM_001326609.1 NM_001326611.1 NM_006303.3 XM_005249847.3 XM_005249848.2 XM_005249850.3 XM_006715772.2 XM_011515519.1 NM_001134335.1 NM_014413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7086154 c.872G>T p.C291F Chr.7: 6023600-6023600 on Build GRCh38
4430250 c.756A>C p.K252N Chr.7: 6023484-6023484 on Build GRCh38
1182325 c.941C>T p.T314M Chr.7: 6023669-6023669 on Build GRCh38
1187256 c.790G>A p.G264R Chr.7: 6023518-6023518 on Build GRCh38
4562271 c.919G>A p.E307K Chr.7: 6023647-6023647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924663778 Chr.7: 6023430-6023430 on Build GRCh38
rs770859885 Chr.7: 6023457-6023457 on Build GRCh38
rs982080297 Chr.7: 6023353-6023353 on Build GRCh38
rs766467029 Chr.7: 6023336-6023336 on Build GRCh38
rs760946189 Chr.7: 6023497-6023497 on Build GRCh38
rs377482895 Chr.7: 6023371-6023371 on Build GRCh38
rs755792627 Chr.7: 6023714-6023714 on Build GRCh38
rs777600343 Chr.7: 6023444-6023444 on Build GRCh38
rs772716032 Chr.7: 6023394-6023394 on Build GRCh38
rs150874145 Chr.7: 6023383-6023383 on Build GRCh38
rs749728733 Chr.7: 6023366-6023366 on Build GRCh38
rs780972148 Chr.7: 6023543-6023543 on Build GRCh38
rs775502742 Chr.7: 6023556-6023556 on Build GRCh38
rs367931587 Chr.7: 6023592-6023592 on Build GRCh38
rs753104633 Chr.7: 6023511-6023511 on Build GRCh38
rs760568809 Chr.7: 6023396-6023396 on Build GRCh38
rs371322289 Chr.7: 6023460-6023460 on Build GRCh38
rs758622796 Chr.7: 6023410-6023411 on Build GRCh38
rs751740812 Chr.7: 6023349-6023349 on Build GRCh38
rs777357626 Chr.7: 6023519-6023519 on Build GRCh38
rs747742165 Chr.7: 6023442-6023442 on Build GRCh38
rs750657628 Chr.7: 6023491-6023491 on Build GRCh38
rs775245397 Chr.7: 6023485-6023485 on Build GRCh38
rs11544566 Chr.7: 6023350-6023350 on Build GRCh38
rs765484017 Chr.7: 6023656-6023656 on Build GRCh38
rs909484169 Chr.7: 6023471-6023471 on Build GRCh38
rs752740782 Chr.7: 6023408-6023409 on Build GRCh38
rs750210959 Chr.7: 6023685-6023698 on Build GRCh38
rs542661636 Chr.7: 6023443-6023443 on Build GRCh38
rs759399018 Chr.7: 6023472-6023472 on Build GRCh38
rs768623646 Chr.7: 6023617-6023617 on Build GRCh38
rs765449737 Chr.7: 6023581-6023581 on Build GRCh38
rs760716532 Chr.7: 6023573-6023573 on Build GRCh38
rs909201219 Chr.7: 6023715-6023715 on Build GRCh38
rs763343312 Chr.7: 6023670-6023670 on Build GRCh38
rs760123492 Chr.7: 6023381-6023384 on Build GRCh38
rs751316385 Chr.7: 6023674-6023674 on Build GRCh38
rs201388339 Chr.7: 6023610-6023610 on Build GRCh38
rs780570820 Chr.7: 6023361-6023361 on Build GRCh38
rs773350468 Chr.7: 6023570-6023570 on Build GRCh38
rs746501522 Chr.7: 6023535-6023535 on Build GRCh38
rs368295654 Chr.7: 6023553-6023553 on Build GRCh38
rs754239135 Chr.7: 6023502-6023502 on Build GRCh38
rs768114421 Chr.7: 6023302-6023302 on Build GRCh38
rs777894287 Chr.7: 6023496-6023496 on Build GRCh38
rs770771902 Chr.7: 6023536-6023536 on Build GRCh38
rs756833002 Chr.7: 6023682-6023682 on Build GRCh38
rs998944405 Chr.7: 6023550-6023550 on Build GRCh38
rs770328913 Chr.7: 6023295-6023295 on Build GRCh38
rs372238513 Chr.7: 6023669-6023669 on Build GRCh38
rs765070549 Chr.7: 6023407-6023407 on Build GRCh38
rs189587013 Chr.7: 6023589-6023589 on Build GRCh38
rs950807957 Chr.7: 6023697-6023697 on Build GRCh38
rs199673160 Chr.7: 6023698-6023698 on Build GRCh38
rs997976538 Chr.7: 6023531-6023531 on Build GRCh38
rs140208111 Chr.7: 6023447-6023447 on Build GRCh38
rs753779282 Chr.7: 6023339-6023339 on Build GRCh38
rs760513787 Chr.7: 6023326-6023326 on Build GRCh38
rs746216887 Chr.7: 6023294-6023294 on Build GRCh38
rs145632958 Chr.7: 6023498-6023498 on Build GRCh38
rs764803060 Chr.7: 6023415-6023415 on Build GRCh38
rs775958032 Chr.7: 6023296-6023296 on Build GRCh38
rs758885310 Chr.7: 6023585-6023585 on Build GRCh38
rs755273615 Chr.7: 6023503-6023503 on Build GRCh38
rs1041316079 Chr.7: 6023434-6023434 on Build GRCh38
rs375888796 Chr.7: 6023505-6023505 on Build GRCh38
rs4560 Chr.7: 6023652-6023652 on Build GRCh38
rs761417289 Chr.7: 6023379-6023379 on Build GRCh38
rs755957474 Chr.7: 6023698-6023699 on Build GRCh38
rs767064905 Chr.7: 6023313-6023313 on Build GRCh38
rs756474990 Chr.7: 6023416-6023416 on Build GRCh38
rs370354397 Chr.7: 6023417-6023417 on Build GRCh38
rs374123170 Chr.7: 6023563-6023563 on Build GRCh38
rs565466567 Chr.7: 6023602-6023602 on Build GRCh38
rs371505435 Chr.7: 6023618-6023618 on Build GRCh38
rs752074098 Chr.7: 6023676-6023676 on Build GRCh38
rs758577601 Chr.7: 6023343-6023343 on Build GRCh38
rs575274304 Chr.7: 6023411-6023411 on Build GRCh38
rs757926814 Chr.7: 6023515-6023515 on Build GRCh38
rs769821226 Chr.7: 6023473-6023473 on Build GRCh38
rs370941772 Chr.7: 6023391-6023391 on Build GRCh38
rs901996125 Chr.7: 6023509-6023509 on Build GRCh38
rs766312290 Chr.7: 6023668-6023668 on Build GRCh38
rs746283944 Chr.7: 6023355-6023355 on Build GRCh38
rs748822585 Chr.7: 6023300-6023300 on Build GRCh38
rs761988750 Chr.7: 6023487-6023487 on Build GRCh38
rs79993082 Chr.7: 6023688-6023688 on Build GRCh38
rs776710794 Chr.7: 6023403-6023403 on Build GRCh38
rs754007971 Chr.7: 6023576-6023576 on Build GRCh38
rs147246097 Chr.7: 6023554-6023554 on Build GRCh38
rs952349709 Chr.7: 6023299-6023299 on Build GRCh38
rs745445513 Chr.7: 6023593-6023593 on Build GRCh38
rs150887968 Chr.7: 6023372-6023372 on Build GRCh38
rs375897273 Chr.7: 6023637-6023637 on Build GRCh38
rs762745274 Chr.7: 6023562-6023562 on Build GRCh38
rs139398981 Chr.7: 6023405-6023405 on Build GRCh38
rs991294520 Chr.7: 6023337-6023337 on Build GRCh38
rs765318758 Chr.7: 6023405-6023406 on Build GRCh38
rs749107344 Chr.7: 6023611-6023611 on Build GRCh38
rs142387037 Chr.7: 6023314-6023314 on Build GRCh38
rs759973375 Chr.7: 6023578-6023578 on Build GRCh38
rs1022792814 Chr.7: 6023615-6023615 on Build GRCh38
rs751152417 Chr.7: 6023590-6023590 on Build GRCh38
rs893082791 Chr.7: 6023566-6023566 on Build GRCh38
rs747111139 Chr.7: 6023622-6023622 on Build GRCh38
rs869312968 Chr.7: 6023301-6023301 on Build GRCh38
rs767430426 Chr.7: 6023488-6023488 on Build GRCh38
rs753260652 Chr.7: 6023583-6023583 on Build GRCh38
rs756425703 Chr.7: 6023356-6023356 on Build GRCh38
rs368862792 Chr.7: 6023646-6023646 on Build GRCh38
rs755955838 Chr.7: 6023604-6023604 on Build GRCh38
rs779256399 Chr.7: 6023436-6023436 on Build GRCh38
rs761471943 Chr.7: 6023307-6023307 on Build GRCh38
rs776366708 Chr.7: 6023306-6023306 on Build GRCh38
rs868626436 Chr.7: 6023420-6023420 on Build GRCh38
rs558931280 Chr.7: 6023448-6023448 on Build GRCh38
rs369386259 Chr.7: 6023401-6023401 on Build GRCh38
rs749640780 Chr.7: 6023422-6023422 on Build GRCh38
rs751703423 Chr.7: 6023410-6023410 on Build GRCh38
rs926430593 Chr.7: 6023354-6023354 on Build GRCh38
rs374243514 Chr.7: 6023431-6023431 on Build GRCh38

View Full Product Details