Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6023657-6024164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIMP2
Entrez Gene ID
7965
Gene Name
aminoacyl tRNA synthetase complex interacting multifunctional protein 2
Gene Aliases
JTV-1, JTV1, P38
UniGene
Hs.301613
Gene Chromosome Location
Chr.7: 6009241-6023834 on Build GRCh38
Transcripts
NM_001326606.1 NM_001326607.1 NM_001326609.1 NM_001326611.1 NM_006303.3 XM_005249847.3 XM_005249848.2 XM_005249850.3 XM_006715772.2 XM_011515519.1 NM_001134335.1 NM_014413.3

Gene Symbol
EIF2AK1
Entrez Gene ID
27102
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 1
Gene Aliases
HCR, HRI
UniGene
Hs.520205
Gene Chromosome Location
Chr.7: 6022247-6059229 on Build GRCh38
Transcripts
NM_001326606.1 NM_001326607.1 NM_001326609.1 NM_001326611.1 NM_006303.3 XM_005249847.3 XM_005249848.2 XM_005249850.3 XM_006715772.2 XM_011515519.1 NM_001134335.1 NM_014413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4162333 c.1624T>C p.S542P Chr.7: 6023976-6023976 on Build GRCh38
6358558 c.1594G>A p.G532S Chr.7: 6024006-6024006 on Build GRCh38
1182325 c.941C>T p.T314M Chr.7: 6023669-6023669 on Build GRCh38
6905622 c.1720G>A p.V574I Chr.7: 6023880-6023880 on Build GRCh38
6284152 c.941C>G p.T314R Chr.7: 6023669-6023669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779534750 Chr.7: 6023747-6023747 on Build GRCh38
rs977878162 Chr.7: 6023972-6023972 on Build GRCh38
rs775012804 Chr.7: 6023816-6023816 on Build GRCh38
rs895055568 Chr.7: 6023938-6023938 on Build GRCh38
rs933557931 Chr.7: 6024020-6024020 on Build GRCh38
rs966550020 Chr.7: 6023964-6023964 on Build GRCh38
rs538466939 Chr.7: 6023835-6023835 on Build GRCh38
rs769880127 Chr.7: 6023897-6023897 on Build GRCh38
rs538382623 Chr.7: 6024094-6024094 on Build GRCh38
rs965721997 Chr.7: 6023990-6023990 on Build GRCh38
rs752527505 Chr.7: 6024141-6024141 on Build GRCh38
rs755792627 Chr.7: 6023714-6023714 on Build GRCh38
rs748008909 Chr.7: 6023752-6023752 on Build GRCh38
rs777895363 Chr.7: 6023871-6023871 on Build GRCh38
rs906157945 Chr.7: 6023907-6023907 on Build GRCh38
rs974700848 Chr.7: 6024016-6024016 on Build GRCh38
rs182045910 Chr.7: 6023833-6023833 on Build GRCh38
rs776400887 Chr.7: 6023881-6023881 on Build GRCh38
rs755957474 Chr.7: 6023698-6023699 on Build GRCh38
rs765844577 Chr.7: 6024017-6024017 on Build GRCh38
rs566553389 Chr.7: 6023792-6023792 on Build GRCh38
rs548198703 Chr.7: 6023749-6023749 on Build GRCh38
rs768503983 Chr.7: 6023787-6023790 on Build GRCh38
rs572400687 Chr.7: 6023806-6023806 on Build GRCh38
rs564832184 Chr.7: 6024117-6024117 on Build GRCh38
rs780145663 Chr.7: 6023900-6023900 on Build GRCh38
rs778617752 Chr.7: 6023840-6023840 on Build GRCh38
rs780898963 Chr.7: 6023805-6023805 on Build GRCh38
rs754791603 Chr.7: 6023768-6023768 on Build GRCh38
rs758166559 Chr.7: 6024143-6024143 on Build GRCh38
rs780826243 Chr.7: 6023816-6023816 on Build GRCh38
rs752475994 Chr.7: 6023843-6023843 on Build GRCh38
rs764864999 Chr.7: 6024123-6024123 on Build GRCh38
rs750210959 Chr.7: 6023685-6023698 on Build GRCh38
rs907090084 Chr.7: 6023896-6023896 on Build GRCh38
rs762472321 Chr.7: 6023820-6023820 on Build GRCh38
rs745955641 Chr.7: 6023772-6023772 on Build GRCh38
rs984875443 Chr.7: 6023943-6023943 on Build GRCh38
rs534281259 Chr.7: 6023825-6023825 on Build GRCh38
rs909201219 Chr.7: 6023715-6023715 on Build GRCh38
rs12657 Chr.7: 6024043-6024043 on Build GRCh38
rs924113142 Chr.7: 6024069-6024069 on Build GRCh38
rs745981867 Chr.7: 6023884-6023884 on Build GRCh38
rs866852204 Chr.7: 6023746-6023746 on Build GRCh38
rs150073939 Chr.7: 6023826-6023826 on Build GRCh38
rs866254180 Chr.7: 6023940-6023940 on Build GRCh38
rs79993082 Chr.7: 6023688-6023688 on Build GRCh38
rs779832748 Chr.7: 6023779-6023780 on Build GRCh38
rs771007640 Chr.7: 6023761-6023761 on Build GRCh38
rs759241074 Chr.7: 6024070-6024070 on Build GRCh38
rs777732966 Chr.7: 6023872-6023872 on Build GRCh38
rs191574702 Chr.7: 6024007-6024007 on Build GRCh38
rs749390730 Chr.7: 6023989-6023989 on Build GRCh38
rs112928096 Chr.7: 6024006-6024006 on Build GRCh38
rs775756518 Chr.7: 6023775-6023775 on Build GRCh38
rs776632578 Chr.7: 6023763-6023763 on Build GRCh38
rs557188645 Chr.7: 6023918-6023918 on Build GRCh38
rs763444661 Chr.7: 6023776-6023776 on Build GRCh38
rs769948642 Chr.7: 6023774-6023774 on Build GRCh38
rs575634604 Chr.7: 6023931-6023931 on Build GRCh38
rs779591426 Chr.7: 6024104-6024104 on Build GRCh38
rs756833002 Chr.7: 6023682-6023682 on Build GRCh38
rs749187455 Chr.7: 6023778-6023779 on Build GRCh38
rs765967124 Chr.7: 6023834-6023834 on Build GRCh38
rs199673160 Chr.7: 6023698-6023698 on Build GRCh38
rs540320232 Chr.7: 6024116-6024116 on Build GRCh38
rs950807957 Chr.7: 6023697-6023697 on Build GRCh38
rs933485276 Chr.7: 6023777-6023777 on Build GRCh38
rs940216360 Chr.7: 6024096-6024096 on Build GRCh38
rs953313479 Chr.7: 6023976-6023976 on Build GRCh38
rs761509596 Chr.7: 6023946-6023946 on Build GRCh38
rs749464292 Chr.7: 6023911-6023911 on Build GRCh38
rs1037148566 Chr.7: 6024126-6024126 on Build GRCh38

View Full Product Details