Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 7234137-7234683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1GALT1
Entrez Gene ID
56913
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1
Gene Aliases
C1GALT, T-synthase
UniGene
Hs.239666
Gene Chromosome Location
Chr.7: 7182547-7248651 on Build GRCh38
Transcripts
NM_020156.4 XM_005249812.1 XM_011515453.2 XM_011515455.2 XM_011515456.2 XM_017012442.1 XM_017012443.1 XM_017012444.1 XM_017012445.1 XM_017012446.1 XM_017012447.1 XM_017012448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667204 c.64T>G p.L22V Chr.7: 7234383-7234383 on Build GRCh38
7106789 c.19C>A p.L7M Chr.7: 7234338-7234338 on Build GRCh38
4943243 c.11A>T p.K4I Chr.7: 7234330-7234330 on Build GRCh38
4667205 c.64T>G p.L22V Chr.7: 7234383-7234383 on Build GRCh38
7106788 c.19C>A p.L7M Chr.7: 7234338-7234338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009591228 Chr.7: 7234473-7234473 on Build GRCh38
rs966732702 Chr.7: 7234266-7234266 on Build GRCh38
rs768018532 Chr.7: 7234451-7234451 on Build GRCh38
rs950949167 Chr.7: 7234653-7234653 on Build GRCh38
rs762672813 Chr.7: 7234298-7234299 on Build GRCh38
rs1049737698 Chr.7: 7234230-7234230 on Build GRCh38
rs997609571 Chr.7: 7234616-7234616 on Build GRCh38
rs748778799 Chr.7: 7234350-7234350 on Build GRCh38
rs141201666 Chr.7: 7234210-7234210 on Build GRCh38
rs921840095 Chr.7: 7234215-7234215 on Build GRCh38
rs748868614 Chr.7: 7234469-7234469 on Build GRCh38
rs756077488 Chr.7: 7234372-7234372 on Build GRCh38
rs892360639 Chr.7: 7234471-7234471 on Build GRCh38
rs982389318 Chr.7: 7234371-7234371 on Build GRCh38
rs780924772 Chr.7: 7234611-7234611 on Build GRCh38
rs562927964 Chr.7: 7234474-7234474 on Build GRCh38
rs549012527 Chr.7: 7234528-7234528 on Build GRCh38
rs973225180 Chr.7: 7234535-7234535 on Build GRCh38
rs754464848 Chr.7: 7234472-7234472 on Build GRCh38
rs746409682 Chr.7: 7234339-7234339 on Build GRCh38
rs770921505 Chr.7: 7234495-7234495 on Build GRCh38
rs775586138 Chr.7: 7234436-7234436 on Build GRCh38
rs919474600 Chr.7: 7234309-7234309 on Build GRCh38
rs756536780 Chr.7: 7234546-7234546 on Build GRCh38
rs376079423 Chr.7: 7234314-7234314 on Build GRCh38
rs773037397 Chr.7: 7234489-7234489 on Build GRCh38
rs767819822 Chr.7: 7234519-7234519 on Build GRCh38
rs757548433 Chr.7: 7234333-7234333 on Build GRCh38
rs1008167112 Chr.7: 7234248-7234248 on Build GRCh38
rs539217147 Chr.7: 7234239-7234239 on Build GRCh38
rs751734811 Chr.7: 7234439-7234439 on Build GRCh38
rs1007637771 Chr.7: 7234562-7234562 on Build GRCh38
rs750887923 Chr.7: 7234521-7234521 on Build GRCh38
rs1043215864 Chr.7: 7234331-7234331 on Build GRCh38
rs557503354 Chr.7: 7234251-7234253 on Build GRCh38
rs746772627 Chr.7: 7234492-7234492 on Build GRCh38
rs944018484 Chr.7: 7234659-7234659 on Build GRCh38
rs973369774 Chr.7: 7234200-7234200 on Build GRCh38
rs768425996 Chr.7: 7234366-7234366 on Build GRCh38
rs73674698 Chr.7: 7234635-7234635 on Build GRCh38
rs770377590 Chr.7: 7234340-7234340 on Build GRCh38
rs772949203 Chr.7: 7234421-7234421 on Build GRCh38
rs770725584 Chr.7: 7234607-7234607 on Build GRCh38
rs976304029 Chr.7: 7234304-7234304 on Build GRCh38
rs747784046 Chr.7: 7234481-7234481 on Build GRCh38
rs895466171 Chr.7: 7234447-7234447 on Build GRCh38
rs747774911 Chr.7: 7234418-7234418 on Build GRCh38
rs1009537386 Chr.7: 7234227-7234227 on Build GRCh38
rs747694469 Chr.7: 7234615-7234618 on Build GRCh38
rs1056404741 Chr.7: 7234456-7234456 on Build GRCh38
rs11976941 Chr.7: 7234216-7234216 on Build GRCh38
rs575865189 Chr.7: 7234523-7234523 on Build GRCh38
rs912525230 Chr.7: 7234231-7234231 on Build GRCh38
rs764112430 Chr.7: 7234515-7234515 on Build GRCh38
rs199619343 Chr.7: 7234512-7234512 on Build GRCh38
rs766796754 Chr.7: 7234560-7234560 on Build GRCh38
rs572875400 Chr.7: 7234313-7234313 on Build GRCh38
rs535326385 Chr.7: 7234218-7234218 on Build GRCh38
rs780706152 Chr.7: 7234463-7234463 on Build GRCh38
rs530324928 Chr.7: 7234573-7234573 on Build GRCh38
rs761991666 Chr.7: 7234516-7234516 on Build GRCh38
rs778692773 Chr.7: 7234603-7234606 on Build GRCh38
rs372878537 Chr.7: 7234561-7234561 on Build GRCh38
rs963544253 Chr.7: 7234195-7234195 on Build GRCh38
rs1053608739 Chr.7: 7234255-7234255 on Build GRCh38
rs773866551 Chr.7: 7234380-7234380 on Build GRCh38
rs753658786 Chr.7: 7234303-7234304 on Build GRCh38
rs751445107 Chr.7: 7234546-7234550 on Build GRCh38
rs776536050 Chr.7: 7234496-7234496 on Build GRCh38
rs746864136 Chr.7: 7234606-7234606 on Build GRCh38
rs926475675 Chr.7: 7234430-7234430 on Build GRCh38
rs781554903 Chr.7: 7234334-7234334 on Build GRCh38
rs35945804 Chr.7: 7234213-7234214 on Build GRCh38
rs764434666 Chr.7: 7234438-7234438 on Build GRCh38
rs1017760962 Chr.7: 7234249-7234249 on Build GRCh38
rs544738309 Chr.7: 7234462-7234462 on Build GRCh38
rs771703576 Chr.7: 7234488-7234488 on Build GRCh38
rs778305710 Chr.7: 7234583-7234583 on Build GRCh38
rs758024658 Chr.7: 7234602-7234602 on Build GRCh38
rs947529408 Chr.7: 7234315-7234315 on Build GRCh38
rs752721656 Chr.7: 7234282-7234282 on Build GRCh38
rs367725837 Chr.7: 7234449-7234449 on Build GRCh38
rs753720249 Chr.7: 7234603-7234603 on Build GRCh38
rs565313805 Chr.7: 7234420-7234420 on Build GRCh38
rs146951691 Chr.7: 7234233-7234233 on Build GRCh38
rs41282685 Chr.7: 7234423-7234423 on Build GRCh38
rs112027213 Chr.7: 7234429-7234429 on Build GRCh38

View Full Product Details