Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 7238114-7238611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1GALT1
Entrez Gene ID
56913
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1
Gene Aliases
C1GALT, T-synthase
UniGene
Hs.239666
Gene Chromosome Location
Chr.7: 7182547-7248651 on Build GRCh38
Transcripts
NM_020156.4 XM_005249812.1 XM_011515453.2 XM_011515455.2 XM_011515456.2 XM_017012442.1 XM_017012443.1 XM_017012444.1 XM_017012445.1 XM_017012446.1 XM_017012447.1 XM_017012448.1 XM_017012449.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4006756 c.359T>G p.F120C Chr.7: 7238393-7238393 on Build GRCh38
6614986 c.558C>T p.Y186Y Chr.7: 7238592-7238592 on Build GRCh38
3833169 c.547C>A p.L183I Chr.7: 7238581-7238581 on Build GRCh38
6420680 c.331T>C p.W111R Chr.7: 7238365-7238365 on Build GRCh38
7087681 c.419G>C p.R140T Chr.7: 7238453-7238453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754178446 Chr.7: 7238267-7238267 on Build GRCh38
rs771348475 Chr.7: 7238444-7238444 on Build GRCh38
rs375614206 Chr.7: 7238352-7238352 on Build GRCh38
rs766223917 Chr.7: 7238549-7238549 on Build GRCh38
rs769684271 Chr.7: 7238295-7238295 on Build GRCh38
rs201415283 Chr.7: 7238560-7238560 on Build GRCh38
rs533762942 Chr.7: 7238520-7238520 on Build GRCh38
rs368614486 Chr.7: 7238463-7238463 on Build GRCh38
rs755008390 Chr.7: 7238557-7238557 on Build GRCh38
rs745643466 Chr.7: 7238292-7238292 on Build GRCh38
rs763383692 Chr.7: 7238362-7238362 on Build GRCh38
rs761004658 Chr.7: 7238347-7238347 on Build GRCh38
rs1021390895 Chr.7: 7238208-7238208 on Build GRCh38
rs372580480 Chr.7: 7238501-7238501 on Build GRCh38
rs753262522 Chr.7: 7238360-7238360 on Build GRCh38
rs149009811 Chr.7: 7238337-7238337 on Build GRCh38
rs371322872 Chr.7: 7238291-7238291 on Build GRCh38
rs200796468 Chr.7: 7238515-7238515 on Build GRCh38
rs763615701 Chr.7: 7238370-7238370 on Build GRCh38
rs918623819 Chr.7: 7238380-7238380 on Build GRCh38
rs780891272 Chr.7: 7238403-7238403 on Build GRCh38
rs554704948 Chr.7: 7238442-7238442 on Build GRCh38
rs756942307 Chr.7: 7238388-7238388 on Build GRCh38
rs757372781 Chr.7: 7238564-7238564 on Build GRCh38
rs754922077 Chr.7: 7238439-7238439 on Build GRCh38
rs775677218 Chr.7: 7238478-7238478 on Build GRCh38
rs759354816 Chr.7: 7238218-7238218 on Build GRCh38
rs779644412 Chr.7: 7238429-7238429 on Build GRCh38
rs145917767 Chr.7: 7238344-7238344 on Build GRCh38
rs138478625 Chr.7: 7238550-7238550 on Build GRCh38
rs761950300 Chr.7: 7238220-7238220 on Build GRCh38
rs376057308 Chr.7: 7238460-7238460 on Build GRCh38
rs76986838 Chr.7: 7238168-7238168 on Build GRCh38
rs763499580 Chr.7: 7238494-7238494 on Build GRCh38
rs772483424 Chr.7: 7238232-7238232 on Build GRCh38
rs189360329 Chr.7: 7238320-7238320 on Build GRCh38
rs549075288 Chr.7: 7238152-7238152 on Build GRCh38
rs377075847 Chr.7: 7238561-7238561 on Build GRCh38
rs765742047 Chr.7: 7238271-7238271 on Build GRCh38
rs765724131 Chr.7: 7238355-7238355 on Build GRCh38
rs941015438 Chr.7: 7238240-7238240 on Build GRCh38
rs747057464 Chr.7: 7238443-7238443 on Build GRCh38
rs773916621 Chr.7: 7238500-7238500 on Build GRCh38
rs760918216 Chr.7: 7238262-7238262 on Build GRCh38
rs760007297 Chr.7: 7238269-7238269 on Build GRCh38
rs766877598 Chr.7: 7238266-7238266 on Build GRCh38
rs184256604 Chr.7: 7238204-7238204 on Build GRCh38
rs756854286 Chr.7: 7238286-7238286 on Build GRCh38
rs755947827 Chr.7: 7238422-7238422 on Build GRCh38
rs3807862 Chr.7: 7238176-7238176 on Build GRCh38
rs181239665 Chr.7: 7238413-7238413 on Build GRCh38
rs909573375 Chr.7: 7238226-7238226 on Build GRCh38
rs567415913 Chr.7: 7238319-7238319 on Build GRCh38
rs982323845 Chr.7: 7238144-7238144 on Build GRCh38
rs764552307 Chr.7: 7238498-7238498 on Build GRCh38
rs61742008 Chr.7: 7238274-7238274 on Build GRCh38
rs755678129 Chr.7: 7238524-7238524 on Build GRCh38
rs906945863 Chr.7: 7238553-7238553 on Build GRCh38
rs757888910 Chr.7: 7238280-7238280 on Build GRCh38
rs931536997 Chr.7: 7238416-7238416 on Build GRCh38
rs751217992 Chr.7: 7238281-7238281 on Build GRCh38
rs751046391 Chr.7: 7238376-7238376 on Build GRCh38
rs746020800 Chr.7: 7238461-7238461 on Build GRCh38
rs766876792 Chr.7: 7238502-7238502 on Build GRCh38
rs572963593 Chr.7: 7238511-7238511 on Build GRCh38
rs773585975 Chr.7: 7238236-7238236 on Build GRCh38
rs771260286 Chr.7: 7238349-7238349 on Build GRCh38
rs112501792 Chr.7: 7238572-7238572 on Build GRCh38
rs750015544 Chr.7: 7238270-7238270 on Build GRCh38
rs767692066 Chr.7: 7238374-7238374 on Build GRCh38
rs749265368 Chr.7: 7238433-7238433 on Build GRCh38
rs568910070 Chr.7: 7238353-7238353 on Build GRCh38

View Full Product Details