Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 7243396-7243903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1GALT1
Entrez Gene ID
56913
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1
Gene Aliases
C1GALT, T-synthase
UniGene
Hs.239666
Gene Chromosome Location
Chr.7: 7182547-7248651 on Build GRCh38
Transcripts
NM_020156.4 XM_005249812.1 XM_011515453.2 XM_011515455.2 XM_011515456.2 XM_017012442.1 XM_017012443.1 XM_017012444.1 XM_017012445.1 XM_017012446.1 XM_017012447.1 XM_017012448.1 XM_017012449.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3663304 c.990T>A p.G330G Chr.7: 7243625-7243625 on Build GRCh38
3640337 c.966C>T p.L322L Chr.7: 7243601-7243601 on Build GRCh38
6431133 c.904T>C p.S302P Chr.7: 7243539-7243539 on Build GRCh38
381595 c.896G>T p.G299V Chr.7: 7243531-7243531 on Build GRCh38
7173352 c.892C>A p.P298T Chr.7: 7243527-7243527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750791945 Chr.7: 7243769-7243769 on Build GRCh38
rs764390159 Chr.7: 7243579-7243579 on Build GRCh38
rs753352734 Chr.7: 7243700-7243700 on Build GRCh38
rs749452892 Chr.7: 7243704-7243704 on Build GRCh38
rs768329939 Chr.7: 7243619-7243619 on Build GRCh38
rs755688614 Chr.7: 7243604-7243604 on Build GRCh38
rs765304552 Chr.7: 7243565-7243565 on Build GRCh38
rs762282109 Chr.7: 7243686-7243686 on Build GRCh38
rs771861888 Chr.7: 7243712-7243712 on Build GRCh38
rs1049955726 Chr.7: 7243833-7243833 on Build GRCh38
rs760543119 Chr.7: 7243656-7243656 on Build GRCh38
rs745366600 Chr.7: 7243495-7243495 on Build GRCh38
rs138563382 Chr.7: 7243580-7243580 on Build GRCh38
rs747935652 Chr.7: 7243648-7243648 on Build GRCh38
rs17165546 Chr.7: 7243870-7243870 on Build GRCh38
rs373753284 Chr.7: 7243486-7243486 on Build GRCh38
rs1010577590 Chr.7: 7243449-7243449 on Build GRCh38
rs112532087 Chr.7: 7243420-7243420 on Build GRCh38
rs761124676 Chr.7: 7243771-7243771 on Build GRCh38
rs150563643 Chr.7: 7243575-7243575 on Build GRCh38
rs932808547 Chr.7: 7243820-7243820 on Build GRCh38
rs762310151 Chr.7: 7243633-7243633 on Build GRCh38
rs776762746 Chr.7: 7243515-7243515 on Build GRCh38
rs765265066 Chr.7: 7243659-7243659 on Build GRCh38
rs146864914 Chr.7: 7243841-7243841 on Build GRCh38
rs779575456 Chr.7: 7243606-7243606 on Build GRCh38
rs748903624 Chr.7: 7243505-7243505 on Build GRCh38
rs532562658 Chr.7: 7243702-7243702 on Build GRCh38
rs775016594 Chr.7: 7243504-7243504 on Build GRCh38
rs772780005 Chr.7: 7243457-7243457 on Build GRCh38
rs764346763 Chr.7: 7243676-7243676 on Build GRCh38
rs375911604 Chr.7: 7243661-7243661 on Build GRCh38
rs777844760 Chr.7: 7243660-7243660 on Build GRCh38
rs772060079 Chr.7: 7243649-7243649 on Build GRCh38
rs371801368 Chr.7: 7243863-7243863 on Build GRCh38
rs752027092 Chr.7: 7243679-7243679 on Build GRCh38
rs747744965 Chr.7: 7243707-7243707 on Build GRCh38
rs142799381 Chr.7: 7243692-7243692 on Build GRCh38
rs774144250 Chr.7: 7243508-7243508 on Build GRCh38
rs993376937 Chr.7: 7243879-7243879 on Build GRCh38
rs748912790 Chr.7: 7243615-7243615 on Build GRCh38
rs777507166 Chr.7: 7243722-7243722 on Build GRCh38
rs769627516 Chr.7: 7243499-7243499 on Build GRCh38
rs768014718 Chr.7: 7243761-7243761 on Build GRCh38
rs757880763 Chr.7: 7243599-7243599 on Build GRCh38
rs763084610 Chr.7: 7243739-7243739 on Build GRCh38
rs762085810 Chr.7: 7243760-7243760 on Build GRCh38
rs1008665801 Chr.7: 7243852-7243852 on Build GRCh38
rs961933173 Chr.7: 7243488-7243488 on Build GRCh38
rs1018998397 Chr.7: 7243869-7243869 on Build GRCh38
rs950189406 Chr.7: 7243683-7243683 on Build GRCh38
rs953194838 Chr.7: 7243469-7243469 on Build GRCh38
rs774050675 Chr.7: 7243647-7243647 on Build GRCh38
rs774718616 Chr.7: 7243754-7243754 on Build GRCh38
rs776366321 Chr.7: 7243637-7243637 on Build GRCh38
rs759603023 Chr.7: 7243531-7243531 on Build GRCh38
rs781761008 Chr.7: 7243600-7243600 on Build GRCh38
rs867576111 Chr.7: 7243817-7243817 on Build GRCh38
rs76676805 Chr.7: 7243693-7243693 on Build GRCh38
rs746319437 Chr.7: 7243601-7243601 on Build GRCh38
rs768365045 Chr.7: 7243507-7243507 on Build GRCh38
rs374068731 Chr.7: 7243653-7243653 on Build GRCh38
rs541042133 Chr.7: 7243742-7243742 on Build GRCh38
rs779476030 Chr.7: 7243695-7243695 on Build GRCh38
rs758608455 Chr.7: 7243578-7243578 on Build GRCh38
rs573123670 Chr.7: 7243734-7243734 on Build GRCh38
rs767924740 Chr.7: 7243688-7243688 on Build GRCh38
rs761783974 Chr.7: 7243509-7243509 on Build GRCh38
rs117309561 Chr.7: 7243789-7243789 on Build GRCh38
rs893265876 Chr.7: 7243763-7243763 on Build GRCh38
rs754650842 Chr.7: 7243703-7243703 on Build GRCh38
rs768559591 Chr.7: 7243741-7243741 on Build GRCh38
rs770881808 Chr.7: 7243730-7243730 on Build GRCh38
rs767408386 Chr.7: 7243512-7243512 on Build GRCh38
rs369861038 Chr.7: 7243481-7243481 on Build GRCh38
rs200061330 Chr.7: 7243478-7243478 on Build GRCh38
rs752869659 Chr.7: 7243571-7243571 on Build GRCh38

View Full Product Details