Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 7246898-7247417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1GALT1
Entrez Gene ID
56913
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1
Gene Aliases
C1GALT, T-synthase
UniGene
Hs.239666
Gene Chromosome Location
Chr.7: 7182547-7248651 on Build GRCh38
Transcripts
NM_020156.4 XM_005249812.1 XM_011515453.2 XM_011515455.2 XM_011515456.2 XM_017012442.1 XM_017012443.1 XM_017012444.1 XM_017012445.1 XM_017012446.1 XM_017012447.1 XM_017012448.1 XM_017012449.1

SNP Detail

SNP ID Chromosome Location
rs556707585 Chr.7: 7247377-7247377 on Build GRCh38
rs925403450 Chr.7: 7247086-7247086 on Build GRCh38
rs962579327 Chr.7: 7247002-7247002 on Build GRCh38
rs1004501039 Chr.7: 7247140-7247140 on Build GRCh38
rs925544696 Chr.7: 7247250-7247250 on Build GRCh38
rs1002484119 Chr.7: 7247012-7247012 on Build GRCh38
rs566832735 Chr.7: 7247053-7247053 on Build GRCh38
rs765053465 Chr.7: 7247132-7247132 on Build GRCh38
rs965357304 Chr.7: 7247267-7247267 on Build GRCh38
rs1004147284 Chr.7: 7246970-7246970 on Build GRCh38
rs1033494848 Chr.7: 7247214-7247214 on Build GRCh38
rs62450012 Chr.7: 7247139-7247139 on Build GRCh38
rs898438104 Chr.7: 7247159-7247159 on Build GRCh38
rs570936811 Chr.7: 7247147-7247147 on Build GRCh38
rs182697508 Chr.7: 7247300-7247300 on Build GRCh38
rs536298024 Chr.7: 7247093-7247094 on Build GRCh38
rs772106428 Chr.7: 7246995-7246995 on Build GRCh38
rs547380234 Chr.7: 7247019-7247019 on Build GRCh38
rs1038289631 Chr.7: 7247150-7247150 on Build GRCh38
rs569704961 Chr.7: 7247201-7247201 on Build GRCh38
rs145740201 Chr.7: 7247121-7247121 on Build GRCh38
rs1039639210 Chr.7: 7247094-7247094 on Build GRCh38
rs766767428 Chr.7: 7247353-7247353 on Build GRCh38
rs1050722411 Chr.7: 7247135-7247135 on Build GRCh38
rs189918095 Chr.7: 7247059-7247059 on Build GRCh38
rs886783281 Chr.7: 7247138-7247138 on Build GRCh38
rs933695537 Chr.7: 7247360-7247360 on Build GRCh38
rs979876383 Chr.7: 7247245-7247245 on Build GRCh38
rs966683507 Chr.7: 7247242-7247242 on Build GRCh38
rs528821933 Chr.7: 7247060-7247060 on Build GRCh38
rs537398016 Chr.7: 7247330-7247330 on Build GRCh38
rs775286875 Chr.7: 7247323-7247323 on Build GRCh38
rs112867379 Chr.7: 7247287-7247287 on Build GRCh38
rs960754380 Chr.7: 7247217-7247217 on Build GRCh38
rs1033759067 Chr.7: 7247020-7247020 on Build GRCh38
rs373828361 Chr.7: 7246967-7246967 on Build GRCh38
rs1020877210 Chr.7: 7247241-7247241 on Build GRCh38
rs979716182 Chr.7: 7247064-7247064 on Build GRCh38
rs1002029838 Chr.7: 7247203-7247203 on Build GRCh38
rs750808246 Chr.7: 7247224-7247224 on Build GRCh38
rs920997376 Chr.7: 7247114-7247114 on Build GRCh38

View Full Product Details