Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 7969401-7969902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLCCI1
Entrez Gene ID
113263
Gene Name
glucocorticoid induced 1
Gene Aliases
FAM117C, GCTR, GIG18, TSSN1
UniGene
Hs.131673
Gene Chromosome Location
Chr.7: 7968743-8089080 on Build GRCh38
Transcripts
NM_138426.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5682585 c.343G>C p.A115P Chr.7: 7969693-7969693 on Build GRCh38
5986336 c.96C>A p.P32P Chr.7: 7969446-7969446 on Build GRCh38
5732344 c.106G>T p.A36S Chr.7: 7969456-7969456 on Build GRCh38
5737868 c.323G>A p.S108N Chr.7: 7969673-7969673 on Build GRCh38
5445823 c.145_146insGCG p.G48_V49insG Chr.7: 7969495-7969496 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045891550 Chr.7: 7969824-7969824 on Build GRCh38
rs752800143 Chr.7: 7969839-7969839 on Build GRCh38
rs777839965 Chr.7: 7969420-7969420 on Build GRCh38
rs911951801 Chr.7: 7969797-7969797 on Build GRCh38
rs751993129 Chr.7: 7969426-7969426 on Build GRCh38
rs543875458 Chr.7: 7969838-7969838 on Build GRCh38
rs892088045 Chr.7: 7969639-7969639 on Build GRCh38
rs1006862581 Chr.7: 7969481-7969482 on Build GRCh38
rs761465295 Chr.7: 7969733-7969733 on Build GRCh38
rs11731 Chr.7: 7969769-7969769 on Build GRCh38
rs1003553266 Chr.7: 7969444-7969444 on Build GRCh38
rs754601634 Chr.7: 7969769-7969780 on Build GRCh38
rs930199784 Chr.7: 7969820-7969820 on Build GRCh38
rs768214939 Chr.7: 7969697-7969697 on Build GRCh38
rs779525035 Chr.7: 7969645-7969645 on Build GRCh38
rs1053762973 Chr.7: 7969632-7969632 on Build GRCh38
rs935182503 Chr.7: 7969798-7969798 on Build GRCh38
rs889836208 Chr.7: 7969666-7969666 on Build GRCh38
rs965965715 Chr.7: 7969504-7969504 on Build GRCh38
rs552933583 Chr.7: 7969491-7969491 on Build GRCh38
rs572463389 Chr.7: 7969542-7969542 on Build GRCh38
rs144036658 Chr.7: 7969446-7969446 on Build GRCh38
rs994218301 Chr.7: 7969719-7969719 on Build GRCh38
rs757751716 Chr.7: 7969431-7969431 on Build GRCh38
rs984550018 Chr.7: 7969791-7969791 on Build GRCh38
rs950603789 Chr.7: 7969522-7969522 on Build GRCh38
rs1019630165 Chr.7: 7969485-7969485 on Build GRCh38
rs530365789 Chr.7: 7969479-7969480 on Build GRCh38
rs753549912 Chr.7: 7969721-7969721 on Build GRCh38
rs1044014359 Chr.7: 7969570-7969570 on Build GRCh38
rs1052251439 Chr.7: 7969826-7969826 on Build GRCh38
rs1044047733 Chr.7: 7969424-7969424 on Build GRCh38
rs781703395 Chr.7: 7969435-7969435 on Build GRCh38
rs938388219 Chr.7: 7969620-7969620 on Build GRCh38
rs928301269 Chr.7: 7969599-7969599 on Build GRCh38
rs1034800868 Chr.7: 7969859-7969859 on Build GRCh38
rs886847672 Chr.7: 7969864-7969864 on Build GRCh38
rs891947790 Chr.7: 7969458-7969458 on Build GRCh38
rs893759649 Chr.7: 7969834-7969834 on Build GRCh38
rs907423800 Chr.7: 7969425-7969425 on Build GRCh38
rs773241200 Chr.7: 7969786-7969786 on Build GRCh38
rs1006998430 Chr.7: 7969674-7969674 on Build GRCh38
rs1021717468 Chr.7: 7969474-7969474 on Build GRCh38
rs984622146 Chr.7: 7969523-7969523 on Build GRCh38
rs950671174 Chr.7: 7969732-7969732 on Build GRCh38
rs1019599024 Chr.7: 7969688-7969688 on Build GRCh38
rs766215827 Chr.7: 7969836-7969836 on Build GRCh38
rs770228032 Chr.7: 7969571-7969571 on Build GRCh38
rs116731086 Chr.7: 7969849-7969849 on Build GRCh38
rs541216640 Chr.7: 7969480-7969480 on Build GRCh38
rs545986371 Chr.7: 7969649-7969649 on Build GRCh38
rs760346550 Chr.7: 7969799-7969799 on Build GRCh38
rs974374127 Chr.7: 7969438-7969438 on Build GRCh38
rs976419070 Chr.7: 7969517-7969517 on Build GRCh38
rs13246559 Chr.7: 7969704-7969704 on Build GRCh38
rs531112603 Chr.7: 7969612-7969614 on Build GRCh38
rs948474188 Chr.7: 7969551-7969551 on Build GRCh38
rs865995749 Chr.7: 7969745-7969745 on Build GRCh38
rs1011773453 Chr.7: 7969463-7969463 on Build GRCh38
rs564447167 Chr.7: 7969700-7969700 on Build GRCh38
rs979806896 Chr.7: 7969848-7969848 on Build GRCh38
rs763164206 Chr.7: 7969847-7969847 on Build GRCh38
rs919203914 Chr.7: 7969518-7969518 on Build GRCh38
rs527406638 Chr.7: 7969854-7969854 on Build GRCh38
rs531755418 Chr.7: 7969761-7969761 on Build GRCh38

View Full Product Details