Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 13891818-13892342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV1
Entrez Gene ID
2115
Gene Name
ETS variant 1
Gene Aliases
ER81
UniGene
Hs.22634
Gene Chromosome Location
Chr.7: 13891229-13991425 on Build GRCh38
Transcripts
NM_001163147.1 NM_001163148.1 NM_001163149.1 NM_001163150.1 NM_001163151.1 NM_001163152.1 NM_004956.4 NR_120445.1

SNP Detail

SNP ID Chromosome Location
rs1002602819 Chr.7: 13892128-13892128 on Build GRCh38
rs1005004970 Chr.7: 13891892-13891892 on Build GRCh38
rs764458873 Chr.7: 13892077-13892077 on Build GRCh38
rs966094330 Chr.7: 13892294-13892294 on Build GRCh38
rs77729573 Chr.7: 13892111-13892111 on Build GRCh38
rs199762151 Chr.7: 13892264-13892265 on Build GRCh38
rs545995087 Chr.7: 13892162-13892162 on Build GRCh38
rs1022540330 Chr.7: 13892160-13892160 on Build GRCh38
rs1053767047 Chr.7: 13892004-13892004 on Build GRCh38
rs1049014504 Chr.7: 13892079-13892079 on Build GRCh38
rs553446193 Chr.7: 13891901-13891901 on Build GRCh38
rs762510409 Chr.7: 13891937-13891937 on Build GRCh38
rs899882549 Chr.7: 13891983-13891983 on Build GRCh38
rs754154176 Chr.7: 13892084-13892084 on Build GRCh38
rs181363770 Chr.7: 13892025-13892025 on Build GRCh38
rs959599209 Chr.7: 13891952-13891952 on Build GRCh38
rs987402647 Chr.7: 13892105-13892105 on Build GRCh38
rs17678858 Chr.7: 13891971-13891971 on Build GRCh38
rs918914861 Chr.7: 13892078-13892078 on Build GRCh38
rs144088916 Chr.7: 13892091-13892091 on Build GRCh38
rs1035180539 Chr.7: 13891947-13891947 on Build GRCh38
rs1014679664 Chr.7: 13891911-13891911 on Build GRCh38
rs1027299349 Chr.7: 13891853-13891853 on Build GRCh38
rs1046634538 Chr.7: 13892272-13892272 on Build GRCh38
rs968738296 Chr.7: 13892161-13892161 on Build GRCh38
rs976177640 Chr.7: 13892298-13892298 on Build GRCh38
rs1033787486 Chr.7: 13892316-13892316 on Build GRCh38
rs974543881 Chr.7: 13892138-13892138 on Build GRCh38
rs959546941 Chr.7: 13892145-13892145 on Build GRCh38
rs970481893 Chr.7: 13891852-13891852 on Build GRCh38
rs991052486 Chr.7: 13891970-13891970 on Build GRCh38
rs539623436 Chr.7: 13891892-13891900 on Build GRCh38
rs531468497 Chr.7: 13892273-13892273 on Build GRCh38
rs114044639 Chr.7: 13892045-13892045 on Build GRCh38
rs1025124979 Chr.7: 13891848-13891848 on Build GRCh38
rs944813645 Chr.7: 13892041-13892041 on Build GRCh38
rs75783477 Chr.7: 13892151-13892151 on Build GRCh38
rs1004550708 Chr.7: 13892087-13892087 on Build GRCh38
rs923959512 Chr.7: 13892233-13892233 on Build GRCh38
rs982237818 Chr.7: 13891941-13891941 on Build GRCh38
rs1002341363 Chr.7: 13892297-13892297 on Build GRCh38
rs887463045 Chr.7: 13891881-13891881 on Build GRCh38
rs548465513 Chr.7: 13892315-13892315 on Build GRCh38
rs186108918 Chr.7: 13892060-13892060 on Build GRCh38
rs946747659 Chr.7: 13891999-13891999 on Build GRCh38
rs1039810939 Chr.7: 13891976-13891976 on Build GRCh38
rs973059968 Chr.7: 13891927-13891927 on Build GRCh38
rs191305402 Chr.7: 13892009-13892009 on Build GRCh38
rs753246986 Chr.7: 13891986-13891986 on Build GRCh38
rs796369614 Chr.7: 13892035-13892035 on Build GRCh38
rs944142747 Chr.7: 13891973-13891973 on Build GRCh38
rs562722674 Chr.7: 13892265-13892265 on Build GRCh38
rs189741754 Chr.7: 13892142-13892142 on Build GRCh38
rs111680196 Chr.7: 13892043-13892043 on Build GRCh38
rs926330396 Chr.7: 13891912-13891912 on Build GRCh38
rs892459922 Chr.7: 13892185-13892185 on Build GRCh38
rs570251391 Chr.7: 13891922-13891922 on Build GRCh38

View Full Product Details