Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 14146977-14147489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKB
Entrez Gene ID
1607
Gene Name
diacylglycerol kinase beta
Gene Aliases
DAGK2, DGK, DGK-BETA
UniGene
Hs.567255
Gene Chromosome Location
Chr.7: 14145049-14974777 on Build GRCh38
Transcripts
NM_004080.2

SNP Detail

SNP ID Chromosome Location
rs146548258 Chr.7: 14147263-14147263 on Build GRCh38
rs950308310 Chr.7: 14147184-14147191 on Build GRCh38
rs762141387 Chr.7: 14147435-14147435 on Build GRCh38
rs182442409 Chr.7: 14147404-14147404 on Build GRCh38
rs879305498 Chr.7: 14147140-14147140 on Build GRCh38
rs555587856 Chr.7: 14147191-14147191 on Build GRCh38
rs879933471 Chr.7: 14147350-14147350 on Build GRCh38
rs968404657 Chr.7: 14147369-14147369 on Build GRCh38
rs1005766774 Chr.7: 14147246-14147246 on Build GRCh38
rs1038216706 Chr.7: 14147029-14147029 on Build GRCh38
rs200398922 Chr.7: 14147219-14147219 on Build GRCh38
rs558180441 Chr.7: 14147179-14147179 on Build GRCh38
rs937409357 Chr.7: 14147240-14147240 on Build GRCh38
rs117097030 Chr.7: 14147349-14147349 on Build GRCh38
rs972063250 Chr.7: 14147378-14147378 on Build GRCh38
rs1020425061 Chr.7: 14147377-14147377 on Build GRCh38
rs978170637 Chr.7: 14147143-14147143 on Build GRCh38
rs568671834 Chr.7: 14147068-14147068 on Build GRCh38
rs573277366 Chr.7: 14147370-14147370 on Build GRCh38
rs746278575 Chr.7: 14147366-14147369 on Build GRCh38
rs760594562 Chr.7: 14147415-14147415 on Build GRCh38
rs1018361319 Chr.7: 14147357-14147357 on Build GRCh38
rs1008289357 Chr.7: 14147121-14147121 on Build GRCh38
rs989053958 Chr.7: 14147169-14147169 on Build GRCh38
rs757580375 Chr.7: 14147299-14147299 on Build GRCh38
rs991076767 Chr.7: 14147336-14147336 on Build GRCh38
rs80210789 Chr.7: 14147196-14147196 on Build GRCh38
rs777724374 Chr.7: 14147079-14147079 on Build GRCh38
rs772562973 Chr.7: 14147380-14147380 on Build GRCh38
rs774515320 Chr.7: 14147270-14147270 on Build GRCh38
rs946629389 Chr.7: 14147142-14147142 on Build GRCh38
rs548339801 Chr.7: 14147065-14147065 on Build GRCh38
rs139426546 Chr.7: 14147007-14147007 on Build GRCh38
rs1035051921 Chr.7: 14147324-14147324 on Build GRCh38
rs957528213 Chr.7: 14147158-14147159 on Build GRCh38
rs919989665 Chr.7: 14147434-14147434 on Build GRCh38
rs73679436 Chr.7: 14147038-14147038 on Build GRCh38
rs183761277 Chr.7: 14147044-14147044 on Build GRCh38
rs957560903 Chr.7: 14147375-14147375 on Build GRCh38
rs144181064 Chr.7: 14147134-14147134 on Build GRCh38
rs931711609 Chr.7: 14147419-14147419 on Build GRCh38
rs891054392 Chr.7: 14147335-14147335 on Build GRCh38
rs1047350782 Chr.7: 14147461-14147461 on Build GRCh38
rs1040131880 Chr.7: 14147181-14147181 on Build GRCh38
rs888698748 Chr.7: 14147227-14147227 on Build GRCh38
rs996209324 Chr.7: 14147033-14147034 on Build GRCh38
rs569361796 Chr.7: 14147025-14147025 on Build GRCh38
rs899506149 Chr.7: 14147360-14147360 on Build GRCh38
rs1003606655 Chr.7: 14147279-14147279 on Build GRCh38
rs771242377 Chr.7: 14147226-14147226 on Build GRCh38
rs368035299 Chr.7: 14147376-14147376 on Build GRCh38
rs563925535 Chr.7: 14147006-14147006 on Build GRCh38
rs541561443 Chr.7: 14147330-14147330 on Build GRCh38
rs555581590 Chr.7: 14147426-14147426 on Build GRCh38
rs910042664 Chr.7: 14147464-14147464 on Build GRCh38
rs773696251 Chr.7: 14147144-14147144 on Build GRCh38
rs1015554503 Chr.7: 14147020-14147020 on Build GRCh38
rs930019605 Chr.7: 14147455-14147455 on Build GRCh38
rs13224657 Chr.7: 14147411-14147411 on Build GRCh38
rs191246895 Chr.7: 14147021-14147021 on Build GRCh38
rs898363967 Chr.7: 14147247-14147247 on Build GRCh38
rs955096655 Chr.7: 14147121-14147122 on Build GRCh38

View Full Product Details