Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 75811596-75812090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL24
Entrez Gene ID
6369
Gene Name
C-C motif chemokine ligand 24
Gene Aliases
Ckb-6, MPIF-2, MPIF2, SCYA24
UniGene
Hs.247838
Gene Chromosome Location
Chr.7: 75810825-75823356 on Build GRCh38
Transcripts
NM_002991.2 XM_011516459.2 XM_011516460.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4802289 c.192-1G>T p.? Chr.7: 75811965-75811965 on Build GRCh38
1698765 c.298G>A p.A100T Chr.7: 75811858-75811858 on Build GRCh38
4802379 c.192-2A>T p.? Chr.7: 75811966-75811966 on Build GRCh38
5935236 c.192-6C>T p.? Chr.7: 75811970-75811970 on Build GRCh38
1735390 c.271G>A p.A91T Chr.7: 75811885-75811885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373981058 Chr.7: 75811875-75811875 on Build GRCh38
rs782415521 Chr.7: 75812007-75812007 on Build GRCh38
rs530225351 Chr.7: 75811820-75811820 on Build GRCh38
rs781855935 Chr.7: 75811770-75811770 on Build GRCh38
rs782667707 Chr.7: 75811783-75811783 on Build GRCh38
rs782337842 Chr.7: 75811806-75811806 on Build GRCh38
rs782361483 Chr.7: 75811879-75811879 on Build GRCh38
rs1010478183 Chr.7: 75811930-75811930 on Build GRCh38
rs1057099239 Chr.7: 75811693-75811693 on Build GRCh38
rs930187371 Chr.7: 75811672-75811672 on Build GRCh38
rs782466627 Chr.7: 75811762-75811762 on Build GRCh38
rs563856016 Chr.7: 75811715-75811715 on Build GRCh38
rs903910519 Chr.7: 75811719-75811719 on Build GRCh38
rs1052931222 Chr.7: 75811895-75811895 on Build GRCh38
rs782647071 Chr.7: 75812000-75812000 on Build GRCh38
rs545492035 Chr.7: 75812038-75812038 on Build GRCh38
rs782287551 Chr.7: 75811797-75811797 on Build GRCh38
rs11465312 Chr.7: 75811852-75811852 on Build GRCh38
rs116910667 Chr.7: 75811659-75811659 on Build GRCh38
rs781832289 Chr.7: 75811862-75811862 on Build GRCh38
rs558869841 Chr.7: 75811870-75811870 on Build GRCh38
rs910938912 Chr.7: 75811617-75811617 on Build GRCh38
rs781840346 Chr.7: 75811756-75811756 on Build GRCh38
rs372921195 Chr.7: 75812054-75812054 on Build GRCh38
rs782148035 Chr.7: 75811892-75811892 on Build GRCh38
rs112210342 Chr.7: 75811766-75811766 on Build GRCh38
rs782784012 Chr.7: 75811765-75811765 on Build GRCh38
rs956054515 Chr.7: 75811749-75811749 on Build GRCh38
rs973805177 Chr.7: 75811815-75811815 on Build GRCh38
rs1056665833 Chr.7: 75811851-75811851 on Build GRCh38
rs781810964 Chr.7: 75811941-75811941 on Build GRCh38
rs574681153 Chr.7: 75811621-75811621 on Build GRCh38
rs145957811 Chr.7: 75811915-75811915 on Build GRCh38
rs375998761 Chr.7: 75811748-75811748 on Build GRCh38
rs539982344 Chr.7: 75812063-75812063 on Build GRCh38
rs537064463 Chr.7: 75811857-75811857 on Build GRCh38
rs948312212 Chr.7: 75811858-75811858 on Build GRCh38
rs782072710 Chr.7: 75811751-75811751 on Build GRCh38
rs782616979 Chr.7: 75811969-75811969 on Build GRCh38
rs918763287 Chr.7: 75811632-75811632 on Build GRCh38
rs11465315 Chr.7: 75811790-75811790 on Build GRCh38
rs200259580 Chr.7: 75811931-75811931 on Build GRCh38
rs782430642 Chr.7: 75811802-75811802 on Build GRCh38
rs377250178 Chr.7: 75812046-75812046 on Build GRCh38
rs11465316 Chr.7: 75811711-75811711 on Build GRCh38
rs782184701 Chr.7: 75811784-75811784 on Build GRCh38
rs549659544 Chr.7: 75812037-75812037 on Build GRCh38
rs11465314 Chr.7: 75811793-75811793 on Build GRCh38
rs782792607 Chr.7: 75811935-75811935 on Build GRCh38
rs782763075 Chr.7: 75811861-75811861 on Build GRCh38
rs1029499696 Chr.7: 75811943-75811943 on Build GRCh38
rs782201883 Chr.7: 75811840-75811840 on Build GRCh38
rs782523132 Chr.7: 75811999-75811999 on Build GRCh38
rs903877934 Chr.7: 75811865-75811865 on Build GRCh38
rs202145310 Chr.7: 75811886-75811886 on Build GRCh38
rs143309023 Chr.7: 75811838-75811838 on Build GRCh38
rs782042253 Chr.7: 75811796-75811796 on Build GRCh38
rs199828405 Chr.7: 75811958-75811958 on Build GRCh38
rs782690263 Chr.7: 75811755-75811755 on Build GRCh38
rs955446524 Chr.7: 75811947-75811947 on Build GRCh38
rs1038521967 Chr.7: 75811630-75811630 on Build GRCh38
rs11465317 Chr.7: 75811622-75811622 on Build GRCh38
rs1045055256 Chr.7: 75811716-75811716 on Build GRCh38
rs372206947 Chr.7: 75811844-75811844 on Build GRCh38
rs560716730 Chr.7: 75811780-75811780 on Build GRCh38
rs143613901 Chr.7: 75811996-75811996 on Build GRCh38
rs782724968 Chr.7: 75811928-75811928 on Build GRCh38
rs145220760 Chr.7: 75811814-75811814 on Build GRCh38
rs575462933 Chr.7: 75812031-75812031 on Build GRCh38
rs545757007 Chr.7: 75811714-75811714 on Build GRCh38
rs973772581 Chr.7: 75812034-75812034 on Build GRCh38
rs782002431 Chr.7: 75811836-75811836 on Build GRCh38
rs1029991279 Chr.7: 75811776-75811776 on Build GRCh38
rs570959258 Chr.7: 75811880-75811880 on Build GRCh38
rs962351297 Chr.7: 75812032-75812032 on Build GRCh38
rs11465313 Chr.7: 75811828-75811828 on Build GRCh38
rs371147438 Chr.7: 75811789-75811789 on Build GRCh38
rs528075437 Chr.7: 75812056-75812056 on Build GRCh38
rs1000925298 Chr.7: 75811723-75811723 on Build GRCh38
rs201733507 Chr.7: 75811925-75811925 on Build GRCh38
rs782574637 Chr.7: 75812008-75812008 on Build GRCh38
rs755938564 Chr.7: 75811977-75811977 on Build GRCh38
rs782286069 Chr.7: 75812006-75812006 on Build GRCh38
rs782164458 Chr.7: 75811668-75811668 on Build GRCh38
rs896771687 Chr.7: 75811704-75811704 on Build GRCh38
rs782373030 Chr.7: 75811884-75811884 on Build GRCh38
rs782325742 Chr.7: 75811900-75811900 on Build GRCh38

View Full Product Details