Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76502214-76502725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTX2
Entrez Gene ID
113878
Gene Name
deltex E3 ubiquitin ligase 2
Gene Aliases
RNF58
UniGene
Hs.187058
Gene Chromosome Location
Chr.7: 76461655-76505995 on Build GRCh38
Transcripts
NM_001102594.1 NM_001102595.1 NM_001102596.1 NM_020892.2 XM_005250131.1 XM_005250132.1 XM_011515772.1 XM_011515773.2 XM_011515774.1 XM_011515775.1 XM_017011723.1 XM_017011724.1 XM_017011725.1 XM_017011726.1 XM_017011727.1 XM_017011728.1 XM_017011730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5520160 c.1389+3G>A p.? Chr.7: 76502459-76502459 on Build GRCh38
1686029 c.1348C>T p.H450Y Chr.7: 76502415-76502415 on Build GRCh38
4162457 c.1261A>G p.T421A Chr.7: 76502328-76502328 on Build GRCh38
5358370 c.1279G>A p.D427N Chr.7: 76502346-76502346 on Build GRCh38
370237 c.1336A>T p.S446C Chr.7: 76502403-76502403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759574652 Chr.7: 76502465-76502465 on Build GRCh38
rs772781142 Chr.7: 76502317-76502319 on Build GRCh38
rs751764209 Chr.7: 76502338-76502338 on Build GRCh38
rs754340583 Chr.7: 76502267-76502267 on Build GRCh38
rs952404723 Chr.7: 76502679-76502679 on Build GRCh38
rs6963784 Chr.7: 76502495-76502495 on Build GRCh38
rs151048323 Chr.7: 76502369-76502369 on Build GRCh38
rs578105583 Chr.7: 76502273-76502273 on Build GRCh38
rs148587797 Chr.7: 76502332-76502332 on Build GRCh38
rs778870566 Chr.7: 76502353-76502353 on Build GRCh38
rs761570621 Chr.7: 76502473-76502473 on Build GRCh38
rs755973929 Chr.7: 76502345-76502345 on Build GRCh38
rs779910965 Chr.7: 76502359-76502359 on Build GRCh38
rs369153706 Chr.7: 76502526-76502526 on Build GRCh38
rs532326466 Chr.7: 76502433-76502433 on Build GRCh38
rs141623125 Chr.7: 76502389-76502389 on Build GRCh38
rs769130113 Chr.7: 76502315-76502315 on Build GRCh38
rs771569668 Chr.7: 76502315-76502315 on Build GRCh38
rs532258927 Chr.7: 76502683-76502683 on Build GRCh38
rs376209701 Chr.7: 76502312-76502312 on Build GRCh38
rs1031787044 Chr.7: 76502537-76502537 on Build GRCh38
rs762997466 Chr.7: 76502373-76502373 on Build GRCh38
rs766376964 Chr.7: 76502481-76502481 on Build GRCh38
rs975076842 Chr.7: 76502629-76502629 on Build GRCh38
rs773707841 Chr.7: 76502371-76502371 on Build GRCh38
rs6964085 Chr.7: 76502657-76502657 on Build GRCh38
rs956254090 Chr.7: 76502346-76502346 on Build GRCh38
rs754477568 Chr.7: 76502484-76502484 on Build GRCh38
rs780763932 Chr.7: 76502485-76502485 on Build GRCh38
rs4728775 Chr.7: 76502378-76502378 on Build GRCh38
rs774439426 Chr.7: 76502448-76502448 on Build GRCh38
rs765101680 Chr.7: 76502382-76502382 on Build GRCh38
rs891959419 Chr.7: 76502555-76502555 on Build GRCh38
rs547899687 Chr.7: 76502477-76502477 on Build GRCh38
rs772539201 Chr.7: 76502252-76502252 on Build GRCh38
rs757012199 Chr.7: 76502379-76502379 on Build GRCh38
rs539330659 Chr.7: 76502500-76502500 on Build GRCh38
rs372062282 Chr.7: 76502527-76502527 on Build GRCh38
rs771655815 Chr.7: 76502305-76502305 on Build GRCh38
rs181043779 Chr.7: 76502472-76502472 on Build GRCh38
rs779329603 Chr.7: 76502496-76502496 on Build GRCh38
rs201157714 Chr.7: 76502367-76502367 on Build GRCh38
rs150141824 Chr.7: 76502445-76502445 on Build GRCh38
rs775217433 Chr.7: 76502308-76502308 on Build GRCh38
rs764703677 Chr.7: 76502259-76502259 on Build GRCh38
rs1019524502 Chr.7: 76502534-76502534 on Build GRCh38
rs185596389 Chr.7: 76502280-76502280 on Build GRCh38
rs576502359 Chr.7: 76502619-76502619 on Build GRCh38
rs778181397 Chr.7: 76502494-76502494 on Build GRCh38
rs369195858 Chr.7: 76502447-76502447 on Build GRCh38
rs750705032 Chr.7: 76502278-76502278 on Build GRCh38
rs561238228 Chr.7: 76502354-76502354 on Build GRCh38
rs569297323 Chr.7: 76502492-76502492 on Build GRCh38
rs901870704 Chr.7: 76502543-76502543 on Build GRCh38
rs778207414 Chr.7: 76502434-76502434 on Build GRCh38
rs931075990 Chr.7: 76502446-76502446 on Build GRCh38
rs756836378 Chr.7: 76502299-76502299 on Build GRCh38
rs6979487 Chr.7: 76502328-76502328 on Build GRCh38
rs746628260 Chr.7: 76502364-76502364 on Build GRCh38
rs578218650 Chr.7: 76502499-76502499 on Build GRCh38
rs556476889 Chr.7: 76502244-76502244 on Build GRCh38
rs757166814 Chr.7: 76502350-76502350 on Build GRCh38
rs1028424074 Chr.7: 76502665-76502665 on Build GRCh38
rs773195533 Chr.7: 76502331-76502331 on Build GRCh38
rs752490607 Chr.7: 76502281-76502281 on Build GRCh38
rs754706871 Chr.7: 76502426-76502426 on Build GRCh38
rs867126610 Chr.7: 76502256-76502256 on Build GRCh38
rs1044800041 Chr.7: 76502525-76502525 on Build GRCh38
rs200474916 Chr.7: 76502348-76502348 on Build GRCh38
rs141587206 Chr.7: 76502327-76502327 on Build GRCh38
rs139847148 Chr.7: 76502316-76502316 on Build GRCh38
rs780924836 Chr.7: 76502432-76502432 on Build GRCh38
rs6958774 Chr.7: 76502333-76502333 on Build GRCh38
rs776021087 Chr.7: 76502253-76502253 on Build GRCh38
rs369495187 Chr.7: 76502493-76502493 on Build GRCh38
rs777742013 Chr.7: 76502287-76502287 on Build GRCh38
rs759082773 Chr.7: 76502251-76502251 on Build GRCh38
rs2690683 Chr.7: 76502269-76502269 on Build GRCh38
rs908363883 Chr.7: 76502461-76502461 on Build GRCh38
rs779688007 Chr.7: 76502408-76502408 on Build GRCh38
rs545321713 Chr.7: 76502276-76502276 on Build GRCh38
rs543877689 Chr.7: 76502620-76502620 on Build GRCh38
rs766439166 Chr.7: 76502374-76502374 on Build GRCh38
rs543111051 Chr.7: 76502538-76502538 on Build GRCh38
rs556815744 Chr.7: 76502498-76502498 on Build GRCh38
rs373955567 Chr.7: 76502545-76502545 on Build GRCh38
rs376289670 Chr.7: 76502478-76502478 on Build GRCh38
rs144760720 Chr.7: 76502329-76502329 on Build GRCh38
rs774600298 Chr.7: 76502376-76502376 on Build GRCh38
rs749222258 Chr.7: 76502289-76502289 on Build GRCh38
rs745337399 Chr.7: 76502302-76502302 on Build GRCh38
rs760353589 Chr.7: 76502393-76502395 on Build GRCh38
rs755843135 Chr.7: 76502286-76502286 on Build GRCh38
rs753825018 Chr.7: 76502349-76502349 on Build GRCh38
rs751117493 Chr.7: 76502483-76502483 on Build GRCh38
rs371261404 Chr.7: 76502246-76502246 on Build GRCh38
rs73703179 Chr.7: 76502247-76502247 on Build GRCh38
rs184250301 Chr.7: 76502641-76502641 on Build GRCh38
rs759736162 Chr.7: 76502377-76502377 on Build GRCh38
rs778266867 Chr.7: 76502300-76502300 on Build GRCh38
rs948711151 Chr.7: 76502517-76502517 on Build GRCh38
rs561402758 Chr.7: 76502592-76502592 on Build GRCh38
rs751064400 Chr.7: 76502412-76502412 on Build GRCh38
rs758570695 Chr.7: 76502279-76502279 on Build GRCh38
rs760921023 Chr.7: 76502507-76502507 on Build GRCh38
rs550083900 Chr.7: 76502370-76502370 on Build GRCh38
rs147908269 Chr.7: 76502388-76502388 on Build GRCh38
rs983822126 Chr.7: 76502682-76502682 on Build GRCh38

View Full Product Details