Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 80638375-80638900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD36
Entrez Gene ID
948
Gene Name
CD36 molecule
Gene Aliases
BDPLT10, CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3
UniGene
Hs.120949
Gene Chromosome Location
Chr.7: 80602188-80679277 on Build GRCh38
Transcripts
NM_000072.3 NM_001001547.2 NM_001001548.2 NM_001289911.1 XM_005250713.1 XM_005250714.1 XM_005250715.4

SNP Detail

SNP ID Chromosome Location
rs562829246 Chr.7: 80638857-80638857 on Build GRCh38
rs951957542 Chr.7: 80638414-80638414 on Build GRCh38
rs189430423 Chr.7: 80638590-80638590 on Build GRCh38
rs532886092 Chr.7: 80638852-80638852 on Build GRCh38
rs115860397 Chr.7: 80638591-80638591 on Build GRCh38
rs976773684 Chr.7: 80638836-80638836 on Build GRCh38
rs1022660395 Chr.7: 80638481-80638481 on Build GRCh38
rs755852179 Chr.7: 80638840-80638840 on Build GRCh38
rs562381974 Chr.7: 80638810-80638810 on Build GRCh38
rs1022691470 Chr.7: 80638786-80638786 on Build GRCh38
rs139016526 Chr.7: 80638823-80638823 on Build GRCh38
rs887573692 Chr.7: 80638645-80638645 on Build GRCh38
rs34829889 Chr.7: 80638401-80638401 on Build GRCh38
rs1013238015 Chr.7: 80638723-80638723 on Build GRCh38
rs899916688 Chr.7: 80638607-80638607 on Build GRCh38
rs180875189 Chr.7: 80638421-80638421 on Build GRCh38
rs1334512 Chr.7: 80638588-80638588 on Build GRCh38
rs994116285 Chr.7: 80638698-80638698 on Build GRCh38
rs1049313916 Chr.7: 80638720-80638720 on Build GRCh38
rs944873798 Chr.7: 80638628-80638628 on Build GRCh38
rs200693535 Chr.7: 80638580-80638581 on Build GRCh38
rs568010205 Chr.7: 80638410-80638410 on Build GRCh38
rs184290852 Chr.7: 80638573-80638573 on Build GRCh38
rs796662034 Chr.7: 80638588-80638588 on Build GRCh38
rs995949901 Chr.7: 80638400-80638400 on Build GRCh38
rs550828696 Chr.7: 80638812-80638812 on Build GRCh38
rs1041468739 Chr.7: 80638596-80638596 on Build GRCh38
rs1014815415 Chr.7: 80638700-80638700 on Build GRCh38
rs1028814010 Chr.7: 80638401-80638401 on Build GRCh38
rs944319725 Chr.7: 80638825-80638825 on Build GRCh38
rs577249587 Chr.7: 80638627-80638627 on Build GRCh38
rs915268669 Chr.7: 80638584-80638584 on Build GRCh38
rs761716433 Chr.7: 80638674-80638674 on Build GRCh38
rs1027698969 Chr.7: 80638433-80638433 on Build GRCh38
rs971231484 Chr.7: 80638494-80638494 on Build GRCh38
rs373889562 Chr.7: 80638780-80638780 on Build GRCh38
rs917656491 Chr.7: 80638539-80638539 on Build GRCh38
rs528880300 Chr.7: 80638811-80638811 on Build GRCh38
rs116540612 Chr.7: 80638469-80638469 on Build GRCh38
rs114535894 Chr.7: 80638799-80638799 on Build GRCh38
rs1002718820 Chr.7: 80638804-80638804 on Build GRCh38
rs116056927 Chr.7: 80638563-80638563 on Build GRCh38
rs1334513 Chr.7: 80638639-80638639 on Build GRCh38
rs375822934 Chr.7: 80638589-80638589 on Build GRCh38
rs373174848 Chr.7: 80638567-80638567 on Build GRCh38
rs898085086 Chr.7: 80638662-80638662 on Build GRCh38
rs570434635 Chr.7: 80638484-80638484 on Build GRCh38
rs532150098 Chr.7: 80638401-80638402 on Build GRCh38
rs951798040 Chr.7: 80638451-80638451 on Build GRCh38
rs762661522 Chr.7: 80638509-80638509 on Build GRCh38
rs141232342 Chr.7: 80638553-80638553 on Build GRCh38
rs559985262 Chr.7: 80638746-80638746 on Build GRCh38
rs887476702 Chr.7: 80638756-80638756 on Build GRCh38
rs980927112 Chr.7: 80638558-80638558 on Build GRCh38
rs796107324 Chr.7: 80638414-80638414 on Build GRCh38
rs763910324 Chr.7: 80638592-80638592 on Build GRCh38
rs910652268 Chr.7: 80638870-80638870 on Build GRCh38
rs992248270 Chr.7: 80638534-80638534 on Build GRCh38

View Full Product Details