Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 80646009-80646506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD36
Entrez Gene ID
948
Gene Name
CD36 molecule
Gene Aliases
BDPLT10, CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3
UniGene
Hs.120949
Gene Chromosome Location
Chr.7: 80602188-80679277 on Build GRCh38
Transcripts
NM_000072.3 NM_001001547.2 NM_001001548.2 NM_001127443.1 NM_001127444.1 NM_001289909.1 NM_001289911.1 NR_110501.1 XM_005250713.1 XM_005250714.1 XM_005250715.4

SNP Detail

SNP ID Chromosome Location
rs1052902306 Chr.7: 80646049-80646049 on Build GRCh38
rs984191174 Chr.7: 80646108-80646108 on Build GRCh38
rs532897983 Chr.7: 80646121-80646121 on Build GRCh38
rs539977067 Chr.7: 80646345-80646345 on Build GRCh38
rs748819650 Chr.7: 80646391-80646391 on Build GRCh38
rs76530424 Chr.7: 80646055-80646056 on Build GRCh38
rs372978118 Chr.7: 80646365-80646365 on Build GRCh38
rs575034427 Chr.7: 80646268-80646268 on Build GRCh38
rs891634610 Chr.7: 80646052-80646052 on Build GRCh38
rs919167387 Chr.7: 80646184-80646184 on Build GRCh38
rs533572011 Chr.7: 80646069-80646069 on Build GRCh38
rs139292358 Chr.7: 80646301-80646301 on Build GRCh38
rs13306228 Chr.7: 80646203-80646203 on Build GRCh38
rs943390741 Chr.7: 80646412-80646412 on Build GRCh38
rs947322628 Chr.7: 80646066-80646066 on Build GRCh38
rs1053517428 Chr.7: 80646277-80646277 on Build GRCh38
rs911100185 Chr.7: 80646399-80646399 on Build GRCh38
rs945536441 Chr.7: 80646334-80646334 on Build GRCh38
rs980142670 Chr.7: 80646259-80646259 on Build GRCh38
rs561862760 Chr.7: 80646421-80646423 on Build GRCh38
rs1039405841 Chr.7: 80646458-80646458 on Build GRCh38
rs563817445 Chr.7: 80646092-80646092 on Build GRCh38
rs779767644 Chr.7: 80646167-80646167 on Build GRCh38
rs398111494 Chr.7: 80646056-80646057 on Build GRCh38
rs997641810 Chr.7: 80646165-80646165 on Build GRCh38
rs543945638 Chr.7: 80646067-80646067 on Build GRCh38
rs969764402 Chr.7: 80646245-80646245 on Build GRCh38
rs567633234 Chr.7: 80646187-80646187 on Build GRCh38
rs143736571 Chr.7: 80646445-80646445 on Build GRCh38
rs938454052 Chr.7: 80646313-80646313 on Build GRCh38
rs3211805 Chr.7: 80646260-80646260 on Build GRCh38
rs1049654 Chr.7: 80646139-80646139 on Build GRCh38
rs947188111 Chr.7: 80646362-80646362 on Build GRCh38
rs1007948092 Chr.7: 80646113-80646113 on Build GRCh38
rs529130978 Chr.7: 80646175-80646175 on Build GRCh38
rs66573838 Chr.7: 80646049-80646050 on Build GRCh38
rs537473704 Chr.7: 80646239-80646239 on Build GRCh38
rs568330281 Chr.7: 80646236-80646236 on Build GRCh38
rs988554359 Chr.7: 80646320-80646320 on Build GRCh38
rs1017700677 Chr.7: 80646133-80646133 on Build GRCh38
rs982440426 Chr.7: 80646246-80646246 on Build GRCh38
rs55849243 Chr.7: 80646425-80646427 on Build GRCh38
rs566379897 Chr.7: 80646147-80646147 on Build GRCh38
rs901848438 Chr.7: 80646476-80646476 on Build GRCh38
rs1043349832 Chr.7: 80646068-80646068 on Build GRCh38

View Full Product Details