Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 80666250-80666779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD36
Entrez Gene ID
948
Gene Name
CD36 molecule
Gene Aliases
BDPLT10, CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3
UniGene
Hs.120949
Gene Chromosome Location
Chr.7: 80602188-80679277 on Build GRCh38
Transcripts
NM_000072.3 NM_001001547.2 NM_001001548.2 NM_001127443.1 NM_001127444.1 NM_001289908.1 NM_001289909.1 NM_001289911.1 NR_110501.1 XM_005250713.1 XM_005250714.1 XM_005250715.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5551041 c.728G>A p.C243Y Chr.7: 80666469-80666469 on Build GRCh38
5560386 c.735G>A p.M245I Chr.7: 80666476-80666476 on Build GRCh38
4556790 c.702G>A p.R234R Chr.7: 80666443-80666443 on Build GRCh38
6226794 c.738T>C p.I246I Chr.7: 80666479-80666479 on Build GRCh38
5551043 c.728G>A p.C243Y Chr.7: 80666469-80666469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771398906 Chr.7: 80666404-80666404 on Build GRCh38
rs924849660 Chr.7: 80666416-80666416 on Build GRCh38
rs768883824 Chr.7: 80666481-80666481 on Build GRCh38
rs897068273 Chr.7: 80666580-80666580 on Build GRCh38
rs916058632 Chr.7: 80666370-80666370 on Build GRCh38
rs760764041 Chr.7: 80666596-80666596 on Build GRCh38
rs748757724 Chr.7: 80666392-80666392 on Build GRCh38
rs1034533016 Chr.7: 80666681-80666681 on Build GRCh38
rs764236775 Chr.7: 80666425-80666425 on Build GRCh38
rs3212012 Chr.7: 80666385-80666385 on Build GRCh38
rs761829928 Chr.7: 80666427-80666427 on Build GRCh38
rs1027734359 Chr.7: 80666330-80666330 on Build GRCh38
rs760939346 Chr.7: 80666424-80666424 on Build GRCh38
rs976070154 Chr.7: 80666717-80666717 on Build GRCh38
rs146885545 Chr.7: 80666447-80666447 on Build GRCh38
rs777291291 Chr.7: 80666451-80666451 on Build GRCh38
rs149985988 Chr.7: 80666470-80666470 on Build GRCh38
rs897075912 Chr.7: 80666570-80666570 on Build GRCh38
rs755730672 Chr.7: 80666450-80666450 on Build GRCh38
rs542883933 Chr.7: 80666551-80666551 on Build GRCh38
rs756466792 Chr.7: 80666497-80666500 on Build GRCh38
rs755888738 Chr.7: 80666490-80666490 on Build GRCh38
rs764002255 Chr.7: 80666498-80666498 on Build GRCh38
rs972390968 Chr.7: 80666304-80666304 on Build GRCh38
rs563185669 Chr.7: 80666664-80666664 on Build GRCh38
rs961982679 Chr.7: 80666292-80666292 on Build GRCh38
rs185009522 Chr.7: 80666502-80666502 on Build GRCh38
rs927432541 Chr.7: 80666353-80666353 on Build GRCh38
rs745387028 Chr.7: 80666511-80666511 on Build GRCh38
rs753405192 Chr.7: 80666500-80666500 on Build GRCh38
rs748787273 Chr.7: 80666454-80666454 on Build GRCh38
rs761535004 Chr.7: 80666402-80666403 on Build GRCh38
rs143762577 Chr.7: 80666442-80666442 on Build GRCh38
rs149178142 Chr.7: 80666487-80666487 on Build GRCh38
rs913513801 Chr.7: 80666696-80666696 on Build GRCh38
rs993863427 Chr.7: 80666601-80666601 on Build GRCh38
rs758909152 Chr.7: 80666512-80666512 on Build GRCh38
rs754945994 Chr.7: 80666439-80666439 on Build GRCh38
rs767021819 Chr.7: 80666428-80666429 on Build GRCh38
rs993452262 Chr.7: 80666317-80666317 on Build GRCh38
rs767706099 Chr.7: 80666514-80666514 on Build GRCh38
rs993481977 Chr.7: 80666602-80666602 on Build GRCh38
rs192303439 Chr.7: 80666341-80666341 on Build GRCh38
rs772536456 Chr.7: 80666418-80666418 on Build GRCh38
rs775025089 Chr.7: 80666410-80666410 on Build GRCh38
rs1012965090 Chr.7: 80666639-80666639 on Build GRCh38
rs74353859 Chr.7: 80666692-80666692 on Build GRCh38
rs934356369 Chr.7: 80666435-80666435 on Build GRCh38
rs957732970 Chr.7: 80666691-80666691 on Build GRCh38
rs3212013 Chr.7: 80666738-80666738 on Build GRCh38
rs3211920 Chr.7: 80666752-80666752 on Build GRCh38
rs139734451 Chr.7: 80666455-80666455 on Build GRCh38
rs557732736 Chr.7: 80666478-80666478 on Build GRCh38
rs752489479 Chr.7: 80666448-80666448 on Build GRCh38
rs776066023 Chr.7: 80666419-80666419 on Build GRCh38
rs778554273 Chr.7: 80666510-80666510 on Build GRCh38
rs3211919 Chr.7: 80666548-80666548 on Build GRCh38
rs767453540 Chr.7: 80666489-80666489 on Build GRCh38
rs1023562756 Chr.7: 80666658-80666658 on Build GRCh38
rs760172383 Chr.7: 80666412-80666412 on Build GRCh38
rs766255139 Chr.7: 80666433-80666433 on Build GRCh38
rs368916360 Chr.7: 80666436-80666436 on Build GRCh38
rs868695492 Chr.7: 80666491-80666491 on Build GRCh38
rs750858271 Chr.7: 80666452-80666453 on Build GRCh38
rs776740918 Chr.7: 80666484-80666484 on Build GRCh38
rs980322237 Chr.7: 80666350-80666350 on Build GRCh38
rs35793888 Chr.7: 80666469-80666469 on Build GRCh38
rs756808645 Chr.7: 80666463-80666463 on Build GRCh38
rs556402579 Chr.7: 80666343-80666343 on Build GRCh38
rs887877850 Chr.7: 80666634-80666634 on Build GRCh38
rs956263901 Chr.7: 80666708-80666708 on Build GRCh38
rs1027598926 Chr.7: 80666605-80666605 on Build GRCh38
rs111305425 Chr.7: 80666680-80666680 on Build GRCh38
rs971624440 Chr.7: 80666666-80666666 on Build GRCh38
rs775941131 Chr.7: 80666471-80666471 on Build GRCh38
rs762143746 Chr.7: 80666485-80666485 on Build GRCh38
rs41281225 Chr.7: 80666513-80666513 on Build GRCh38
rs145187459 Chr.7: 80666486-80666486 on Build GRCh38

View Full Product Details