Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81964070-81964569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA2D1
Entrez Gene ID
781
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 1
Gene Aliases
CACNA2, CACNL2A, CCHL2A, LINC01112, lncRNA-N3
UniGene
Hs.282151
Gene Chromosome Location
Chr.7: 81946444-82443921 on Build GRCh38
Transcripts
NM_000722.3 XM_005250570.2 XM_005250572.2 XM_005250573.2 XM_005250574.2 XM_006716118.2 XM_006716119.3 XM_006716120.3 XM_006716121.3 XM_011516570.2 XM_011516571.2 XM_011516572.2 XM_017012588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6178716 c.2699A>G p.Q900R Chr.7: 81964235-81964235 on Build GRCh38
7424872 c.2633C>A p.S878* Chr.7: 81964301-81964301 on Build GRCh38
4363443 c.2750G>A p.R917H Chr.7: 81964220-81964220 on Build GRCh38
5032306 c.2691T>G p.S897S Chr.7: 81964279-81964279 on Build GRCh38
4363440 c.2776G>T p.D926Y Chr.7: 81964096-81964096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200274400 Chr.7: 81964192-81964192 on Build GRCh38
rs771624597 Chr.7: 81964253-81964253 on Build GRCh38
rs758571613 Chr.7: 81964112-81964112 on Build GRCh38
rs746860492 Chr.7: 81964219-81964219 on Build GRCh38
rs566320066 Chr.7: 81964146-81964146 on Build GRCh38
rs3735517 Chr.7: 81964138-81964138 on Build GRCh38
rs532517591 Chr.7: 81964126-81964126 on Build GRCh38
rs369327479 Chr.7: 81964157-81964157 on Build GRCh38
rs768238513 Chr.7: 81964153-81964153 on Build GRCh38
rs933300474 Chr.7: 81964466-81964466 on Build GRCh38
rs779639873 Chr.7: 81964144-81964144 on Build GRCh38
rs187849336 Chr.7: 81964106-81964106 on Build GRCh38
rs183290212 Chr.7: 81964368-81964368 on Build GRCh38
rs771501535 Chr.7: 81964394-81964394 on Build GRCh38
rs568107538 Chr.7: 81964174-81964174 on Build GRCh38
rs757217470 Chr.7: 81964123-81964123 on Build GRCh38
rs765175609 Chr.7: 81964195-81964195 on Build GRCh38
rs770266829 Chr.7: 81964163-81964163 on Build GRCh38
rs907776127 Chr.7: 81964504-81964504 on Build GRCh38
rs749817139 Chr.7: 81964230-81964231 on Build GRCh38
rs773868134 Chr.7: 81964166-81964166 on Build GRCh38
rs767702491 Chr.7: 81964181-81964181 on Build GRCh38
rs925743459 Chr.7: 81964130-81964130 on Build GRCh38
rs748874797 Chr.7: 81964393-81964393 on Build GRCh38
rs772889955 Chr.7: 81964251-81964251 on Build GRCh38
rs1015793463 Chr.7: 81964217-81964217 on Build GRCh38
rs760765395 Chr.7: 81964289-81964289 on Build GRCh38
rs1052996296 Chr.7: 81964145-81964145 on Build GRCh38
rs753964891 Chr.7: 81964323-81964323 on Build GRCh38
rs775601344 Chr.7: 81964276-81964276 on Build GRCh38
rs778525285 Chr.7: 81964158-81964158 on Build GRCh38
rs147728344 Chr.7: 81964252-81964252 on Build GRCh38
rs138827246 Chr.7: 81964161-81964161 on Build GRCh38
rs747636283 Chr.7: 81964247-81964247 on Build GRCh38
rs780710873 Chr.7: 81964241-81964241 on Build GRCh38
rs770219374 Chr.7: 81964152-81964160 on Build GRCh38
rs199542348 Chr.7: 81964198-81964198 on Build GRCh38
rs956142469 Chr.7: 81964476-81964476 on Build GRCh38
rs890360772 Chr.7: 81964147-81964147 on Build GRCh38
rs543950666 Chr.7: 81964537-81964537 on Build GRCh38
rs150815216 Chr.7: 81964540-81964540 on Build GRCh38
rs759826842 Chr.7: 81964256-81964256 on Build GRCh38
rs71557107 Chr.7: 81964430-81964430 on Build GRCh38
rs367790770 Chr.7: 81964103-81964103 on Build GRCh38
rs373544416 Chr.7: 81964387-81964387 on Build GRCh38
rs755839246 Chr.7: 81964391-81964391 on Build GRCh38
rs142553634 Chr.7: 81964261-81964261 on Build GRCh38
rs1037541999 Chr.7: 81964543-81964543 on Build GRCh38
rs774855335 Chr.7: 81964254-81964254 on Build GRCh38
rs751287413 Chr.7: 81964371-81964371 on Build GRCh38
rs575022176 Chr.7: 81964467-81964467 on Build GRCh38
rs746269947 Chr.7: 81964120-81964122 on Build GRCh38
rs201782873 Chr.7: 81964191-81964191 on Build GRCh38
rs561899173 Chr.7: 81964461-81964461 on Build GRCh38
rs769232295 Chr.7: 81964368-81964369 on Build GRCh38
rs538712996 Chr.7: 81964199-81964199 on Build GRCh38
rs974772133 Chr.7: 81964431-81964431 on Build GRCh38
rs556022468 Chr.7: 81964342-81964342 on Build GRCh38
rs750388768 Chr.7: 81964199-81964199 on Build GRCh38
rs764123114 Chr.7: 81964310-81964310 on Build GRCh38
rs752262317 Chr.7: 81964388-81964388 on Build GRCh38
rs201755371 Chr.7: 81964224-81964224 on Build GRCh38
rs774906295 Chr.7: 81964171-81964171 on Build GRCh38
rs1013862654 Chr.7: 81964220-81964220 on Build GRCh38
rs769219661 Chr.7: 81964248-81964248 on Build GRCh38
rs763370860 Chr.7: 81964167-81964167 on Build GRCh38
rs745745737 Chr.7: 81964478-81964478 on Build GRCh38
rs758369222 Chr.7: 81964213-81964213 on Build GRCh38
rs17240359 Chr.7: 81964453-81964453 on Build GRCh38
rs532555875 Chr.7: 81964545-81964545 on Build GRCh38
rs766822279 Chr.7: 81964170-81964170 on Build GRCh38
rs754742897 Chr.7: 81964380-81964380 on Build GRCh38
rs989037031 Chr.7: 81964505-81964505 on Build GRCh38
rs377181609 Chr.7: 81964096-81964096 on Build GRCh38
rs202188055 Chr.7: 81964149-81964149 on Build GRCh38

View Full Product Details