Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87163405-87163901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMTF1
Entrez Gene ID
9988
Gene Name
cyclin D binding myb like transcription factor 1
Gene Aliases
DMP1, DMTF, MRUL, hDMP1
UniGene
Hs.196129
Gene Chromosome Location
Chr.7: 87152361-87196332 on Build GRCh38
Transcripts
NM_001142326.1 NM_001142327.1 NM_021145.3 NR_024549.1 NR_024550.1 XM_011516735.2 XM_011516737.1 XM_011516739.2 XM_017012865.1 XM_017012866.1 XM_017012867.1 XM_017012868.1 XM_017012869.1 XM_017012870.1 XM_017012871.1 XM_017012872.1 XM_017012873.1 XM_017012874.1 XM_017012875.1 XM_017012876.1 XM_017012877.1 XM_017012878.1

SNP Detail

SNP ID Chromosome Location
rs987479445 Chr.7: 87163438-87163438 on Build GRCh38
rs566215534 Chr.7: 87163808-87163808 on Build GRCh38
rs1053792618 Chr.7: 87163503-87163503 on Build GRCh38
rs913470423 Chr.7: 87163506-87163506 on Build GRCh38
rs543792575 Chr.7: 87163572-87163572 on Build GRCh38
rs866861849 Chr.7: 87163562-87163562 on Build GRCh38
rs1038088505 Chr.7: 87163608-87163608 on Build GRCh38
rs951652727 Chr.7: 87163434-87163434 on Build GRCh38
rs891028753 Chr.7: 87163703-87163703 on Build GRCh38
rs942436987 Chr.7: 87163624-87163624 on Build GRCh38
rs1045399885 Chr.7: 87163550-87163550 on Build GRCh38
rs974814788 Chr.7: 87163766-87163766 on Build GRCh38
rs964641378 Chr.7: 87163761-87163761 on Build GRCh38
rs184705669 Chr.7: 87163446-87163446 on Build GRCh38
rs998426904 Chr.7: 87163675-87163675 on Build GRCh38
rs935187344 Chr.7: 87163560-87163560 on Build GRCh38
rs1039359976 Chr.7: 87163636-87163636 on Build GRCh38
rs949302963 Chr.7: 87163505-87163505 on Build GRCh38
rs974762360 Chr.7: 87163458-87163458 on Build GRCh38
rs1053532719 Chr.7: 87163568-87163568 on Build GRCh38
rs923785130 Chr.7: 87163542-87163542 on Build GRCh38
rs761907656 Chr.7: 87163537-87163537 on Build GRCh38
rs548005897 Chr.7: 87163580-87163580 on Build GRCh38
rs900850527 Chr.7: 87163669-87163669 on Build GRCh38
rs768670693 Chr.7: 87163643-87163643 on Build GRCh38
rs964756412 Chr.7: 87163455-87163455 on Build GRCh38
rs887273656 Chr.7: 87163847-87163847 on Build GRCh38
rs115004336 Chr.7: 87163874-87163874 on Build GRCh38
rs898352460 Chr.7: 87163627-87163627 on Build GRCh38
rs577986983 Chr.7: 87163481-87163481 on Build GRCh38
rs539556677 Chr.7: 87163811-87163811 on Build GRCh38
rs1026013676 Chr.7: 87163696-87163696 on Build GRCh38
rs1018906604 Chr.7: 87163747-87163747 on Build GRCh38
rs950792373 Chr.7: 87163430-87163430 on Build GRCh38
rs559191428 Chr.7: 87163472-87163472 on Build GRCh38
rs926124923 Chr.7: 87163468-87163468 on Build GRCh38
rs936146964 Chr.7: 87163485-87163485 on Build GRCh38
rs937082188 Chr.7: 87163594-87163594 on Build GRCh38
rs1009345292 Chr.7: 87163708-87163708 on Build GRCh38
rs537024393 Chr.7: 87163876-87163876 on Build GRCh38
rs1026343052 Chr.7: 87163801-87163801 on Build GRCh38
rs949777358 Chr.7: 87163510-87163510 on Build GRCh38
rs957562602 Chr.7: 87163834-87163834 on Build GRCh38
rs993988159 Chr.7: 87163658-87163658 on Build GRCh38
rs186739661 Chr.7: 87163869-87163869 on Build GRCh38
rs540635938 Chr.7: 87163447-87163447 on Build GRCh38
rs774462688 Chr.7: 87163794-87163794 on Build GRCh38
rs36096650 Chr.7: 87163618-87163618 on Build GRCh38

View Full Product Details