Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87170904-87171411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMTF1
Entrez Gene ID
9988
Gene Name
cyclin D binding myb like transcription factor 1
Gene Aliases
DMP1, DMTF, MRUL, hDMP1
UniGene
Hs.196129
Gene Chromosome Location
Chr.7: 87152361-87196332 on Build GRCh38
Transcripts
NM_001142326.1 NM_001142327.1 NM_021145.3 NR_024549.1 NR_024550.1 XM_011516735.2 XM_011516737.1 XM_011516739.2 XM_017012865.1 XM_017012866.1 XM_017012867.1 XM_017012868.1 XM_017012869.1 XM_017012870.1 XM_017012871.1 XM_017012872.1 XM_017012873.1 XM_017012874.1 XM_017012875.1 XM_017012876.1 XM_017012877.1 XM_017012878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5778570 c.234T>A p.L78L Chr.7: 87170996-87170996 on Build GRCh38
5778569 c.234T>A p.L78L Chr.7: 87170996-87170996 on Build GRCh38
4162483 c.273A>G p.A91A Chr.7: 87171035-87171035 on Build GRCh38
5496156 c.315G>A p.V105V Chr.7: 87171077-87171077 on Build GRCh38
4162484 c.273A>G p.A91A Chr.7: 87171035-87171035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965683612 Chr.7: 87171269-87171269 on Build GRCh38
rs769586724 Chr.7: 87170960-87170960 on Build GRCh38
rs982579160 Chr.7: 87171204-87171204 on Build GRCh38
rs759631186 Chr.7: 87170958-87170960 on Build GRCh38
rs772937512 Chr.7: 87170965-87170965 on Build GRCh38
rs928667859 Chr.7: 87171045-87171045 on Build GRCh38
rs185358584 Chr.7: 87171142-87171142 on Build GRCh38
rs796986869 Chr.7: 87171349-87171349 on Build GRCh38
rs371999785 Chr.7: 87171113-87171113 on Build GRCh38
rs200542814 Chr.7: 87171094-87171094 on Build GRCh38
rs974757119 Chr.7: 87171026-87171026 on Build GRCh38
rs368198198 Chr.7: 87171131-87171131 on Build GRCh38
rs1010395674 Chr.7: 87171060-87171060 on Build GRCh38
rs569673003 Chr.7: 87170978-87170978 on Build GRCh38
rs931984567 Chr.7: 87171363-87171363 on Build GRCh38
rs779513352 Chr.7: 87171312-87171312 on Build GRCh38
rs200898825 Chr.7: 87171097-87171097 on Build GRCh38
rs117360089 Chr.7: 87171048-87171048 on Build GRCh38
rs745313472 Chr.7: 87171119-87171119 on Build GRCh38
rs775851936 Chr.7: 87171013-87171013 on Build GRCh38
rs770462236 Chr.7: 87171111-87171111 on Build GRCh38
rs995681230 Chr.7: 87171183-87171183 on Build GRCh38
rs1054975104 Chr.7: 87171369-87171369 on Build GRCh38
rs751871447 Chr.7: 87170946-87170946 on Build GRCh38
rs779083172 Chr.7: 87171179-87171188 on Build GRCh38
rs189279801 Chr.7: 87171317-87171317 on Build GRCh38
rs754126527 Chr.7: 87171032-87171032 on Build GRCh38
rs114136038 Chr.7: 87171008-87171008 on Build GRCh38
rs117318646 Chr.7: 87171104-87171104 on Build GRCh38
rs747021860 Chr.7: 87170954-87170954 on Build GRCh38
rs748948300 Chr.7: 87170966-87170966 on Build GRCh38
rs772578350 Chr.7: 87171120-87171120 on Build GRCh38
rs758498760 Chr.7: 87171160-87171160 on Build GRCh38
rs576082807 Chr.7: 87171182-87171182 on Build GRCh38
rs749022283 Chr.7: 87171109-87171109 on Build GRCh38
rs932073234 Chr.7: 87171046-87171046 on Build GRCh38
rs151141381 Chr.7: 87171057-87171057 on Build GRCh38
rs764002217 Chr.7: 87170983-87170984 on Build GRCh38
rs137914812 Chr.7: 87171274-87171275 on Build GRCh38
rs764300746 Chr.7: 87171027-87171027 on Build GRCh38
rs778851191 Chr.7: 87170951-87170951 on Build GRCh38
rs912358556 Chr.7: 87171268-87171268 on Build GRCh38
rs773944974 Chr.7: 87171116-87171116 on Build GRCh38
rs912472622 Chr.7: 87170971-87170971 on Build GRCh38
rs117653228 Chr.7: 87171124-87171124 on Build GRCh38
rs754967697 Chr.7: 87171069-87171069 on Build GRCh38
rs773987320 Chr.7: 87170988-87170989 on Build GRCh38
rs368708718 Chr.7: 87170997-87170997 on Build GRCh38
rs981608681 Chr.7: 87171037-87171037 on Build GRCh38
rs768453100 Chr.7: 87170956-87170956 on Build GRCh38
rs760968422 Chr.7: 87171014-87171014 on Build GRCh38
rs35662461 Chr.7: 87171275-87171276 on Build GRCh38
rs374553113 Chr.7: 87170957-87170957 on Build GRCh38
rs747905428 Chr.7: 87170958-87170958 on Build GRCh38
rs3747807 Chr.7: 87171035-87171035 on Build GRCh38
rs981577638 Chr.7: 87171336-87171337 on Build GRCh38
rs757961895 Chr.7: 87171196-87171196 on Build GRCh38
rs139522170 Chr.7: 87171250-87171250 on Build GRCh38
rs371775057 Chr.7: 87171237-87171237 on Build GRCh38
rs775695375 Chr.7: 87170938-87170938 on Build GRCh38
rs551363065 Chr.7: 87170977-87170977 on Build GRCh38
rs956091741 Chr.7: 87171382-87171382 on Build GRCh38
rs781219753 Chr.7: 87171092-87171092 on Build GRCh38
rs116680384 Chr.7: 87171191-87171191 on Build GRCh38
rs181048450 Chr.7: 87171034-87171034 on Build GRCh38
rs958700230 Chr.7: 87170941-87170941 on Build GRCh38
rs1023182756 Chr.7: 87171282-87171282 on Build GRCh38
rs775905048 Chr.7: 87171121-87171121 on Build GRCh38
rs541609135 Chr.7: 87171384-87171384 on Build GRCh38
rs74574229 Chr.7: 87171385-87171385 on Build GRCh38
rs767803355 Chr.7: 87171021-87171021 on Build GRCh38
rs1050894147 Chr.7: 87171133-87171133 on Build GRCh38

View Full Product Details