Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87174451-87174957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMTF1
Entrez Gene ID
9988
Gene Name
cyclin D binding myb like transcription factor 1
Gene Aliases
DMP1, DMTF, MRUL, hDMP1
UniGene
Hs.196129
Gene Chromosome Location
Chr.7: 87152361-87196332 on Build GRCh38
Transcripts
NM_001142326.1 NM_001142327.1 NM_021145.3 NR_024549.1 NR_024550.1 XM_011516735.2 XM_011516737.1 XM_011516739.2 XM_017012865.1 XM_017012866.1 XM_017012867.1 XM_017012868.1 XM_017012869.1 XM_017012870.1 XM_017012871.1 XM_017012872.1 XM_017012873.1 XM_017012874.1 XM_017012875.1 XM_017012876.1 XM_017012877.1 XM_017012878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3950891 c.512A>C p.Y171S Chr.7: 87174662-87174662 on Build GRCh38
7303487 c.519+9T>C p.? Chr.7: 87174678-87174678 on Build GRCh38
7303488 c.519+9T>C p.? Chr.7: 87174678-87174678 on Build GRCh38
7290813 c.492G>C p.L164F Chr.7: 87174642-87174642 on Build GRCh38
7290814 c.492G>C p.L164F Chr.7: 87174642-87174642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770707081 Chr.7: 87174731-87174731 on Build GRCh38
rs780078428 Chr.7: 87174803-87174803 on Build GRCh38
rs561097467 Chr.7: 87174778-87174778 on Build GRCh38
rs1052682817 Chr.7: 87174845-87174845 on Build GRCh38
rs999796661 Chr.7: 87174827-87174827 on Build GRCh38
rs1040217814 Chr.7: 87174867-87174867 on Build GRCh38
rs11976589 Chr.7: 87174790-87174790 on Build GRCh38
rs771241966 Chr.7: 87174681-87174681 on Build GRCh38
rs746481922 Chr.7: 87174612-87174612 on Build GRCh38
rs530773348 Chr.7: 87174761-87174761 on Build GRCh38
rs375005251 Chr.7: 87174716-87174716 on Build GRCh38
rs559006713 Chr.7: 87174658-87174658 on Build GRCh38
rs763311153 Chr.7: 87174678-87174678 on Build GRCh38
rs111706183 Chr.7: 87174608-87174608 on Build GRCh38
rs546263307 Chr.7: 87174850-87174850 on Build GRCh38
rs764166529 Chr.7: 87174502-87174502 on Build GRCh38
rs374767399 Chr.7: 87174566-87174567 on Build GRCh38
rs990061782 Chr.7: 87174833-87174833 on Build GRCh38
rs750097741 Chr.7: 87174560-87174560 on Build GRCh38
rs144397319 Chr.7: 87174678-87174679 on Build GRCh38
rs746142573 Chr.7: 87174560-87174561 on Build GRCh38
rs774597825 Chr.7: 87174695-87174695 on Build GRCh38
rs773607716 Chr.7: 87174667-87174667 on Build GRCh38
rs764094189 Chr.7: 87174712-87174712 on Build GRCh38
rs758023617 Chr.7: 87174569-87174569 on Build GRCh38
rs879048994 Chr.7: 87174718-87174718 on Build GRCh38
rs943169601 Chr.7: 87174864-87174864 on Build GRCh38
rs1025446081 Chr.7: 87174738-87174738 on Build GRCh38
rs1025996921 Chr.7: 87174520-87174523 on Build GRCh38
rs754092574 Chr.7: 87174926-87174926 on Build GRCh38
rs993539467 Chr.7: 87174494-87174494 on Build GRCh38
rs542460368 Chr.7: 87174777-87174777 on Build GRCh38
rs570941124 Chr.7: 87174912-87174912 on Build GRCh38
rs770114803 Chr.7: 87174659-87174659 on Build GRCh38
rs961942445 Chr.7: 87174593-87174593 on Build GRCh38
rs753782328 Chr.7: 87174714-87174714 on Build GRCh38
rs957273609 Chr.7: 87174795-87174795 on Build GRCh38
rs1013242541 Chr.7: 87174892-87174892 on Build GRCh38
rs769167320 Chr.7: 87174620-87174620 on Build GRCh38
rs11976564 Chr.7: 87174737-87174737 on Build GRCh38
rs911310053 Chr.7: 87174769-87174769 on Build GRCh38
rs896167171 Chr.7: 87174846-87174846 on Build GRCh38
rs545349795 Chr.7: 87174728-87174728 on Build GRCh38
rs903995877 Chr.7: 87174797-87174797 on Build GRCh38
rs966833146 Chr.7: 87174750-87174750 on Build GRCh38
rs759449691 Chr.7: 87174770-87174770 on Build GRCh38
rs375925236 Chr.7: 87174709-87174709 on Build GRCh38
rs189245835 Chr.7: 87174635-87174635 on Build GRCh38
rs929092567 Chr.7: 87174893-87174893 on Build GRCh38
rs909009875 Chr.7: 87174584-87174584 on Build GRCh38
rs1006506029 Chr.7: 87174699-87174699 on Build GRCh38
rs762793132 Chr.7: 87174599-87174599 on Build GRCh38
rs922988300 Chr.7: 87174882-87174882 on Build GRCh38
rs759628229 Chr.7: 87174698-87174698 on Build GRCh38

View Full Product Details