Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87359482-87360012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROT
Entrez Gene ID
54677
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
COT
UniGene
Hs.125039
Gene Chromosome Location
Chr.7: 87345635-87399796 on Build GRCh38
Transcripts
NM_001143935.1 NM_001243745.1 NM_021151.3 XM_011516337.2 XM_017012370.1

SNP Detail

SNP ID Chromosome Location
rs959728174 Chr.7: 87359532-87359532 on Build GRCh38
rs959778784 Chr.7: 87359818-87359818 on Build GRCh38
rs991159729 Chr.7: 87359550-87359550 on Build GRCh38
rs527638855 Chr.7: 87359718-87359718 on Build GRCh38
rs991130067 Chr.7: 87359827-87359827 on Build GRCh38
rs116979197 Chr.7: 87359731-87359731 on Build GRCh38
rs894232868 Chr.7: 87359534-87359534 on Build GRCh38
rs193133754 Chr.7: 87359775-87359775 on Build GRCh38
rs139491665 Chr.7: 87359835-87359835 on Build GRCh38
rs908367528 Chr.7: 87359957-87359957 on Build GRCh38
rs957585755 Chr.7: 87359656-87359656 on Build GRCh38
rs553995913 Chr.7: 87359849-87359849 on Build GRCh38
rs547705063 Chr.7: 87359505-87359505 on Build GRCh38
rs112050733 Chr.7: 87359753-87359753 on Build GRCh38
rs376007371 Chr.7: 87359653-87359653 on Build GRCh38
rs556413556 Chr.7: 87359786-87359786 on Build GRCh38
rs1012647029 Chr.7: 87359567-87359567 on Build GRCh38
rs908397363 Chr.7: 87359644-87359644 on Build GRCh38
rs915774457 Chr.7: 87359565-87359565 on Build GRCh38
rs568303379 Chr.7: 87359808-87359808 on Build GRCh38
rs889345260 Chr.7: 87359719-87359719 on Build GRCh38
rs941233060 Chr.7: 87359814-87359814 on Build GRCh38
rs770509465 Chr.7: 87359722-87359722 on Build GRCh38
rs984076941 Chr.7: 87359907-87359907 on Build GRCh38
rs998603755 Chr.7: 87359780-87359780 on Build GRCh38
rs113106254 Chr.7: 87359695-87359695 on Build GRCh38
rs948512180 Chr.7: 87359837-87359837 on Build GRCh38
rs1024417829 Chr.7: 87359580-87359580 on Build GRCh38
rs57676881 Chr.7: 87359847-87359847 on Build GRCh38
rs908472690 Chr.7: 87359811-87359811 on Build GRCh38
rs1036441817 Chr.7: 87359680-87359680 on Build GRCh38
rs952723227 Chr.7: 87359891-87359891 on Build GRCh38
rs802026 Chr.7: 87359723-87359723 on Build GRCh38
rs575225164 Chr.7: 87359802-87359802 on Build GRCh38
rs984329867 Chr.7: 87359639-87359639 on Build GRCh38
rs766205901 Chr.7: 87359710-87359710 on Build GRCh38
rs761870754 Chr.7: 87359603-87359603 on Build GRCh38
rs998486811 Chr.7: 87359915-87359915 on Build GRCh38
rs565990141 Chr.7: 87359650-87359650 on Build GRCh38
rs983416651 Chr.7: 87359740-87359740 on Build GRCh38
rs1035337674 Chr.7: 87359523-87359523 on Build GRCh38
rs538323786 Chr.7: 87359784-87359784 on Build GRCh38

View Full Product Details